Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01574181_m1
          See other DPP10 GE Assays ›
          Gene Symbol:
          DPP10
          Gene Name:
          dipeptidyl peptidase like 10
          Gene Aliases:
          DPL2, DPPY, DPRP-3, DPRP3
          Chromosome Location:
          Chr.2: 114442358 - 115845752 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01574181_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DPP10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004360.3 NP_001004360.2 12 - 13 1217 121
          NM_001178034.1 NP_001171505.1 12 - 13 1144 121
          NM_001178036.1 NP_001171507.1 13 - 14 1621 121
          NM_001178037.1 NP_001171508.1 12 - 13 1187 121
          NM_001321905.1 NP_001308834.1 13 - 14 1647 121
          NM_001321906.1 NP_001308835.1 13 - 14 1263 121
          NM_001321908.1 NP_001308837.1 12 - 13 1506 121
          NM_001321909.1 NP_001308838.1 11 - 12 1121 121
          NM_001321910.1 NP_001308839.1 13 - 14 1590 121
          NM_001321911.1 NP_001308840.1 12 - 13 1589 121
          NM_001321912.1 NP_001308841.1 13 - 14 1621 121
          NM_001321913.1 NP_001308842.1 11 - 12 1417 121
          NM_001321914.1 NP_001308843.1 12 - 13 1142 121
          NM_020868.4 NP_065919.2 12 - 13 1532 121
          XM_017004566.1 XP_016860055.1 11 - 12 1136 121
          GenBank mRNA AB040925.2 - 12 - 13 1099 121
          AK289967.1 - 12 - 13 1231 121
          AK296490.1 - 12 - 13 1219 121
          AK315147.1 - 12 - 13 1336 121
          AY172661.3 - 12 - 13 1111 121
          AY387785.1 - 12 - 13 1133 121
          BC030832.1 - 12 - 13 1568 4811771 121
          DQ857321.1 - 12 - 13 1099 121
          DQ857322.1 - 12 - 13 1123 121
          DQ857323.1 - 12 - 13 1090 121

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200009005.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02000738.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene DPP10

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          protein localization to plasma membrane
          regulation of potassium ion transmembrane transport
          positive regulation of protein localization to plasma membrane
          serine-type peptidase activity
          dipeptidyl-peptidase activity
          potassium channel regulator activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02011527.hg.1 ENST00000310323.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02011527.hg.1 ENST00000393147.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02011527.hg.1 ENST00000409163.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02011527.hg.1 ENST00000410059.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02011527.hg.1 ENST00000476155.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02011527.hg.1 NM_001004360.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02011527.hg.1 NM_001178034.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02011527.hg.1 NM_001178036.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02011527.hg.1 NM_001178037.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02011527.hg.1 NM_020868.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02011527.hg.1 TC02000738.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0200163084.hg.1 ENST00000310323.12 Ensembl
          Affymetrix Human Clariom D Assay PSR0200163084.hg.1 ENST00000393147.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0200163084.hg.1 ENST00000409163.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200163084.hg.1 ENST00000410059.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200163084.hg.1 NM_001004360.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200163084.hg.1 NM_001178034.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200163084.hg.1 NM_001178036.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200163084.hg.1 NM_001178037.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200163084.hg.1 NM_020868.4 Refseq
          Affymetrix Human Clariom D Assay PSR0200163084.hg.1 TC0200009005.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200163084.hg.1 ENST00000310323.12 Ensembl
          Affymetrix Human Clariom D Assay PSR0200163084.hg.1 ENST00000393147.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0200163084.hg.1 ENST00000409163.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200163084.hg.1 ENST00000410059.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0200163084.hg.1 NM_001004360.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200163084.hg.1 NM_001178034.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200163084.hg.1 NM_001178036.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200163084.hg.1 NM_001178037.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200163084.hg.1 NM_020868.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0