Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01574212_g1
          See other SLC17A7 GE Assays ›
          Gene Symbol:
          SLC17A7
          Gene Name:
          solute carrier family 17 member 7
          Gene Aliases:
          BNPI, VGLUT1
          Chromosome Location:
          Chr.19: 49429398 - 49441551 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01574212_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SLC17A7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020309.3 NP_064705.1 5 - 6 802 69
          GenBank mRNA AB032436.1 - 5 - 6 651 69
          AK057879.1 - 748 69
          AK091061.1 - 1 - 2 53 69
          AK095659.2 - 4 - 5 2081 69
          AK289622.1 - 2 - 3 210 69
          AK294226.1 - 5 - 6 608 69
          AK294405.1 - 5 - 6 651 69
          BC059379.1 - 5 - 6 777 5262415 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900011139.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19001715.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SLC17A7

          Gene Ontology Categories:

          Function(s) Process(es)

          neural retina development
          ion transport
          phosphate ion transport
          neurotransmitter transport
          brain development
          long-term memory
          glutamate secretion
          L-glutamate transmembrane transport
          synaptic transmission, glutamatergic
          phosphate ion transmembrane transport
          sodium ion transmembrane transport
          sequestering of neurotransmitter
          sodium-dependent phosphate transport
          L-glutamate import
          excitatory postsynaptic potential
          synaptic vesicle lumen acidification
          chloride transmembrane transport
          L-glutamate transmembrane transporter activity
          inorganic phosphate transmembrane transporter activity
          sodium:phosphate symporter activity
          extracellularly glutamate-gated chloride channel activity
          sodium:inorganic phosphate symporter activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR19023424.hg.1 TC19001715.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19023424.hg.1 ENST00000221485.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19023424.hg.1 ENST00000543531.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19023424.hg.1 NM_020309.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000596689.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000596689.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000596689.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000596689.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000596689.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000596689.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000598018.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000598018.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000598018.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000598018.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000598018.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000598018.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000598018.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000598018.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000598018.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000600672.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000600672.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000600672.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000600672.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000600672.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000600672.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000600672.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000600672.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000600672.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000600672.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000600672.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000600672.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 ENST00000600672.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900193260.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900193260.hg.1 ENST00000596689.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900193260.hg.1 ENST00000598018.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900193260.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900193260.hg.1 ENST00000600672.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900193260.hg.1 NM_020309.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900193260.hg.1 TC1900011139.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900193260.hg.1 ENST00000598018.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900193260.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1900193260.hg.1 ENST00000596689.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900193260.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900193260.hg.1 ENST00000600672.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900193260.hg.1 NM_020309.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000221485.7 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000596689.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000596689.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000596689.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000596689.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000596689.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000596689.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000598018.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000598018.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000598018.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000598018.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000598018.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000598018.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000598018.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000598018.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000598018.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000600601.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000600672.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000600672.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000600672.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000600672.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000600672.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000600672.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000600672.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000600672.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000600672.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000600672.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000600672.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000600672.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 ENST00000600672.5 Ensembl
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 NM_020309.3 Refseq
          Affymetrix Human Clariom D Assay TC1900011139.hg.1 NM_020309.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-ztd5s:80/100.66.134.6:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0