Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01574709_m1
          See other UBR4 GE Assays ›
          Gene Symbol:
          UBR4
          Gene Name:
          ubiquitin protein ligase E3 component n-recognin 4
          Gene Aliases:
          RBAF600, ZUBR1, p600
          Chromosome Location:
          Chr.1: 19074506 - 19210334 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01574709_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: UBR4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020765.2 NP_065816.2 88 - 89 13028 65
          XM_011541108.2 XP_011539410.1 90 - 91 13263 65
          XM_011541109.2 XP_011539411.1 90 - 91 13260 65
          XM_011541110.2 XP_011539412.1 90 - 91 13260 65
          XM_011541111.2 XP_011539413.1 90 - 91 13260 65
          XM_011541112.2 XP_011539414.1 90 - 91 13248 65
          XM_011541113.2 XP_011539415.1 90 - 91 13245 65
          XM_011541114.2 XP_011539416.1 90 - 91 13245 65
          XM_011541115.2 XP_011539417.1 90 - 91 13239 65
          XM_011541116.2 XP_011539418.1 89 - 90 13230 65
          XM_011541117.2 XP_011539419.1 89 - 90 13179 65
          XM_011541118.2 XP_011539420.1 89 - 90 13176 65
          XM_011541119.2 XP_011539421.1 90 - 91 13263 65
          XM_011541120.2 XP_011539422.1 89 - 90 13140 65
          XM_011541121.2 XP_011539423.1 88 - 89 13107 65
          XM_017000822.1 XP_016856311.1 90 - 91 13242 65
          XM_017000823.1 XP_016856312.1 89 - 90 13215 65
          XM_017000824.1 XP_016856313.1 89 - 90 13161 65
          XM_017000825.1 XP_016856314.1 88 - 89 13146 65
          XM_017000826.1 XP_016856315.1 89 - 90 13143 65
          XM_017000827.1 XP_016856316.1 88 - 89 13128 65
          XM_017000828.1 XP_016856317.1 88 - 89 13104 65
          XM_017000829.1 XP_016856318.1 88 - 89 13056 65
          XM_017000830.1 XP_016856319.1 87 - 88 13005 65
          GenBank mRNA AB007931.1 - 29 - 30 4304 65
          AF348492.1 - 88 - 89 13024 65
          AK022573.1 - 1 - 2 50 65
          AK023826.1 - 26 - 27 3859 65
          AK025149.1 - 5 - 6 546 65
          AK123857.1 - 3 - 4 629 65
          AK302623.1 - 1 - 2 109 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100013123.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01002293.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AK022573.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene UBR4

          Gene Ontology Categories:

          Function(s) Process(es)

          ubiquitin-dependent protein catabolic process
          viral process
          ubiquitin-protein transferase activity
          protein binding
          calmodulin binding
          zinc ion binding
          ligase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01035009.hg.1 ENST00000375217.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01035009.hg.1 ENST00000375224.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01035009.hg.1 ENST00000375226.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01035009.hg.1 ENST00000375254.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01035009.hg.1 ENST00000375267.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01035009.hg.1 ENST00000543981.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01035009.hg.1 NM_020765.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01035009.hg.1 TC01002293.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100196992.hg.1 ENST00000375224.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100196992.hg.1 ENST00000375254.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100196992.hg.1 NM_020765.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100196992.hg.1 TC0100013123.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100196992.hg.1 ENST00000375224.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100196992.hg.1 ENST00000375254.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100196992.hg.1 NM_020765.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 155235 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-f85dfd666-px2hc:80/100.66.130.62:80.
          git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0