Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL7A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000094.3 | NP_000085.1 | 40 - 41 | 4406 | 87 | |
XM_011533337.1 | XP_011531639.1 | 41 - 42 | 4441 | 87 | ||
XM_017005688.1 | XP_016861177.1 | 41 - 42 | 4441 | 87 | ||
XM_017005689.1 | XP_016861178.1 | 41 - 42 | 4441 | 87 | ||
XM_017005690.1 | XP_016861179.1 | 41 - 42 | 4441 | 87 | ||
XM_017005691.1 | XP_016861180.1 | 41 - 42 | 4441 | 87 | ||
XM_017005692.1 | XP_016861181.1 | 41 - 42 | 4441 | 87 | ||
GenBank mRNA | AB209645.1 | - | 6 - 7 | 322 | 87 | |
L02870.1 | - | 40 - 41 | 4518 | 87 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |