Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01579626_m1
          See other PDE4D GE Assays ›
          Gene Symbol:
          PDE4D
          Gene Name:
          phosphodiesterase 4D
          Gene Aliases:
          ACRDYS2, DPDE3, HSPDE4D, PDE43, PDE4DN2, STRK1
          Chromosome Location:
          Chr.5: 58969038 - 60488098 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01579626_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PDE4D
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001104631.1 NP_001098101.1 11 - 12 1722 118
          NM_001165899.1 NP_001159371.1 13 - 14 1613 118
          NM_001197218.1 NP_001184147.1 11 - 12 1461 118
          NM_001197219.1 NP_001184148.1 11 - 12 1239 118
          NM_001197220.1 NP_001184149.1 11 - 12 1877 118
          NM_001197221.1 NP_001184150.1 7 - 8 1197 118
          NM_001197222.1 NP_001184151.1 6 - 7 1283 118
          NM_001197223.1 NP_001184152.1 6 - 7 1073 118
          NM_006203.4 NP_006194.2 11 - 12 1265 118
          XM_005248537.3 XP_005248594.1 11 - 12 2419 118
          XM_005248538.4 XP_005248595.1 12 - 13 4320 118
          XM_011543469.1 XP_011541771.1 14 - 15 1702 118
          XM_011543470.2 XP_011541772.1 14 - 15 1626 118
          XM_011543471.1 XP_011541773.1 13 - 14 1579 118
          XM_011543473.1 XP_011541775.1 13 - 14 1527 118
          XM_011543477.2 XP_011541779.1 13 - 14 1529 118
          XM_017009565.1 XP_016865054.1 14 - 15 1739 118
          XM_017009566.1 XP_016865055.1 14 - 15 1629 118
          XM_017009567.1 XP_016865056.1 12 - 13 1367 118
          XM_017009568.1 XP_016865057.1 14 - 15 1613 118
          XM_017009569.1 XP_016865058.1 13 - 14 1492 118
          XM_017009570.1 XP_016865059.1 10 - 11 2312 118
          XM_017009571.1 XP_016865060.1 13 - 14 1535 118
          GenBank mRNA AF012073.1 - 11 - 12 1475 118
          AF012074.1 - 7 - 8 1211 118
          AF536975.1 - 6 - 7 880 118
          AF536976.1 - 13 - 14 1436 118
          AF536977.1 - 11 - 12 1239 118
          AJ250852.1 - 7 - 8 807 118
          AJ250854.1 - 12 - 13 1402 118
          AY245866.1 - 13 - 14 1578 118
          AY245867.1 - 11 - 12 1877 118
          AY388960.1 - 11 - 12 1160 118
          BC036319.1 - 6 - 7 1075 4828512 118
          L20970.1 - 11 - 12 1273 118
          U02882.1 - 10 - 11 1043 118
          U50157.1 - 6 - 7 881 118
          U50158.1 - 7 - 8 795 118
          U50159.1 - 11 - 12 1216 118

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PDE4D

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of heart rate
          cAMP catabolic process
          smooth muscle contraction
          aging
          regulation of signaling receptor activity
          regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
          cAMP-mediated signaling
          neutrophil chemotaxis
          positive regulation of interferon-gamma production
          positive regulation of interleukin-2 production
          positive regulation of interleukin-5 production
          negative regulation of peptidyl-serine phosphorylation
          multicellular organism growth
          negative regulation of heart contraction
          T cell receptor signaling pathway
          regulation of ryanodine-sensitive calcium-release channel activity
          establishment of endothelial barrier
          cellular response to lipopolysaccharide
          cellular response to epinephrine stimulus
          adrenergic receptor signaling pathway
          regulation of cardiac muscle cell contraction
          adenylate cyclase-activating adrenergic receptor signaling pathway involved in positive regulation of heart rate
          regulation of cell communication by electrical coupling involved in cardiac conduction
          negative regulation of relaxation of cardiac muscle
          3',5'-cyclic-nucleotide phosphodiesterase activity
          3',5'-cyclic-AMP phosphodiesterase activity
          protein binding
          enzyme binding
          cAMP binding
          beta-2 adrenergic receptor binding
          transmembrane transporter binding
          metal ion binding
          ATPase binding
          scaffold protein binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0