Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01585093_s1
          See other TPBGL GE Assays ›
          Gene Symbol:
          TPBGL
          Gene Name:
          trophoblast glycoprotein-like
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 75240905 - 75243704 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01585093_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TPBGL
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001195528.1 NP_001182457.1 1 2738 145
          GenBank mRNA BC007804.2 - 1 1448 145
          N54218.1 - 1 164 145
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AW083054.1 - 1 Chr. 11 - 75243302 - 75243704 [-] on Build GRCh38 145
          GenBankmRNA AW294830.1 - 1 Chr. 11 - 75243469 - 75243699 [-] on Build GRCh38 145
          GenBankmRNA AW592098.1 - 1 Chr. 11 - 75243399 - 75243705 [-] on Build GRCh38 145
          GenBankmRNA BP395974.1 - 1 302 Chr. 11 - 75243342 - 75243683 [+] on Build GRCh38 145
          GenBankmRNA AW183461.1 - 1 Chr. 11 - 75243479 - 75243704 [-] on Build GRCh38 145
          GenBankmRNA N51092.1 - 1 Chr. 11 - 75243431 - 75243703 [-] on Build GRCh38 144
          GenBankmRNA BM675893.1 - 1 Chr. 11 - 75243128 - 75243699 [-] on Build GRCh38 145

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream solute carrier organic anion transporter family member 2B1 SLCO2B1 OATP-B,
          OATP2B1,
          OATPB,
          SLC21A9
          -34.5kb Sense Chr. 11 - 75150987 - 75206400 on NCBI Build GRCh38
          Downstream arrestin beta 1 ARRB1 ARB1,
          ARR1
          16.4kb Anti-sense Chr. 11 - 75260122 - 75351831 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100008454.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11000803.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          Roundabout signaling pathway
          axon extension involved in axon guidance
          regulation of axonogenesis
          heparin binding
          Roundabout binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879868 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879870 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879788 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28298240 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879852 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879814 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28298298 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879824 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16474336 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879795 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28298274 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879813 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879857 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879822 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28298223 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16474333 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879875 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28298297 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16474346 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16474331 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879874 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879850 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16474337 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879837 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879811 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28298253 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16474345 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28298285 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879828 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16474332 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879821 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879831 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28298238 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16474329 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28298201 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879842 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28298270 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879834 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28298216 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28298259 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879861 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879816 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28298277 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879879 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28298182 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879847 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879791 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879854 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879867 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28298268 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28298196 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879864 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879872 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879838 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879809 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28298188 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16474344 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28298226 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879803 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28298273 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879819 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28298219 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879815 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28298200 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879793 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879797 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879853 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879876 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879851 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879843 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879812 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879840 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28298271 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879817 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16474348 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879787 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07879786 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 444777 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3423220 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02470936 - -
          Agilent Human 1B Microarray G4111A A_32_P528354 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1100011641.hg.1 ENST00000529491.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100011641.hg.1 ENST00000529491.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100011641.hg.1 ENST00000529491.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100011641.hg.1 ENST00000529491.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100011641.hg.1 ENST00000530792.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100011641.hg.1 ENST00000530792.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100011641.hg.1 ENST00000530792.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100011641.hg.1 ENST00000530792.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100011641.hg.1 ENST00000530792.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100011641.hg.1 ENST00000530792.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100011641.hg.1 ENST00000603012.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100011641.hg.1 ENST00000603012.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100011641.hg.1 ENST00000603012.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100011641.hg.1 ENST00000603012.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008454.hg.1 ENST00000562197.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008454.hg.1 ENST00000562197.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008454.hg.1 ENST00000562197.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008454.hg.1 ENST00000562197.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100008454.hg.1 NM_001195528.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008454.hg.1 NM_001195528.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008454.hg.1 NM_001195528.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100008454.hg.1 NM_001195528.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11010287.hg.1 NM_001195528.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11010287.hg.1 TC11000803.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1100162581.hg.1 ENST00000562197.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100162581.hg.1 NM_001195528.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100162581.hg.1 TC1100008454.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100162581.hg.1 NM_001195528.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100162581.hg.1 ENST00000562197.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008454.hg.1 ENST00000562197.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008454.hg.1 ENST00000562197.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008454.hg.1 ENST00000562197.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008454.hg.1 ENST00000562197.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100008454.hg.1 NM_001195528.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008454.hg.1 NM_001195528.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008454.hg.1 NM_001195528.1 Refseq
          Affymetrix Human Clariom D Assay TC1100008454.hg.1 NM_001195528.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100162579.hg.1 ENST00000562197.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100162579.hg.1 NM_001195528.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100162579.hg.1 TC1100008454.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100162579.hg.1 ENST00000562197.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100162579.hg.1 NM_001195528.1 Refseq
          Affymetrix Human Clariom D Assay TC1100011641.hg.1 ENST00000529491.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100011641.hg.1 ENST00000529491.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100011641.hg.1 ENST00000529491.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100011641.hg.1 ENST00000529491.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100011641.hg.1 ENST00000530792.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100011641.hg.1 ENST00000530792.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100011641.hg.1 ENST00000530792.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100011641.hg.1 ENST00000530792.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100011641.hg.1 ENST00000530792.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100011641.hg.1 ENST00000530792.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100011641.hg.1 ENST00000603012.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100011641.hg.1 ENST00000603012.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100011641.hg.1 ENST00000603012.1 Ensembl
          Affymetrix Human Clariom D Assay TC1100011641.hg.1 ENST00000603012.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1100162580.hg.1 ENST00000562197.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100162580.hg.1 NM_001195528.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100162580.hg.1 TC1100008454.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100162580.hg.1 ENST00000562197.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100162580.hg.1 NM_001195528.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100162578.hg.1 ENST00000562197.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100162578.hg.1 NM_001195528.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100162578.hg.1 TC1100008454.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100162578.hg.1 ENST00000562197.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100162578.hg.1 NM_001195528.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046766 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046766 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 194263 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0