Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01586550_s1
          See other LOC100131510 GE Assays ›
          Gene Symbol:
          LOC100131510
          Gene Name:
          uncharacterized LOC100131510
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01586550_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100131510
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AF052121.1 - 1 1372 154
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA F10927.1 - Chr. - - [-] on Build 154
          GenBankmRNA AI631015.1 - 1 Chr. 2 - 25369136 - 25369634 [+] on Build GRCh38 154

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream DNA methyltransferase 3 alpha DNMT3A DNMT3A2,
          M.HsaIIIA,
          TBRS
          -26.5kb Anti-sense Chr. 2 - 25232961 - 25342590 on NCBI Build GRCh38
          Downstream dystrobrevin beta DTNB 7.6kb Anti-sense Chr. 2 - 25377220 - 25673647 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01055055 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15226825 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01055093 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01055066 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01055079 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01055052 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01055094 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21134022 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01055084 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01055096 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01055064 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21133997 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21134000 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01055069 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01055054 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01055083 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01055087 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21134029 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21133987 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21134011 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21134001 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01055063 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01055078 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01055057 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01055090 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01055062 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21133994 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21134015 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21134013 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01055059 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01055082 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21133978 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15226830 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01055099 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01055075 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01055060 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15226824 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01055091 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15226822 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 36341 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 227081 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 8554 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 15525 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 9415 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P00335328 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P323754 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P00335328 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00335328 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P00335328 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8050845 - -
          Affymetrix Human Gene 1.1 ST Array 8050844 - -
          Affymetrix HT HG-U133+ PM Array Plate 215421_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0200012008.hg.1 ENST00000313031.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012008.hg.1 ENST00000313031.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012008.hg.1 ENST00000313031.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012008.hg.1 ENST00000313031.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012008.hg.1 ENST00000313031.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012008.hg.1 ENST00000431557.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012008.hg.1 ENST00000431557.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012008.hg.1 ENST00000313031.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200012008.hg.1 ENST00000313031.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200012008.hg.1 ENST00000313031.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200012008.hg.1 ENST00000313031.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200012008.hg.1 ENST00000313031.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200012008.hg.1 ENST00000431557.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012008.hg.1 ENST00000431557.1 Ensembl
          Human Genome U133 Plus 2.0 Array 215421_at - -
          Human Genome U133 Plus 2.0 Array 215421_PM_at - -
          Human Genome U133A 2.0 Array 215421_PM_at - -
          Human Genome U133A 2.0 Array 215421_at - -
          Human Genome U95 Set 36752_PM_at - -
          Human Genome U95 Set 36752_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215421_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215421_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215421_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215421_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215421_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215421_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215421_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215421_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215421_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215421_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 215421_at2 - -
          Human Genome U133 Set 215421_PM_at - -
          Human Genome U133 Set 215421_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 231014 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3292114 AF052121.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc