Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01587574_m1
          See other CLASP2 GE Assays ›
          Gene Symbol:
          CLASP2
          Gene Name:
          cytoplasmic linker associated protein 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 33496245 - 33718356 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01587574_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CLASP2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_015097.2 NP_055912.2 23 - 24 2562 127
          XM_006713048.1 XP_006713111.1 23 - 24 2705 127
          XM_006713050.1 XP_006713113.1 23 - 24 2705 127
          XM_006713052.1 XP_006713115.1 23 - 24 2702 127
          XM_006713053.1 XP_006713116.1 23 - 24 2705 127
          XM_017005954.1 XP_016861443.1 23 - 24 2702 127
          XM_017005957.1 XP_016861446.1 23 - 24 2705 127
          XM_017005958.1 XP_016861447.1 23 - 24 2705 127
          XM_017005959.1 XP_016861448.1 23 - 24 2702 127
          XM_017005960.1 XP_016861449.1 21 - 22 2663 127
          XM_017005961.1 XP_016861450.1 21 - 22 2506 127
          XM_017005962.1 XP_016861451.1 23 - 24 2702 127
          XM_017005963.1 XP_016861452.1 23 - 24 2705 127
          XM_017005964.1 XP_016861453.1 23 - 24 2702 127
          XM_017005965.1 XP_016861454.1 21 - 22 2663 127
          XM_017005966.1 XP_016861455.1 21 - 22 2663 127
          XM_017005967.1 XP_016861456.1 21 - 22 2506 127
          XM_017005968.1 XP_016861457.1 17 - 18 1845 127
          XM_017005969.1 XP_016861458.1 17 - 18 1855 127
          XM_017005970.1 XP_016861459.1 17 - 18 2104 127
          XM_017005971.1 XP_016861460.1 17 - 18 1855 127
          XM_017005972.1 XP_016861461.1 17 - 18 2432 127
          XM_017005973.1 XP_016861462.1 17 - 18 2101 127
          XM_017005974.1 XP_016861463.1 17 - 18 2101 127
          XM_017005975.1 XP_016861464.1 17 - 18 1845 127
          XM_017005976.1 XP_016861465.1 15 - 16 1813 127
          XM_017005977.1 XP_016861466.1 17 - 18 2104 127
          XM_017005978.1 XP_016861467.1 17 - 18 1845 127
          XM_017005979.1 XP_016861468.1 17 - 18 1855 127
          XM_017005980.1 XP_016861469.1 17 - 18 2432 127
          XM_017005981.1 XP_016861470.1 17 - 18 1845 127
          XM_017005982.1 XP_016861471.1 17 - 18 1855 127
          XM_017005983.1 XP_016861472.1 17 - 18 2104 127
          XM_017005984.1 XP_016861473.1 17 - 18 2101 127
          XM_017005985.1 XP_016861474.1 17 - 18 2432 127
          XM_017005986.1 XP_016861475.1 15 - 16 2062 127
          GenBank mRNA AK297322.1 - 18 - 19 1818 127
          BC011530.1 - 4 - 5 401 127
          BC140778.1 - 23 - 24 2398 9021646 127

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CLASP2

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          vesicle targeting
          microtubule nucleation
          negative regulation of microtubule depolymerization
          Golgi organization
          mitotic spindle organization
          sister chromatid cohesion
          establishment or maintenance of cell polarity
          exit from mitosis
          regulation of gastrulation
          positive regulation of epithelial cell migration
          regulation of epithelial to mesenchymal transition
          microtubule organizing center organization
          regulation of microtubule polymerization or depolymerization
          regulation of microtubule-based process
          microtubule anchoring
          positive regulation of exocytosis
          cell division
          negative regulation of stress fiber assembly
          negative regulation of focal adhesion assembly
          protein localization to plasma membrane
          positive regulation of extracellular matrix disassembly
          negative regulation of wound healing, spreading of epidermal cells
          positive regulation of basement membrane assembly involved in embryonic body morphogenesis
          dystroglycan binding
          protein binding
          microtubule binding
          microtubule plus-end binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0