Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPAMD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015692.2 | NP_056507.2 | 3 - 4 | 443 | 136 | |
XM_011527918.1 | XP_011526220.1 | 3 - 4 | 446 | 136 | ||
XM_011527919.1 | XP_011526221.1 | 3 - 4 | 446 | 136 | ||
XM_011527920.2 | XP_011526222.1 | 3 - 4 | 446 | 136 | ||
XM_011527921.2 | XP_011526223.1 | 3 - 4 | 446 | 136 | ||
XM_011527922.1 | XP_011526224.1 | 3 - 4 | 446 | 136 | ||
XM_011527923.1 | XP_011526225.1 | 3 - 4 | 446 | 136 | ||
XM_011527924.2 | XP_011526226.1 | 3 - 4 | 446 | 136 | ||
XM_017026594.1 | XP_016882083.1 | 3 - 4 | 446 | 136 | ||
GenBank mRNA | AB033109.1 | - | 3 - 4 | 268 | 136 | |
AY101765.1 | - | 3 - 4 | 287 | 136 | ||
BC067854.1 | - | 3 - 4 | 443 | 136 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |