Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01592327_m1
          See other ITGB1BP1 GE Assays ›
          Gene Symbol:
          ITGB1BP1
          Gene Name:
          integrin subunit beta 1 binding protein 1
          Gene Aliases:
          ICAP-1A, ICAP-1B, ICAP-1alpha, ICAP1, ICAP1A, ICAP1B
          Chromosome Location:
          Chr.2: 9405681 - 9423569 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01592327_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ITGB1BP1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_134951.1 1 - 2 178 179
          XM_005246183.4 XP_005246240.1 1 - 2 768 179
          XM_005246189.4 XP_005246246.1 1 - 2 733 179
          XM_017005267.1 XP_016860756.1 1 - 2 725 179
          XM_017005270.1 XP_016860759.1 1 - 2 734 179

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the XM_005246183.4, XM_005246189.4, XM_017005267.1, XM_017005270.1 transcripts.

          Set Membership:

          Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel endothelial cell proliferation involved in sprouting angiogenesis
          transcription, DNA-templated
          negative regulation of protein kinase activity
          negative regulation of cell adhesion involved in substrate-bound cell migration
          cell-matrix adhesion
          Notch signaling pathway
          integrin-mediated signaling pathway
          positive regulation of cell population proliferation
          negative regulation of cell population proliferation
          positive regulation of endothelial cell migration
          negative regulation of fibroblast migration
          protein transport
          cell migration
          cell differentiation
          biomineral tissue development
          negative regulation of protein binding
          activation of protein kinase B activity
          integrin activation
          regulation of cell adhesion mediated by integrin
          tube formation
          intracellular signal transduction
          cellular response to vascular endothelial growth factor stimulus
          regulation of GTPase activity
          receptor clustering
          cellular response to fibroblast growth factor stimulus
          positive regulation of Notch signaling pathway
          positive regulation of transcription by RNA polymerase II
          myoblast migration
          positive regulation of stress fiber assembly
          positive regulation of cell division
          positive regulation of focal adhesion assembly
          negative regulation of focal adhesion assembly
          positive regulation of protein kinase B signaling
          negative regulation of ERK1 and ERK2 cascade
          negative regulation of cell migration involved in sprouting angiogenesis
          positive regulation of protein targeting to membrane
          negative regulation of protein targeting to membrane
          blood vessel diameter maintenance
          negative regulation of substrate adhesion-dependent cell spreading
          regulation of integrin-mediated signaling pathway
          GDP-dissociation inhibitor activity
          integrin binding
          protein binding
          protein kinase binding
          protein-containing complex binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0