Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01594967_m1
          See other SP7 GE Assays ›
          Gene Symbol:
          SP7
          Gene Name:
          Sp7 transcription factor
          Gene Aliases:
          OI11, OI12, OSX, osterix
          Chromosome Location:
          Chr.12: 53326575 - 53344793 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01594967_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SP7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001300837.1 NP_001287766.1 2 - 3 349 127
          XM_011537900.2 XP_011536202.1 2 - 3 400 127
          GenBank mRNA AY150674.1 - 1 - 2 80 127
          HY020346.1 - 2 - 3 350 127

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200010799.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the AY150674.1, NM_001300837.1, XM_011537900.2 transcripts.

          Set Membership:

          Amplicon length greater than or equal to 101 Homo sapiens Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          positive regulation of transcription by RNA polymerase II
          hematopoietic stem cell differentiation
          positive regulation of stem cell differentiation
          DNA binding
          DEAD/H-box RNA helicase binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 ENST00000303846.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 ENST00000303846.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 ENST00000303846.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 ENST00000303846.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 ENST00000303846.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 ENST00000303846.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 ENST00000303846.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 ENST00000536324.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 ENST00000536324.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 ENST00000536324.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 ENST00000536324.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 ENST00000536324.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 ENST00000536324.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 ENST00000536324.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 ENST00000536324.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 ENST00000537210.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 ENST00000537210.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 ENST00000537210.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 ENST00000537210.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 ENST00000547755.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 ENST00000547755.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 ENST00000547755.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 NM_001173467.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 NM_001173467.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 NM_001173467.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 NM_001173467.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 NM_001173467.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 NM_001173467.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 NM_001173467.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 NM_001173467.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 NM_152860.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 NM_152860.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 NM_152860.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 NM_152860.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 NM_152860.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 NM_152860.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010799.hg.1 NM_152860.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200182680.hg.1 ENST00000536324.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182680.hg.1 ENST00000537210.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182680.hg.1 NM_001173467.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200182680.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200182680.hg.1 TC1200010799.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200182680.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200182680.hg.1 ENST00000536324.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182680.hg.1 ENST00000537210.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182680.hg.1 NM_001173467.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200182670.hg.1 ENST00000303846.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182670.hg.1 ENST00000536324.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182670.hg.1 NM_001173467.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200182670.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200182670.hg.1 NM_152860.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200182670.hg.1 TC1200010799.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200182670.hg.1 ENST00000303846.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182670.hg.1 ENST00000536324.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182670.hg.1 NM_001173467.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200182670.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200182670.hg.1 NM_152860.1 Refseq
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 ENST00000303846.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 ENST00000303846.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 ENST00000303846.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 ENST00000303846.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 ENST00000303846.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 ENST00000303846.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 ENST00000303846.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 ENST00000536324.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 ENST00000536324.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 ENST00000536324.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 ENST00000536324.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 ENST00000536324.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 ENST00000536324.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 ENST00000536324.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 ENST00000536324.3 Ensembl
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 ENST00000537210.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 ENST00000537210.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 ENST00000537210.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 ENST00000537210.2 Ensembl
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 ENST00000547755.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 ENST00000547755.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 ENST00000547755.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 NM_001173467.2 Refseq
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 NM_001173467.2 Refseq
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 NM_001173467.2 Refseq
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 NM_001173467.2 Refseq
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 NM_001173467.2 Refseq
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 NM_001173467.2 Refseq
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 NM_001173467.2 Refseq
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 NM_001173467.2 Refseq
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 NM_152860.1 Refseq
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 NM_152860.1 Refseq
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 NM_152860.1 Refseq
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 NM_152860.1 Refseq
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 NM_152860.1 Refseq
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 NM_152860.1 Refseq
          Affymetrix Human Clariom D Assay TC1200010799.hg.1 NM_152860.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200182683.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200182683.hg.1 TC1200010799.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200182683.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200182673.hg.1 ENST00000303846.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182673.hg.1 ENST00000536324.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182673.hg.1 ENST00000537210.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182673.hg.1 ENST00000547755.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182673.hg.1 NM_001173467.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200182673.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200182673.hg.1 NM_152860.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200182673.hg.1 TC1200010799.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200182673.hg.1 ENST00000303846.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182673.hg.1 ENST00000536324.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182673.hg.1 ENST00000537210.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182673.hg.1 ENST00000547755.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182673.hg.1 NM_001173467.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200182673.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200182673.hg.1 NM_152860.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200182674.hg.1 ENST00000303846.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182674.hg.1 ENST00000536324.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182674.hg.1 ENST00000537210.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182674.hg.1 ENST00000547755.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182674.hg.1 NM_001173467.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200182674.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200182674.hg.1 NM_152860.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200182674.hg.1 TC1200010799.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200182674.hg.1 ENST00000303846.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182674.hg.1 ENST00000536324.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182674.hg.1 ENST00000537210.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182674.hg.1 ENST00000547755.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182674.hg.1 NM_001173467.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200182674.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200182674.hg.1 NM_152860.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200182681.hg.1 ENST00000536324.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182681.hg.1 NM_001173467.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200182681.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200182681.hg.1 TC1200010799.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200182681.hg.1 ENST00000536324.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182681.hg.1 NM_001173467.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200182681.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200182671.hg.1 ENST00000303846.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182671.hg.1 ENST00000536324.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182671.hg.1 NM_001173467.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200182671.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200182671.hg.1 NM_152860.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200182671.hg.1 TC1200010799.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200182671.hg.1 ENST00000303846.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182671.hg.1 ENST00000536324.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182671.hg.1 NM_001173467.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200182671.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200182671.hg.1 NM_152860.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200182672.hg.1 ENST00000303846.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182672.hg.1 ENST00000536324.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182672.hg.1 ENST00000537210.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182672.hg.1 NM_001173467.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200182672.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200182672.hg.1 NM_152860.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200182672.hg.1 TC1200010799.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200182672.hg.1 ENST00000303846.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182672.hg.1 ENST00000536324.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182672.hg.1 ENST00000537210.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1200182672.hg.1 NM_001173467.2 Refseq
          Affymetrix Human Clariom D Assay PSR1200182672.hg.1 NM_001300837.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200182672.hg.1 NM_152860.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d58dc5f8-fsfvr:80/100.66.134.75:80.
          git-commit: 38b8a393e477ae4d9d89cc5f7a29948d21ae29e2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc