Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01596056_m1
          See other LOC100652777 GE Assays ›
          Gene Symbol:
          LOC100652777 PLA2G10
          Gene Name:
          group 10 secretory phospholipase A2-like
          phospholipase A2 group X
          Gene Aliases:
          -
          GXPLA2, GXSPLA2, SPLA2, sPLA2-X
          Chromosome Location:
          Chr.16: 14709055 - 14733781 on Build GRCh38
          Chromosome Location:
          Chr.16: 14672548 - 14694663 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs01596056_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100652777
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_011522789.1 XP_011521091.1 1 - 2 533 116
          XM_011522790.2 XP_011521092.1 1 - 2 533 116
          Gene Symbol: PLA2G10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003561.2 NP_003552.1 1 - 2 443 116
          NR_133651.1 1 - 2 443 116
          XM_011522695.2 XP_011520997.1 1 - 2 533 116
          XM_011522696.2 XP_011520998.1 1 - 2 533 116
          XM_011522697.1 XP_011520999.1 1 - 2 533 116
          XM_011522698.1 XP_011521000.1 1 - 2 533 116
          GenBank mRNA BC069539.1 - 1 - 2 283 7262248 116
          BC106731.1 - 1 - 2 443 40015660 116
          BC106732.1 - 1 - 2 443 40015664 116
          BC111804.1 - 1 - 2 443 40069347 116
          BM739473.1 - 1 - 2 177 121
          CR456885.1 - 1 - 2 99 116
          U95301.1 - 1 - 2 539 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1600009486.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC16000885.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC100652777
          Click here to find siRNAs for Gene PLA2G10

          Gene Ontology Categories:

          Function(s) Process(es)

          phosphatidic acid biosynthetic process
          axon guidance
          positive regulation of macrophage derived foam cell differentiation
          positive regulation of lipid storage
          lipid catabolic process
          arachidonic acid metabolic process
          positive regulation of cellular protein metabolic process
          positive regulation of prostaglandin secretion
          phosphatidylglycerol acyl-chain remodeling
          phosphatidylinositol acyl-chain remodeling
          phosphatidylserine acyl-chain remodeling
          phosphatidylcholine acyl-chain remodeling
          phosphatidylethanolamine acyl-chain remodeling
          cholesterol homeostasis
          regulation of macrophage activation
          negative regulation of DNA-binding transcription factor activity
          lysophospholipid transport
          positive regulation of arachidonic acid secretion
          negative regulation of cholesterol efflux
          phospholipase activity
          phospholipase A2 activity
          calcium ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR16012682.hg.1 ENST00000438167.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16012682.hg.1 NM_003561.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16012682.hg.1 TC16000885.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1600009486.hg.1 ENST00000438167.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009486.hg.1 ENST00000438167.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009486.hg.1 ENST00000438167.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009486.hg.1 ENST00000438167.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009486.hg.1 ENST00000438167.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009486.hg.1 ENST00000438167.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009486.hg.1 ENST00000438167.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009486.hg.1 ENST00000438167.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009486.hg.1 ENST00000438167.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009486.hg.1 ENST00000438167.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009486.hg.1 ENST00000567462.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009486.hg.1 ENST00000567462.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009486.hg.1 ENST00000567462.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009486.hg.1 ENST00000567462.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009486.hg.1 ENST00000567462.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009486.hg.1 ENST00000567462.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009486.hg.1 NM_003561.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600009486.hg.1 NM_003561.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600009486.hg.1 NM_003561.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600009486.hg.1 NM_003561.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600009486.hg.1 NM_003561.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600009486.hg.1 NM_003561.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600009486.hg.1 NM_003561.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600009486.hg.1 NM_003561.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600009486.hg.1 NM_003561.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600009486.hg.1 NM_003561.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600009491.hg.1 ENST00000528546.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009491.hg.1 ENST00000528546.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16012684.hg.1 ENST00000438167.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16012684.hg.1 NM_003561.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16012684.hg.1 TC16000885.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1600009871.hg.1 ENST00000562217.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009871.hg.1 ENST00000562217.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009871.hg.1 ENST00000562217.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009564.hg.1 ENST00000564015.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009564.hg.1 ENST00000564015.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009648.hg.1 ENST00000491689.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009489.hg.1 ENST00000526173.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009489.hg.1 ENST00000526173.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009489.hg.1 ENST00000526173.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009489.hg.1 ENST00000526173.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007344.hg.1 ENST00000616703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007344.hg.1 ENST00000616703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16012683.hg.1 ENST00000438167.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16012683.hg.1 NM_003561.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16012683.hg.1 TC16000885.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1600007408.hg.1 ENST00000546408.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007408.hg.1 ENST00000546408.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007073.hg.1 ENST00000622756.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007073.hg.1 ENST00000622756.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009899.hg.1 ENST00000562618.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009899.hg.1 ENST00000562618.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009899.hg.1 ENST00000562618.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009899.hg.1 ENST00000562618.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009899.hg.1 ENST00000565800.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009899.hg.1 ENST00000565800.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009899.hg.1 ENST00000567688.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009899.hg.1 ENST00000567688.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009899.hg.1 ENST00000567688.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009877.hg.1 ENST00000619561.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600009877.hg.1 ENST00000619561.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007011.hg.1 ENST00000611162.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007011.hg.1 ENST00000611162.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007452.hg.1 ENST00000563429.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600007452.hg.1 ENST00000563429.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600172592.hg.1 ENST00000438167.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1600172592.hg.1 ENST00000567462.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600172592.hg.1 NM_003561.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600172592.hg.1 TC1600009486.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600172592.hg.1 NM_003561.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600172592.hg.1 ENST00000438167.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1600172592.hg.1 ENST00000567462.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600172593.hg.1 ENST00000438167.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1600172593.hg.1 NM_003561.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600172593.hg.1 TC1600009486.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600172593.hg.1 ENST00000438167.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1600172593.hg.1 NM_003561.2 Refseq
          Affymetrix Human Clariom D Assay TC1600009871.hg.1 ENST00000562217.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009871.hg.1 ENST00000562217.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009871.hg.1 ENST00000562217.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009877.hg.1 ENST00000619561.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009877.hg.1 ENST00000619561.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009489.hg.1 ENST00000526173.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009489.hg.1 ENST00000526173.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009489.hg.1 ENST00000526173.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009489.hg.1 ENST00000526173.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600007073.hg.1 ENST00000622756.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600007073.hg.1 ENST00000622756.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600172591.hg.1 ENST00000438167.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1600172591.hg.1 ENST00000567462.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600172591.hg.1 NM_003561.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600172591.hg.1 TC1600009486.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600172591.hg.1 ENST00000438167.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1600172591.hg.1 ENST00000567462.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600172591.hg.1 NM_003561.2 Refseq
          Affymetrix Human Clariom D Assay TC1600009899.hg.1 ENST00000562618.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009899.hg.1 ENST00000562618.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009899.hg.1 ENST00000562618.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009899.hg.1 ENST00000562618.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009899.hg.1 ENST00000565800.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009899.hg.1 ENST00000565800.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009899.hg.1 ENST00000567688.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009899.hg.1 ENST00000567688.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009899.hg.1 ENST00000567688.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600007408.hg.1 ENST00000546408.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600007408.hg.1 ENST00000546408.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600007344.hg.1 ENST00000616703.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600007344.hg.1 ENST00000616703.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600007452.hg.1 ENST00000563429.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600007452.hg.1 ENST00000563429.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600172594.hg.1 ENST00000438167.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1600172594.hg.1 NM_003561.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600172594.hg.1 TC1600009486.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600172594.hg.1 ENST00000438167.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1600172594.hg.1 NM_003561.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600172595.hg.1 ENST00000438167.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1600172595.hg.1 NM_003561.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600172595.hg.1 TC1600009486.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600172595.hg.1 ENST00000438167.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1600172595.hg.1 NM_003561.2 Refseq
          Affymetrix Human Clariom D Assay TC1600009491.hg.1 ENST00000528546.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009491.hg.1 ENST00000528546.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600007011.hg.1 ENST00000611162.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600007011.hg.1 ENST00000611162.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009486.hg.1 ENST00000438167.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600009486.hg.1 ENST00000438167.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600009486.hg.1 ENST00000438167.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600009486.hg.1 ENST00000438167.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600009486.hg.1 ENST00000438167.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600009486.hg.1 ENST00000438167.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600009486.hg.1 ENST00000438167.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600009486.hg.1 ENST00000438167.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600009486.hg.1 ENST00000438167.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600009486.hg.1 ENST00000438167.7 Ensembl
          Affymetrix Human Clariom D Assay TC1600009486.hg.1 ENST00000567462.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009486.hg.1 ENST00000567462.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009486.hg.1 ENST00000567462.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009486.hg.1 ENST00000567462.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009486.hg.1 ENST00000567462.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009486.hg.1 ENST00000567462.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009486.hg.1 NM_003561.2 Refseq
          Affymetrix Human Clariom D Assay TC1600009486.hg.1 NM_003561.2 Refseq
          Affymetrix Human Clariom D Assay TC1600009486.hg.1 NM_003561.2 Refseq
          Affymetrix Human Clariom D Assay TC1600009486.hg.1 NM_003561.2 Refseq
          Affymetrix Human Clariom D Assay TC1600009486.hg.1 NM_003561.2 Refseq
          Affymetrix Human Clariom D Assay TC1600009486.hg.1 NM_003561.2 Refseq
          Affymetrix Human Clariom D Assay TC1600009486.hg.1 NM_003561.2 Refseq
          Affymetrix Human Clariom D Assay TC1600009486.hg.1 NM_003561.2 Refseq
          Affymetrix Human Clariom D Assay TC1600009486.hg.1 NM_003561.2 Refseq
          Affymetrix Human Clariom D Assay TC1600009486.hg.1 NM_003561.2 Refseq
          Affymetrix Human Clariom D Assay TC1600009564.hg.1 ENST00000564015.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600009564.hg.1 ENST00000564015.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009648.hg.1 ENST00000491689.1 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0