Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01598309_m1
          See other PIN1 GE Assays ›
          Gene Symbol:
          PIN1
          Gene Name:
          peptidylprolyl cis/trans isomerase, NIMA-interacting 1
          Gene Aliases:
          DOD, UBL5
          Chromosome Location:
          Chr.19: 9835207 - 9849689 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01598309_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PIN1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_006221.3 NP_006212.1 3 - 4 519 148
          NR_038422.2 4 - 5 572 148
          NR_038830.1 4 - 5 572 148
          XM_011528068.2 XP_011526370.1 5 - 6 510 148
          GenBank mRNA AK092970.1 - 2 - 3 2531 148
          AK291074.1 - 3 - 4 403 148
          BC002899.1 - 3 - 4 408 3941595 148
          BC031971.1 - 4 - 5 1416 148
          BI827672.1 - 3 - 4 521 148
          BT019331.1 - 3 - 4 381 148
          CD675162.1 - 4 - 5 432 148
          CR407654.1 - 3 - 4 381 148
          U49070.1 - 3 - 4 405 148

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900006956.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19000165.hg.1

          Additional Information:

          This assay is in the 5'UTR region of the BC031971.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene PIN1

          Gene Ontology Categories:

          Function(s) Process(es)

          protein peptidyl-prolyl isomerization
          positive regulation of protein phosphorylation
          cell cycle
          regulation of mitotic nuclear division
          neuron differentiation
          negative regulation of transforming growth factor beta receptor signaling pathway
          negative regulation of protein binding
          regulation of cytokinesis
          negative regulation of type I interferon production
          positive regulation of protein dephosphorylation
          negative regulation of protein catabolic process
          negative regulation of neuron apoptotic process
          positive regulation of neuron apoptotic process
          positive regulation of GTPase activity
          positive regulation of transcription by RNA polymerase II
          synapse organization
          protein stabilization
          positive regulation of ubiquitin-protein transferase activity
          regulation of pathway-restricted SMAD protein phosphorylation
          positive regulation of cell growth involved in cardiac muscle cell development
          negative regulation of ERK1 and ERK2 cascade
          positive regulation of canonical Wnt signaling pathway
          regulation of protein localization to nucleus
          regulation of signal transduction by p53 class mediator
          negative regulation of cell motility
          peptidyl-prolyl cis-trans isomerase activity
          cytoskeletal motor activity
          protein binding
          beta-catenin binding
          mitogen-activated protein kinase kinase binding
          GTPase activating protein binding
          phosphoserine residue binding
          phosphothreonine residue binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR19002493.hg.1 ENST00000247970.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19002493.hg.1 ENST00000380889.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19002493.hg.1 ENST00000424497.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19002493.hg.1 NM_006221.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19002493.hg.1 NR_038422.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19002493.hg.1 NR_038830.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19002493.hg.1 TC19000165.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1900149335.hg.1 ENST00000247970.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1900149335.hg.1 ENST00000380889.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1900149335.hg.1 ENST00000585442.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900149335.hg.1 ENST00000586025.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900149335.hg.1 ENST00000586352.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900149335.hg.1 ENST00000588695.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900149335.hg.1 ENST00000589058.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900149335.hg.1 ENST00000590540.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900149335.hg.1 ENST00000591777.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900149335.hg.1 NM_006221.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900149335.hg.1 NR_038422.2 Refseq
          Affymetrix Human Clariom D Assay PSR1900149335.hg.1 NR_038830.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900149335.hg.1 TC1900006956.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900149335.hg.1 ENST00000586025.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900149335.hg.1 ENST00000247970.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1900149335.hg.1 ENST00000380889.6 Ensembl
          Affymetrix Human Clariom D Assay PSR1900149335.hg.1 ENST00000585442.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900149335.hg.1 ENST00000586352.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900149335.hg.1 ENST00000588695.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900149335.hg.1 ENST00000589058.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900149335.hg.1 ENST00000590540.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1900149335.hg.1 ENST00000591777.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900149335.hg.1 NM_006221.3 Refseq
          Affymetrix Human Clariom D Assay PSR1900149335.hg.1 NR_038422.2 Refseq
          Affymetrix Human Clariom D Assay PSR1900149335.hg.1 NR_038830.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0