Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01669495_m1
          See other HECW1-IT1 GE Assays ›
          Gene Symbol:
          HECW1-IT1
          Gene Name:
          HECW1 intronic transcript 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 43117896 - 43163187 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01669495_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HECW1-IT1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_135295.1 5 - 6 1473 112
          GenBank mRNA AK093262.1 - 5 - 6 1473 112

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3390924 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3390924 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3390924 - -
          Agilent Human 1B Microarray G4111A A_32_P918201 - -
          Agilent Human 1B Microarray G4111A A_32_P768323 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8132498 - -
          Affymetrix Human Gene 1.0 ST Array 8132496 - -
          Affymetrix Human Gene 1.0 ST Array 8132494 - -
          Affymetrix Human Gene 1.0 ST Array 8132495 - -
          Affymetrix Human Gene 1.0 ST Array 8132497 - -
          Affymetrix Human Gene 1.0 ST Array 8132500 - -
          Affymetrix Human Gene 1.0 ST Array 8132499 - -
          Affymetrix Human Gene 1.1 ST Array 8132493 - -
          Affymetrix HT HG-U133+ PM Array Plate 1560950_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0700007337.hg.1 ENST00000322220.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700007337.hg.1 ENST00000322220.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700007337.hg.1 ENST00000322220.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700007337.hg.1 ENST00000322220.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700007337.hg.1 ENST00000322220.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700007337.hg.1 ENST00000322220.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700007337.hg.1 ENST00000322220.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700007337.hg.1 ENST00000322220.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700007337.hg.1 ENST00000322220.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700007337.hg.1 ENST00000322220.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700007337.hg.1 ENST00000322220.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700007337.hg.1 ENST00000322220.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700007337.hg.1 ENST00000322220.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700007337.hg.1 ENST00000322220.1 Ensembl
          Human Genome U133 Plus 2.0 Array 1560950_PM_at - -
          Human Genome U133 Plus 2.0 Array 1560950_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560950_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560950_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560950_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560950_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560950_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560950_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560950_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560950_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560950_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560950_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560950_at6 - -
          Human Genome U95 Set 66933_at - -
          Human Genome U95 Set 66933_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina human-6 v2.0 expression beadchip ILMN_1854701 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 4210333 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_77055 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1854701 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1854701 - -
          Illumina HumanWG-6_V2_0_R2 ILMN_1854701 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0