Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01676670_g1
          See other POU5F1P4 GE Assays ›
          Gene Symbol:
          POU5F1P4 POU5F1B POU5F1P3
          Gene Name:
          POU class 5 homeobox 1 pseudogene 4
          POU class 5 homeobox 1B
          POU class 5 homeobox 1 pseudogene 3
          Gene Aliases:
          Oct4-pg4, POU5FLC1
          OCT4-PG1, OCT4PG1, OTF3C, OTF3P1, POU5F1P1, POU5F1P4, POU5FLC20, POU5FLC8
          OCT4-pg3, OTF3L, POU5F1L, POU5FLC12
          Chromosome Location:
          Chr.1: 155433180 - 155434262 on Build GRCh38
          Chromosome Location:
          Chr.8: 127415612 - 127417210 on Build GRCh38
          Chromosome Location:
          Chr.12: 8133769 - 8134852 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          Assay ID Hs01676670_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: POU5F1P4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_034180.1 1 624 98
          GenBank mRNA AF268613.1 - 1 624 98
          DQ851566.1 - 1 624 98
          GU480814.1 - 1 625 98
          GU480831.1 - 1 626 98
          GU480833.1 - 1 625 98
          GU480834.1 - 1 625 98
          GU480836.1 - 1 625 98
          GU480839.1 - 1 625 98
          GU480848.1 - 1 625 98
          GU480853.1 - 1 626 98
          GU480855.1 - 1 623 98
          GU480856.1 - 1 625 98
          GU480857.1 - 1 625 98
          GU480858.1 - 1 625 98
          GU480859.1 - 1 625 98
          GU480860.1 - 1 624 98
          GU480861.1 - 1 626 98
          GU480865.1 - 1 626 98
          GU480867.1 - 1 625 98
          GU480871.1 - 1 625 98
          GU480873.1 - 1 625 98
          GU480875.1 - 1 625 98
          GU480879.1 - 1 625 98
          GU480881.1 - 1 625 98
          GU480882.1 - 1 625 98
          GU480890.1 - 1 624 98
          GU480895.1 - 1 625 98
          GU480898.1 - 1 624 98
          GU480900.1 - 1 624 98
          GU480904.1 - 1 625 98
          HQ122676.1 - 1 625 98
          HQ122677.1 - 1 625 98
          HQ122678.1 - 1 623 98
          HQ122679.1 - 1 624 98
          HQ122680.1 - 1 625 98
          HQ122681.1 - 1 625 98
          HQ122682.1 - 1 625 98
          HQ122683.1 - 1 626 98
          HQ122684.1 - 1 625 98
          HQ122685.1 - 1 623 98
          HQ122686.1 - 1 625 98
          HQ122687.1 - 1 625 98
          HQ122689.1 - 1 623 98
          HQ122690.1 - 1 625 98
          HQ122691.1 - 1 625 98
          HQ122695.1 - 1 624 98
          HQ122696.1 - 1 624 98
          HQ122697.1 - 1 624 98
          HQ122698.1 - 1 625 98
          HQ122699.1 - 1 625 98
          HQ122700.1 - 1 625 98
          HQ122701.1 - 1 625 98
          HQ122702.1 - 1 625 98
          HQ122703.1 - 1 625 98
          HQ122704.1 - 1 625 98
          HQ122705.1 - 1 625 98
          HQ122706.1 - 1 625 98
          HQ122707.1 - 1 624 98
          HQ122708.1 - 1 625 98
          HQ122709.1 - 1 625 98
          HQ122731.1 - 1 625 98
          HQ122732.1 - 1 625 98
          HQ122733.1 - 1 625 98
          HQ122734.1 - 1 625 98
          HQ122735.1 - 1 625 98
          HQ122736.1 - 1 625 98
          HQ122740.1 - 1 625 98
          HQ122741.1 - 1 625 98
          HQ122742.1 - 1 625 98
          HQ122743.1 - 1 625 98
          HQ122744.1 - 1 625 98
          HQ122745.1 - 1 625 98
          HQ122746.1 - 1 625 98
          HQ122747.1 - 1 625 98
          HQ122748.1 - 1 625 98
          HQ122756.1 - 1 625 98
          HQ122757.1 - 1 625 98
          HQ122758.1 - 1 625 98
          HQ122760.1 - 1 625 98
          HQ122761.1 - 1 625 98
          HQ122762.1 - 1 625 98
          HQ122763.1 - 1 625 98
          HQ122764.1 - 1 625 98
          HQ122765.1 - 1 625 98
          HQ122766.1 - 1 625 98
          HQ122767.1 - 1 625 98
          HQ122768.1 - 1 625 98
          HQ122769.1 - 1 624 98
          HQ122771.1 - 1 625 98
          HQ122772.1 - 1 625 98
          HQ122774.1 - 1 625 98
          HQ122775.1 - 1 625 98
          HQ122777.1 - 1 624 98
          HQ122778.1 - 1 625 98
          HQ122779.1 - 1 625 98
          HQ122780.1 - 1 625 98
          HQ122798.1 - 1 625 98
          HQ122799.1 - 1 624 98
          HQ122801.1 - 1 625 98
          HQ122802.1 - 1 623 98
          HQ122803.1 - 1 626 98
          HQ122804.1 - 1 625 98
          HQ122805.1 - 1 625 98
          HQ122807.1 - 1 625 98
          HQ122808.1 - 1 625 98
          HQ122809.1 - 1 623 98
          HQ122810.1 - 1 625 98
          HQ122811.1 - 1 625 98
          HQ122812.1 - 1 625 98
          HQ122813.1 - 1 625 98
          HQ122814.1 - 1 623 98
          HQ122833.1 - 1 625 98
          HQ122835.1 - 1 623 98
          HQ122836.1 - 1 625 98
          HQ122838.1 - 1 623 98
          HQ122839.1 - 1 625 98
          HQ122840.1 - 1 625 98
          HQ122858.1 - 1 625 98
          HQ122859.1 - 1 625 98
          HQ122860.1 - 1 625 98
          HQ122861.1 - 1 625 98
          HQ122862.1 - 1 625 98
          HQ122904.1 - 1 625 98
          HQ122905.1 - 1 625 98
          HQ122906.1 - 1 626 98
          HQ122907.1 - 1 625 98
          HQ122909.1 - 1 626 98
          HQ122910.1 - 1 625 98
          HQ122911.1 - 1 625 98
          HQ122912.1 - 1 625 98
          HQ122913.1 - 1 625 98
          Z21963.1 - 1 63 98
          Gene Symbol: POU5F1B
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA GU480844.1 - 1 624 98
          Gene Symbol: POU5F1P3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA GU480891.1 - 1 623 98

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0