Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01686869_sH
          See other POU5F1P3 GE Assays ›
          Gene Symbol:
          POU5F1P3 POU5F1P4
          Gene Name:
          POU class 5 homeobox 1 pseudogene 3
          POU class 5 homeobox 1 pseudogene 4
          Gene Aliases:
          OCT4-pg3, OTF3L, POU5F1L, POU5FLC12
          Oct4-pg4, POU5FLC1
          Chromosome Location:
          Chr.12: 8133769 - 8134852 on Build GRCh38
          Chromosome Location:
          Chr.1: 155433180 - 155434262 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs01686869_sH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: POU5F1P3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_036440.1 1 153
          GenBank mRNA AF268617.1 - 1 153
          GU480832.1 - 1 153
          GU480837.1 - 1 153
          GU480838.1 - 1 153
          GU480843.1 - 1 153
          GU480846.1 - 1 153
          GU480847.1 - 1 153
          GU480876.1 - 1 153
          GU480877.1 - 1 153
          GU480887.1 - 1 153
          GU480891.1 - 1 153
          GU480892.1 - 1 153
          GU480893.1 - 1 153
          GU480897.1 - 1 153
          GU480903.1 - 1 153
          GU480907.1 - 1 153
          GU480909.1 - 1 153
          HQ122692.1 - 1 153
          HQ122693.1 - 1 153
          HQ122694.1 - 1 153
          HQ122722.1 - 1 153
          HQ122723.1 - 1 153
          HQ122724.1 - 1 153
          HQ122725.1 - 1 153
          HQ122726.1 - 1 153
          HQ122727.1 - 1 153
          HQ122728.1 - 1 153
          HQ122729.1 - 1 153
          HQ122730.1 - 1 153
          HQ122754.1 - 1 153
          HQ122755.1 - 1 153
          HQ122791.1 - 1 153
          HQ122792.1 - 1 114 153
          HQ122793.1 - 1 153
          HQ122794.1 - 1 153
          HQ122795.1 - 1 153
          HQ122796.1 - 1 153
          HQ122797.1 - 1 153
          HQ122828.1 - 1 153
          HQ122829.1 - 1 153
          HQ122830.1 - 1 153
          HQ122831.1 - 1 153
          Gene Symbol: POU5F1P4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA GU480900.1 - 1 153
          HQ122697.1 - 1 153

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200009810.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12001173.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR12015591.hg.1 NR_036440.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12015591.hg.1 TC12001173.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1200009810.hg.1 ENST00000428824.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200009810.hg.1 NR_036440.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200173736.hg.1 ENST00000428824.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1200173736.hg.1 NR_036440.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200173736.hg.1 TC1200009810.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200173736.hg.1 NR_036440.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200173736.hg.1 ENST00000428824.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200009810.hg.1 ENST00000428824.4 Ensembl
          Affymetrix Human Clariom D Assay TC1200009810.hg.1 NR_036440.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 193216 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0