Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01689249_s1
          See other SUMO1P3 GE Assays ›
          Gene Symbol:
          SUMO1P3
          Gene Name:
          SUMO1 pseudogene 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 160317265 - 160318474 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01689249_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SUMO1P3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002190.1 1 753 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100010294.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01001366.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P223189 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P223189 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 3150 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 24177 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3318465 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P223189 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P319354 - -
          SurePrint G3 Human GE Microarray G4851A A_32_P223189 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00545409 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00545438 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00545437 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00545435 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604077 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604089 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604094 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00545416 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15136382 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604059 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00545439 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604063 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00545426 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604070 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15136387 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604098 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604061 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00545419 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604078 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15136380 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604110 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604073 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00545442 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00545434 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604085 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604100 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00545429 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604074 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604065 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604067 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604087 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00545411 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15136387 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00545426 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604077 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604110 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604061 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604074 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604078 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00545419 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604098 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00545435 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604085 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15136380 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00545409 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604063 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00545411 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604100 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604070 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604059 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604094 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15136382 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604067 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00545434 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00545442 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604073 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604087 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00545416 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604089 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00545439 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00545429 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00545437 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00545438 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20604065 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P223189 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P223189 - -
          Agilent Human 1B Microarray G4111A A_32_P223189 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00545419 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00545436 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00545431 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00545426 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00545442 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00545410 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00545428 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00545413 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15136375 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00545411 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00545424 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00545422 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00545420 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00545433 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15136378 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00545435 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00545414 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00545402 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00545404 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00545444 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00545439 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00545417 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00545407 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15136373 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NR_002190_at - -
          Affymetrix Human Gene 1.0 ST Array 7906575 - -
          Affymetrix Human Gene 1.1 ST Array 7906574 - -
          Affymetrix Human Transcriptome Array 2.0 PSR01021414.hg.1 NR_002190.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01021414.hg.1 TC01001366.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100175546.hg.1 NR_002190.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100175546.hg.1 TC0100010294.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100175546.hg.1 NR_002190.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100175545.hg.1 ENST00000439961.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100175545.hg.1 NR_002190.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100175545.hg.1 TC0100010294.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100175545.hg.1 ENST00000439961.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100175545.hg.1 NR_002190.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100175544.hg.1 NR_002190.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100175544.hg.1 TC0100010294.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100175544.hg.1 NR_002190.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A033356 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A033358 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A033356 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A033358 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 105431 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1732080 NR_002190.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2172221 NR_002190.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2172221 AA021599.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1732080 NR_002190.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1732080 NR_002190.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2172221 NR_002190.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2172221 AA021599.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2172221 NR_002190.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2172221 AA021599.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2172221 NR_002190.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2172221 AA021599.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1732080 NR_002190.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 430010 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_16906 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2172221 NR_002190.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2172221 AA021599.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1732080 NR_002190.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2172221 NR_002190.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2172221 AA021599.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2172221 NR_002190.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2172221 AA021599.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1732080 NR_002190.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1732080 NR_002190.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2172221 NR_002190.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2172221 AA021599.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0