Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01894347_s1
          See other DIAPH2-AS1 GE Assays ›
          Gene Symbol:
          DIAPH2-AS1
          Gene Name:
          DIAPH2 antisense RNA 1
          Gene Aliases:
          EPAG
          Chromosome Location:
          Chr.X: 97528364 - 97564535 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01894347_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DIAPH2-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA L22650.1 - 1 106 90

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567096 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11838878 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11838871 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11838893 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567058 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11838896 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11838900 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567067 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567081 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11838892 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567073 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567093 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11838880 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567080 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567091 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567046 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567066 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567059 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11838851 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11838864 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11838932 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567034 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11838891 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567085 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11838934 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567072 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567071 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567055 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11838869 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567095 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567092 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11838904 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11838933 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567083 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567068 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11838881 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567076 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567029 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11838924 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567028 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567104 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11838866 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11838858 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567088 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567078 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567053 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11838877 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567087 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567045 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567035 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11838936 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11838870 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17378558 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11838938 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567075 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567036 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11838874 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567064 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567030 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11838928 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11838931 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11838901 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567106 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32567098 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11838872 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11838868 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 86160 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 692847 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 118100 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 698077 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 396465 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 34980 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03734026 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P33309 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 25173 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P21528214 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P21528207 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03734026 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P17237560 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P41735898 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P21528200 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P21528187 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P17237560 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P03734026 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P21528187 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8173932 - -
          Affymetrix Human Gene 1.0 ST Array 8173931 - -
          Affymetrix Human Gene 1.1 ST Array 8173930 - -
          Affymetrix HT HG-U133+ PM Array Plate 217245_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 217050_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0X00010310.hg.1 ENST00000439759.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010310.hg.1 ENST00000439759.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010310.hg.1 ENST00000439759.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010310.hg.1 ENST00000439759.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010310.hg.1 ENST00000439759.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010310.hg.1 ENST00000439759.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010310.hg.1 ENST00000439759.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010310.hg.1 ENST00000439759.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010310.hg.1 ENST00000439759.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010310.hg.1 ENST00000439759.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010310.hg.1 ENST00000439759.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010310.hg.1 ENST00000439759.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010310.hg.1 ENST00000445414.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010310.hg.1 ENST00000445414.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010310.hg.1 ENST00000445414.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010310.hg.1 ENST00000445414.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010310.hg.1 ENST00000542084.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010310.hg.1 ENST00000542084.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010310.hg.1 ENST00000542084.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010310.hg.1 ENST00000579945.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010310.hg.1 ENST00000579945.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010310.hg.1 ENST00000579945.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010310.hg.1 ENST00000579945.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00010310.hg.1 NR_125391.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010310.hg.1 NR_125391.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010310.hg.1 NR_125391.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00010310.hg.1 NR_125391.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00010310.hg.1 ENST00000439759.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010310.hg.1 ENST00000439759.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010310.hg.1 ENST00000439759.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010310.hg.1 ENST00000439759.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010310.hg.1 ENST00000439759.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010310.hg.1 ENST00000439759.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010310.hg.1 ENST00000439759.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010310.hg.1 ENST00000439759.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010310.hg.1 ENST00000439759.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010310.hg.1 ENST00000439759.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010310.hg.1 ENST00000439759.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010310.hg.1 ENST00000439759.2 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010310.hg.1 ENST00000445414.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010310.hg.1 ENST00000445414.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010310.hg.1 ENST00000445414.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010310.hg.1 ENST00000445414.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010310.hg.1 ENST00000542084.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010310.hg.1 ENST00000542084.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010310.hg.1 ENST00000542084.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010310.hg.1 ENST00000579945.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010310.hg.1 ENST00000579945.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010310.hg.1 ENST00000579945.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010310.hg.1 ENST00000579945.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00010310.hg.1 NR_125391.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00010310.hg.1 NR_125391.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00010310.hg.1 NR_125391.1 Refseq
          Affymetrix Human Clariom D Assay TC0X00010310.hg.1 NR_125391.1 Refseq
          Human Genome U133 Plus 2.0 Array 217246_PM_s_at - -
          Human Genome U133 Plus 2.0 Array 217245_at - -
          Human Genome U133 Plus 2.0 Array 217050_at - -
          Human Genome U133 Plus 2.0 Array 217246_s_at - -
          Human Genome U133 Plus 2.0 Array 217245_PM_at - -
          Human Genome U133 Plus 2.0 Array 217050_PM_at - -
          Human Genome U133A 2.0 Array 217050_PM_at - -
          Human Genome U133A 2.0 Array 217050_at - -
          Human Genome U133A 2.0 Array 217245_at - -
          Human Genome U133A 2.0 Array 217245_PM_at - -
          Human Genome U133A 2.0 Array 217246_s_at - -
          Human Genome U133A 2.0 Array 217246_PM_s_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) XM_938050_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) XM_926826_at - -
          AffymetrixPathwork Diagnostics Pathchip pw543077 - -
          AffymetrixPathwork Diagnostics Pathchip pw522755 - -
          AffymetrixPathwork Diagnostics Pathchip pw500645 - -
          Human Genome U95 Set 31388_at - -
          Human Genome U95 Set 31388_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217246_s_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217246_s_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217050_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217050_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217246_s_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217050_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217246_s_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217246_s_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217050_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217245_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217245_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217050_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217245_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217246_s_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217245_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217050_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217050_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217245_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217245_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217246_s_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217245_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217246_s_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217245_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217246_s_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217050_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217245_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217245_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217246_s_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217245_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217050_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217050_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217050_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 217246_s_at4 - -
          Human Genome U133 Set 217246_s_at - -
          Human Genome U133 Set 217050_at - -
          Human Genome U133 Set 217245_at - -
          Human Genome U133 Set 217246_PM_s_at - -
          Human Genome U133 Set 217245_PM_at - -
          Human Genome U133 Set 217050_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 191832 - -
          Array Name Probe IDs Targets Target Type
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1651309 Y15909.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 2190088 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_32433 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1651309 Y15909.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1651309 Y15909.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0