Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01903755_g1
          See other WAC-AS1 GE Assays ›
          Gene Symbol:
          WAC-AS1
          Gene Name:
          WAC antisense RNA 1 (head to head)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 28519917 - 28532354 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01903755_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: WAC-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033805.1 3 3476 128

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000010145.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10001140.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P27507282 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 80888 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 134237 AF151855.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 134237 AK001779.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 134237 AK289609.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 134237 AY169963.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 134237 BC065700.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 134237 CR591576.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 134237 CR593490.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 134237 CR596032.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 134237 CR597378.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 134237 CR599805.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 134237 CR601234.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 134237 CR601377.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 134237 CR604577.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 134237 CR606525.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 134237 CR606624.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 134237 CR610261.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 134237 CR612387.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 134237 CR613995.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 134237 CR622210.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 134237 CR622442.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 134237 CR626007.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 134237 NM_016038.2 refseq_mrna_human_NM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 675534 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 702109 CR600852.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 702109 XM_001130620.1 refseq_mrna_human_XM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 702109 XM_001130637.1 refseq_mrna_human_XM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 702109 XM_001130649.1 refseq_mrna_human_XM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 702109 XM_001130661.1 refseq_mrna_human_XM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 702109 XM_001130669.1 refseq_mrna_human_XM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 702109 XM_001130681.1 refseq_mrna_human_XM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 702109 XM_001131584.1 refseq_mrna_human_XM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 702109 XM_001131589.1 refseq_mrna_human_XM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 702109 XM_001131594.1 refseq_mrna_human_XM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 702109 XM_001131599.1 refseq_mrna_human_XM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 702109 XM_001131602.1 refseq_mrna_human_XM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 702109 XM_001131605.1 refseq_mrna_human_XM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 702109 XM_001131621.1 refseq_mrna_human_XM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 702109 XM_001131627.1 refseq_mrna_human_XM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 702109 XM_932750.2 refseq_mrna_human_XM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 743130 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P879563 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 10651 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P18876822 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P18876822 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3374718 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P879563 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P18876822 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P879563 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18876822 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18876819 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18876827 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P18876827 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P18876822 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 233013_PM_x_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR10014293.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR10014293.hg.1 NR_033805.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10014293.hg.1 TC10001140.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010145.hg.1 ENST00000528337.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010145.hg.1 ENST00000528337.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010145.hg.1 ENST00000528337.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010145.hg.1 ENST00000528337.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010145.hg.1 ENST00000528337.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010145.hg.1 ENST00000528337.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000010145.hg.1 NR_033805.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010145.hg.1 NR_033805.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010145.hg.1 NR_033805.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010145.hg.1 NR_033805.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010145.hg.1 NR_033805.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010145.hg.1 NR_033805.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010145.hg.1 NR_033805.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000010145.hg.1 NR_033805.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10014292.hg.1 NR_033805.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10014292.hg.1 TC10001140.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1000169165.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000169165.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000169165.hg.1 ENST00000528337.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000169165.hg.1 NR_033805.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000169165.hg.1 TC1000010145.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000169165.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000169165.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000169165.hg.1 ENST00000528337.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000169165.hg.1 NR_033805.1 Refseq
          Affymetrix Human Clariom D Assay TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010145.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010145.hg.1 ENST00000528337.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010145.hg.1 ENST00000528337.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010145.hg.1 ENST00000528337.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010145.hg.1 ENST00000528337.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010145.hg.1 ENST00000528337.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010145.hg.1 ENST00000528337.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000010145.hg.1 NR_033805.1 Refseq
          Affymetrix Human Clariom D Assay TC1000010145.hg.1 NR_033805.1 Refseq
          Affymetrix Human Clariom D Assay TC1000010145.hg.1 NR_033805.1 Refseq
          Affymetrix Human Clariom D Assay TC1000010145.hg.1 NR_033805.1 Refseq
          Affymetrix Human Clariom D Assay TC1000010145.hg.1 NR_033805.1 Refseq
          Affymetrix Human Clariom D Assay TC1000010145.hg.1 NR_033805.1 Refseq
          Affymetrix Human Clariom D Assay TC1000010145.hg.1 NR_033805.1 Refseq
          Affymetrix Human Clariom D Assay TC1000010145.hg.1 NR_033805.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000169163.hg.1 NR_033805.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000169163.hg.1 TC1000010145.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000169163.hg.1 NR_033805.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000169164.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000169164.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000169164.hg.1 NR_033805.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000169164.hg.1 TC1000010145.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000169164.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000169164.hg.1 ENST00000527986.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000169164.hg.1 NR_033805.1 Refseq
          Human Genome U133 Plus 2.0 Array 225515_s_at - -
          Human Genome U133 Plus 2.0 Array 233013_x_at - -
          Human Genome U133 Plus 2.0 Array 233013_PM_x_at - -
          Human Genome U133 Plus 2.0 Array 225515_PM_s_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233013_x_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233013_x_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233013_x_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233013_x_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233013_x_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233013_x_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233013_x_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233013_x_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233013_x_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233013_x_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 233013_x_at7 - -
          Human Genome U95 Set 48946_PM_at - -
          Human Genome U95 Set 48946_at - -
          Human Genome U95 Set 84705_PM_at - -
          Human Genome U95 Set 84705_at - -
          Human Genome U95 Set 89154_at - -
          Human Genome U95 Set 89523_r_at - -
          Human Genome U95 Set 89154_PM_at - -
          Human Genome U95 Set 89523_PM_r_at - -
          Human Genome U133 Set 233013_PM_x_at - -
          Human Genome U133 Set 233013_x_at - -
          Human Genome U133 Set 225515_s_at - -
          Human Genome U133 Set 225515_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A042560 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A042560 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 151395 - -
          ABI Human Genome Survey Microarray Version 2.0 243149 - -
          ABI Human Genome Survey Microarray Version 2.0 590611 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-595d89cdf5-c6dj8:80/100.66.135.190:80.
          git-commit: bccefdb7e3170cc2d597cd09baf15c702f2b69d9
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0