Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01904942_s1
          See other FASLG GE Assays ›
          Gene Symbol:
          FASLG
          Gene Name:
          Fas ligand
          Gene Aliases:
          ALPS1B, APT1LG1, APTL, CD178, CD95-L, CD95L, FASL, TNFSF6, TNLG1A
          Chromosome Location:
          Chr.1: 172659008 - 172666873 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01904942_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FASLG
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000639.2 NP_000630.1 1 498 69
          NM_001302746.1 NP_001289675.1 1 498 69
          GenBank mRNA AF288573.1 - 1 362 69
          AK314874.1 - 1 462 69
          AY225406.1 - 1 304 69
          AY858799.1 - 1 304 69
          BC017502.1 - 1 391 4849770 69
          D38122.1 - 1 492 69
          DA916336.1 - 1 495 69
          KJ948655.1 - 1 461 69
          U08137.1 - 1 389 69
          U11821.1 - 1 368 69
          X89102.1 - 1 461 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100010625.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01001506.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene FASLG

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          transcription, DNA-templated
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          inflammatory cell apoptotic process
          immune response
          signal transduction
          cell-cell signaling
          positive regulation of cell population proliferation
          extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of angiogenesis
          cellular chloride ion homeostasis
          response to lipopolysaccharide
          positive regulation of apoptotic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of neuron apoptotic process
          positive regulation of epidermal growth factor receptor signaling pathway
          retinal cell programmed cell death
          endosomal lumen acidification
          T cell apoptotic process
          necroptotic process
          response to growth factor
          apoptotic signaling pathway
          extrinsic apoptotic signaling pathway
          activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
          necroptotic signaling pathway
          regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          positive regulation of endothelial cell apoptotic process
          signaling receptor binding
          death receptor binding
          cytokine activity
          tumor necrosis factor receptor binding
          protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01023368.hg.1 ENST00000340030.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01023368.hg.1 ENST00000367721.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01023368.hg.1 NM_000639.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01023368.hg.1 TC01001506.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01023369.hg.1 ENST00000367721.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01023369.hg.1 NM_000639.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01023369.hg.1 TC01001506.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 ENST00000367721.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 ENST00000367721.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 ENST00000367721.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 ENST00000367721.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 ENST00000367721.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 ENST00000367721.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 NM_000639.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 NM_000639.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 NM_000639.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 NM_000639.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 NM_000639.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 NM_000639.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 NM_000639.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 NM_000639.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 NM_000639.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 NM_001302746.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 NM_001302746.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 NM_001302746.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 NM_001302746.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 NM_001302746.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 NM_001302746.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 NM_001302746.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 NM_001302746.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 ENST00000340030.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 ENST00000340030.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 ENST00000340030.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 ENST00000340030.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 ENST00000340030.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 ENST00000367721.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 ENST00000367721.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100010625.hg.1 ENST00000367721.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01023367.hg.1 ENST00000340030.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01023367.hg.1 ENST00000367721.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01023367.hg.1 NM_000639.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01023367.hg.1 TC01001506.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100178235.hg.1 ENST00000340030.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100178235.hg.1 ENST00000367721.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100178235.hg.1 NM_000639.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100178235.hg.1 NM_001302746.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100178235.hg.1 TC0100010625.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100178235.hg.1 NM_001302746.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100178235.hg.1 ENST00000340030.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100178235.hg.1 ENST00000367721.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100178235.hg.1 NM_000639.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100178232.hg.1 ENST00000367721.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100178232.hg.1 NM_000639.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100178232.hg.1 NM_001302746.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100178232.hg.1 TC0100010625.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100178232.hg.1 ENST00000367721.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100178232.hg.1 NM_000639.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100178232.hg.1 NM_001302746.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100178234.hg.1 ENST00000340030.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100178234.hg.1 ENST00000367721.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100178234.hg.1 NM_000639.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100178234.hg.1 NM_001302746.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100178234.hg.1 TC0100010625.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100178234.hg.1 ENST00000340030.4 Ensembl
          Affymetrix Human Clariom D Assay PSR0100178234.hg.1 ENST00000367721.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100178234.hg.1 NM_000639.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100178234.hg.1 NM_001302746.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 ENST00000367721.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 ENST00000367721.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 ENST00000367721.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 ENST00000367721.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 ENST00000367721.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 ENST00000367721.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 NM_000639.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 NM_000639.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 NM_000639.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 NM_000639.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 NM_000639.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 NM_000639.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 NM_000639.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 NM_000639.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 NM_000639.2 Refseq
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 NM_001302746.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 NM_001302746.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 NM_001302746.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 NM_001302746.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 NM_001302746.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 NM_001302746.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 NM_001302746.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 NM_001302746.1 Refseq
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 ENST00000340030.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 ENST00000340030.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 ENST00000340030.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 ENST00000340030.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 ENST00000340030.4 Ensembl
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 ENST00000367721.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 ENST00000367721.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100010625.hg.1 ENST00000367721.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100178233.hg.1 ENST00000367721.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100178233.hg.1 NM_000639.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100178233.hg.1 NM_001302746.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100178233.hg.1 TC0100010625.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100178233.hg.1 ENST00000367721.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100178233.hg.1 NM_000639.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100178233.hg.1 NM_001302746.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 116304 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0