Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs01942239_s1
          See other LINC00649 GE Assays ›
          Gene Symbol:
          LINC00649
          Gene Name:
          long intergenic non-protein coding RNA 649
          Gene Aliases:
          -
          Chromosome Location:
          Chr.21: 33931122 - 33971186 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs01942239_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC00649
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK127913.1 - 2 2159 87

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361549 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361503 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361496 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361509 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361555 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361540 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361514 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361524 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361543 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361527 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361530 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32021738 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32021779 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32021776 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361534 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32021705 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361499 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32021712 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32021732 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361501 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361545 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361525 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361544 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32021742 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361535 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361562 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32021709 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361508 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361570 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32021735 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361546 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32021728 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361556 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361552 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361491 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361538 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361506 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361571 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32021777 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361532 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32021723 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32021736 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361505 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32021737 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361547 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32021747 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32021727 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32021751 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361554 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361561 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361517 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361504 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361502 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361569 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32021745 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361500 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32021706 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361559 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361560 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11361550 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 14641 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3263851 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8068352 - -
          Affymetrix Human Gene 1.0 ST Array 8068351 - -
          Affymetrix Human Gene 1.1 ST Array 8068350 - -
          Human Genome U95 Set 51337_PM_at - -
          Human Genome U95 Set 51337_at - -
          Human Genome U95 Set 57360_PM_at - -
          Human Genome U95 Set 61137_PM_at - -
          Human Genome U95 Set 65330_at - -
          Human Genome U95 Set 65330_PM_at - -
          Human Genome U95 Set 57360_at - -
          Human Genome U95 Set 61137_at - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1786559 AK127913.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1786559 AK127913.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1786559 AK127913.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1786559 AK127913.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1786559 AK127913.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1786559 AK127913.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0