Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02330075_g1
          See other MYOD1 GE Assays ›
          Gene Symbol:
          MYOD1
          Gene Name:
          myogenic differentiation 1
          Gene Aliases:
          MYF3, MYOD, PUM, bHLHc1
          Chromosome Location:
          Chr.11: 17719563 - 17722131 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02330075_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MYOD1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002478.4 NP_002469.2 1 - 2 853 87
          GenBank mRNA BC064493.1 - 1 - 2 695 5022419 87
          BT007157.1 - 1 - 2 633 87
          X17650.1 - 1 - 2 474 87
          X56677.1 - 1 - 2 750 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100006976.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11000232.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MYOD1

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of alternative mRNA splicing, via spliceosome
          regulation of transcription by RNA polymerase II
          transcription by RNA polymerase II
          protein phosphorylation
          muscle organ development
          myoblast fate determination
          skeletal muscle tissue development
          myoblast fusion
          cellular response to starvation
          myotube cell development
          myotube differentiation involved in skeletal muscle regeneration
          skeletal muscle cell differentiation
          muscle cell fate commitment
          positive regulation of skeletal muscle tissue regeneration
          regulation of RNA splicing
          skeletal muscle fiber adaptation
          histone H3 acetylation
          histone H4 acetylation
          positive regulation of myoblast differentiation
          positive regulation of transcription by RNA polymerase II
          skeletal muscle fiber development
          positive regulation of skeletal muscle fiber development
          positive regulation of muscle cell differentiation
          cellular response to glucocorticoid stimulus
          cellular response to estradiol stimulus
          cellular response to oxygen levels
          positive regulation of myoblast fusion
          negative regulation of myoblast proliferation
          RNA polymerase II transcription regulatory region sequence-specific DNA binding
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          DNA-binding transcription factor activity, RNA polymerase II-specific
          core promoter sequence-specific DNA binding
          DNA-binding transcription activator activity, RNA polymerase II-specific
          chromatin binding
          transcription coactivator activity
          protein binding
          transcription factor binding
          chromatin DNA binding
          ubiquitin protein ligase binding
          protein heterodimerization activity
          E-box binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR11003084.hg.1 ENST00000250003.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11003084.hg.1 NM_002478.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11003084.hg.1 TC11000232.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR11003086.hg.1 ENST00000250003.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11003086.hg.1 NM_002478.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11003086.hg.1 TC11000232.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1100006976.hg.1 ENST00000250003.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006976.hg.1 ENST00000250003.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006976.hg.1 ENST00000250003.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006976.hg.1 ENST00000250003.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006976.hg.1 ENST00000250003.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006976.hg.1 ENST00000250003.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006976.hg.1 ENST00000250003.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006976.hg.1 NM_002478.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006976.hg.1 NM_002478.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006976.hg.1 NM_002478.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006976.hg.1 NM_002478.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006976.hg.1 NM_002478.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006976.hg.1 NM_002478.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100006976.hg.1 NM_002478.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11003088.hg.1 ENST00000250003.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11003088.hg.1 NM_002478.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11003088.hg.1 TC11000232.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR11003087.hg.1 ENST00000250003.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR11003087.hg.1 NM_002478.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11003087.hg.1 TC11000232.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1100149135.hg.1 ENST00000250003.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100149135.hg.1 NM_002478.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100149135.hg.1 TC1100006976.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100149135.hg.1 NM_002478.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100149135.hg.1 ENST00000250003.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100149137.hg.1 ENST00000250003.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100149137.hg.1 NM_002478.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100149137.hg.1 TC1100006976.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100149137.hg.1 ENST00000250003.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100149137.hg.1 NM_002478.4 Refseq
          Affymetrix Human Clariom D Assay TC1100006976.hg.1 ENST00000250003.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100006976.hg.1 ENST00000250003.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100006976.hg.1 ENST00000250003.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100006976.hg.1 ENST00000250003.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100006976.hg.1 ENST00000250003.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100006976.hg.1 ENST00000250003.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100006976.hg.1 ENST00000250003.3 Ensembl
          Affymetrix Human Clariom D Assay TC1100006976.hg.1 NM_002478.4 Refseq
          Affymetrix Human Clariom D Assay TC1100006976.hg.1 NM_002478.4 Refseq
          Affymetrix Human Clariom D Assay TC1100006976.hg.1 NM_002478.4 Refseq
          Affymetrix Human Clariom D Assay TC1100006976.hg.1 NM_002478.4 Refseq
          Affymetrix Human Clariom D Assay TC1100006976.hg.1 NM_002478.4 Refseq
          Affymetrix Human Clariom D Assay TC1100006976.hg.1 NM_002478.4 Refseq
          Affymetrix Human Clariom D Assay TC1100006976.hg.1 NM_002478.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100149140.hg.1 ENST00000250003.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100149140.hg.1 NM_002478.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100149140.hg.1 TC1100006976.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100149140.hg.1 ENST00000250003.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100149140.hg.1 NM_002478.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100149139.hg.1 ENST00000250003.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100149139.hg.1 NM_002478.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100149139.hg.1 TC1100006976.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100149139.hg.1 ENST00000250003.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100149139.hg.1 NM_002478.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100149138.hg.1 ENST00000250003.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100149138.hg.1 NM_002478.4 Refseq
          Affymetrix Human Clariom D Assay PSR1100149138.hg.1 TC1100006976.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100149138.hg.1 ENST00000250003.3 Ensembl
          Affymetrix Human Clariom D Assay PSR1100149138.hg.1 NM_002478.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline