Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02341925_s1
          See other GPR152 GE Assays ›
          Gene Symbol:
          GPR152
          Gene Name:
          G protein-coupled receptor 152
          Gene Aliases:
          PGR5
          Chromosome Location:
          Chr.11: 67451301 - 67452729 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02341925_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: GPR152
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_206997.1 NP_996880.1 1 841 157
          GenBank mRNA AY569571.1 - 1 841 157
          BC119779.1 - 1 841 40109407 157
          BC122869.1 - 1 841 40109404 157

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene GPR152

          Gene Ontology Categories:

          Function(s) Process(es)

          angiotensin-activated signaling pathway
          angiotensin receptor activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07851091 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28264980 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28264979 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07851094 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07851095 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07851103 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07851096 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07851101 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07851100 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07851092 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16464841 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07851097 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07851099 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3243429 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3509 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3510 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 57445 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3243429 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3243429 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07851103 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07851096 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07851095 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07851097 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28264979 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16464841 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07851093 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07851100 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07851094 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07851102 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07851092 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07851101 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16464841 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07851096 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07851093 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07851103 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07851092 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07851101 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07851095 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28264979 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07851094 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07851102 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07851097 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07851100 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07851108 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07851109 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07851093 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07851095 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07851090 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07851097 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07851096 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07851092 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07851101 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07851111 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07851112 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07851104 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07851102 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07851106 - -
          Agilent Human Promoter ChIP-on-Chip A) A_17_P07851099 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16464818 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851137 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851126 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851096 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851122 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851063 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851111 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851104 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851118 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851056 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851115 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16464831 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851057 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851109 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851060 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851112 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851119 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851066 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851106 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16464842 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16464817 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851138 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851093 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851135 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851095 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851140 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851108 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851124 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851099 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851145 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851144 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851132 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851097 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16464849 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851062 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851101 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16464852 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851090 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16464821 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16464825 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16464829 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851117 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851059 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851054 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851092 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16464851 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07851129 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16464846 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_206997_at - -
          Affymetrix Human Gene 1.0 ST Array 7949797 - -
          Affymetrix Human Gene 1.1 ST Array 7949796 - -
          Affymetrix Human Gene 1.1 ST Array 7941917 - -
          Affymetrix Human Genome U219 Array 11734634_PM_at - -
          Affymetrix Human Genome U219 Array 11734634_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A045413 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A045413 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1659982 NM_206997.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1659982 AY569571.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1659982 BC119779.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1659982 BC122869.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1659982 NM_206997.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1659982 AY569571.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1659982 BC119779.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1659982 BC122869.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1659982 NM_206997.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1659982 AY569571.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1659982 BC119779.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1659982 BC122869.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1659982 NM_206997.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1659982 AY569571.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1659982 BC119779.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1659982 BC122869.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1659982 NM_206997.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1659982 AY569571.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1659982 BC119779.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1659982 BC122869.1 human_transcriptome
          Illumina HumanRef-8 v2 Expression BeadChip GI_7047-S - -
          Illumina human-6 v2.0 expression beadchip ILMN_1659982 NM_206997.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1659982 AY569571.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1659982 BC119779.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1659982 BC122869.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1659982 NM_206997.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1659982 AY569571.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1659982 BC119779.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1659982 BC122869.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 380270 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_7047 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1659982 NM_206997.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1659982 AY569571.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1659982 BC119779.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_1659982 BC122869.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1659982 NM_206997.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1659982 AY569571.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1659982 BC119779.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1659982 BC122869.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1659982 NM_206997.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1659982 AY569571.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1659982 BC119779.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1659982 BC122869.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1659982 NM_206997.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1659982 AY569571.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1659982 BC119779.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1659982 BC122869.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1659982 NM_206997.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1659982 AY569571.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1659982 BC119779.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1659982 BC122869.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0