Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02384547_gH
          See other LOC401127 GE Assays ›
          Gene Symbol:
          LOC401127
          Gene Name:
          WD repeat domain 5 pseudogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 39480255 - 39481905 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02384547_gH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC401127
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_026854.1 1 1443 116
          GenBank mRNA BC010526.2 - 2 1324 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0400007286.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P41267 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P41267 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 10126 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 46824 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P41267 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P41267 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23206615 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23206634 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23206628 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15560979 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03053310 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03053319 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03053313 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03053308 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23206631 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15560976 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03053304 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03053301 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23206621 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03053300 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23206618 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03053315 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03053309 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03053306 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23206626 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23206634 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23206621 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15560976 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03053310 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03053315 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03053319 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03053313 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23206618 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03053308 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23206631 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23206626 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23206628 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03053301 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03053309 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15560979 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03053306 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03053300 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03053304 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23206615 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P41267 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P41267 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03053318 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03053304 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03053302 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15560974 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03053314 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03053324 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03053313 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03053315 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03053299 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03053305 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03053311 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03053320 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15560978 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P03053309 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8094702 - -
          Affymetrix Human Gene 1.0 ST Array 8094703 - -
          Affymetrix Human Gene 1.1 ST Array 8094701 - -
          Affymetrix Human Transcriptome Array 2.0 TC0400007286.hg.1 ENST00000507033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007286.hg.1 ENST00000507033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007286.hg.1 ENST00000507033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007286.hg.1 ENST00000507033.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007286.hg.1 ENST00000513652.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007286.hg.1 ENST00000513652.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007286.hg.1 ENST00000513652.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007286.hg.1 ENST00000513652.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007286.hg.1 ENST00000513652.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007286.hg.1 ENST00000513652.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007286.hg.1 ENST00000597991.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007286.hg.1 ENST00000597991.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007286.hg.1 ENST00000597991.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007286.hg.1 ENST00000597991.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007286.hg.1 ENST00000597991.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007286.hg.1 ENST00000597991.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400007286.hg.1 NR_026854.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007286.hg.1 NR_026854.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007286.hg.1 NR_026854.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007286.hg.1 NR_026854.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007286.hg.1 NR_026854.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007286.hg.1 NR_026854.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007286.hg.1 NR_026854.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400007286.hg.1 NR_026854.1 Refseq
          Affymetrix Human Clariom D Assay TC0400007286.hg.1 ENST00000507033.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400007286.hg.1 ENST00000507033.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400007286.hg.1 ENST00000507033.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400007286.hg.1 ENST00000507033.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400007286.hg.1 ENST00000513652.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400007286.hg.1 ENST00000513652.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400007286.hg.1 ENST00000513652.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400007286.hg.1 ENST00000513652.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400007286.hg.1 ENST00000513652.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400007286.hg.1 ENST00000513652.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400007286.hg.1 ENST00000597991.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400007286.hg.1 ENST00000597991.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400007286.hg.1 ENST00000597991.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400007286.hg.1 ENST00000597991.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400007286.hg.1 ENST00000597991.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400007286.hg.1 ENST00000597991.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400007286.hg.1 NR_026854.1 Refseq
          Affymetrix Human Clariom D Assay TC0400007286.hg.1 NR_026854.1 Refseq
          Affymetrix Human Clariom D Assay TC0400007286.hg.1 NR_026854.1 Refseq
          Affymetrix Human Clariom D Assay TC0400007286.hg.1 NR_026854.1 Refseq
          Affymetrix Human Clariom D Assay TC0400007286.hg.1 NR_026854.1 Refseq
          Affymetrix Human Clariom D Assay TC0400007286.hg.1 NR_026854.1 Refseq
          Affymetrix Human Clariom D Assay TC0400007286.hg.1 NR_026854.1 Refseq
          Affymetrix Human Clariom D Assay TC0400007286.hg.1 NR_026854.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400149867.hg.1 ENST00000513652.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400149867.hg.1 ENST00000597991.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400149867.hg.1 NR_026854.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400149867.hg.1 TC0400007286.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400149867.hg.1 NR_026854.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400149867.hg.1 ENST00000513652.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400149867.hg.1 ENST00000597991.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400149868.hg.1 ENST00000513652.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400149868.hg.1 NR_026854.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400149868.hg.1 TC0400007286.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400149868.hg.1 ENST00000513652.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400149868.hg.1 NR_026854.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A025434 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A025434 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 498521 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3200378 CR604500.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3200378 CR619269.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3200378 NR_026854.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3200378 BC010526.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3200378 CR592370.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3276859 CR604500.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3276859 CR619269.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3276859 NR_026854.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3276859 BC010526.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3276859 CR592370.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0