Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02561709_m1
          See other PDE4DIP GE Assays ›
          Gene Symbol:
          PDE4DIP
          Gene Name:
          phosphodiesterase 4D interacting protein
          Gene Aliases:
          CMYA2, MMGL
          Chromosome Location:
          Chr.1: 148808167 - 149051277 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02561709_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PDE4DIP
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001002810.3 NP_001002810.1 1 - 2 440 81
          NM_001002812.2 NP_001002812.2 1 - 2 440 81
          NM_001195260.1 NP_001182189.1 2 - 3 137 81
          NM_001195261.1 NP_001182190.1 2 - 3 235 81
          NM_001198832.2 NP_001185761.2 4 - 5 722 81
          NM_001198834.3 NP_001185763.3 1 - 2 440 81
          NM_014644.5 NP_055459.5 1 - 2 440 81
          NM_022359.5 NP_071754.3 4 - 5 873 81
          XM_006711646.1 XP_006711709.1 4 - 5 729 81
          XM_006711650.3 XP_006711713.1 2 - 3 214 81
          XM_006711651.1 XP_006711714.1 1 - 2 332 81
          XM_006711652.1 XP_006711715.1 1 - 2 332 81
          XM_011510172.1 XP_011508474.1 4 - 5 887 81
          XM_011510173.1 XP_011508475.1 4 - 5 887 81
          XM_011510175.1 XP_011508477.1 4 - 5 887 81
          XM_011510176.1 XP_011508478.1 4 - 5 887 81
          XM_011510179.1 XP_011508481.1 1 - 2 332 81
          XM_017002878.1 XP_016858367.1 4 - 5 887 81
          XM_017002880.1 XP_016858369.1 4 - 5 887 81
          XM_017002887.1 XP_016858376.1 5 - 6 1164 81
          XM_017002888.1 XP_016858377.1 5 - 6 1137 81
          XM_017002889.1 XP_016858378.1 5 - 6 809 81
          XM_017002898.1 XP_016858387.1 4 - 5 729 81
          XM_017002899.1 XP_016858388.1 4 - 5 729 81
          XM_017002902.1 XP_016858391.1 4 - 5 729 81
          XM_017002903.1 XP_016858392.1 4 - 5 729 81
          GenBank mRNA AB007923.2 - 4 - 5 601 81
          AL831815.2 - 1 - 2 283 81
          AL833273.1 - 1 - 2 328 81
          BP286699.1 - 3 - 4 286 81
          BX647660.1 - 1 - 2 368 81
          DA764537.1 - 1 - 2 388 81
          DA766011.1 - 1 - 2 332 81
          DA860918.1 - 1 - 2 378 81

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the NM_001195260.1, XM_017002887.1, XM_017002888.1, XM_017002889.1 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular protein-containing complex assembly
          protein binding
          enzyme binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0