Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs02721466_g1
See other R3HCC1L GE Assays ›
Gene Symbol:
R3HCC1L
Gene Name:
R3H domain and coiled-coil containing 1 like
Gene Aliases:
C10orf28, GIDRP86, GIDRP88, PSORT
Chromosome Location:
Chr.10: 98134608 - 98244897 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs02721466_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: R3HCC1L
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001256619.1
NP_001243548.1
9 - 10
2522
101
NM_001256620.1
NP_001243549.1
7 - 8
2429
101
NM_001256621.1
NP_001243550.1
6 - 7
626
101
NM_014472.4
NP_055287.4
7 - 8
2425
101
NM_138469.2
NP_612478.2
8 - 9
2522
101
XM_011539639.1
XP_011537941.1
9 - 10
2779
101
XM_011539640.1
XP_011537942.1
10 - 11
2584
101
XM_011539641.1
XP_011537943.1
9 - 10
2536
101
XM_011539642.1
XP_011537944.1
10 - 11
2876
101
XM_011539643.1
XP_011537945.1
8 - 9
2463
101
XM_011539644.1
XP_011537946.1
8 - 9
2467
101
XM_011539645.2
XP_011537947.1
8 - 9
2710
101
XM_011539646.1
XP_011537948.1
7 - 8
2342
101
XM_011539647.1
XP_011537949.1
7 - 8
2344
101
XM_011539648.1
XP_011537950.1
6 - 7
2217
101
XM_011539649.1
XP_011537951.1
8 - 9
2737
101
XM_017016074.1
XP_016871563.1
8 - 9
3675
101
XM_017016075.1
XP_016871564.1
5 - 6
2175
101
XM_017016076.1
XP_016871565.1
8 - 9
2469
101
XM_017016077.1
XP_016871566.1
10 - 11
2632
101
XM_017016078.1
XP_016871567.1
9 - 10
2554
101
XM_017016079.1
XP_016871568.1
9 - 10
2568
101
XM_017016080.1
XP_016871569.1
7 - 8
2474
101
XM_017016081.1
XP_016871570.1
8 - 9
2556
101
XM_017016082.1
XP_016871571.1
6 - 7
2348
101
XM_017016083.1
XP_016871572.1
7 - 8
2364
101
GenBank mRNA
AF525304.1
-
9 - 10
2475
101
AK058187.1
-
3 - 4
216
101
AK094479.1
-
4 - 5
936
101
AK292546.1
-
7 - 8
2424
101
BC009231.2
-
3 - 4
351
101
BC047908.1
-
6 - 7
572
101
BC098175.1
-
4 - 5
369
101
BC098268.1
-
8 - 9
2463
40026913
101
BC098359.1
-
7 - 8
2366
40026912
101
BC099848.1
-
7 - 8
2366
40026916
101
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length greater than or equal to 101
Mammalian Gene Collection
Detect Genomic DNA
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene R3HCC1L
Gene Ontology Categories:
Process(es)
nucleotide binding
protein binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0