Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs02742336_s1
          See other EHMT1-IT1 GE Assays ›
          Gene Symbol:
          EHMT1-IT1
          Gene Name:
          EHMT1 intronic transcript 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 137763022 - 137764772 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs02742336_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: EHMT1-IT1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_024341.1 1 735 96
          GenBank mRNA AK097611.1 - 1 744 96
          AK127990.1 - 1 744 96
          BC132855.1 - 1 252 96
          BC132857.1 - 1 252 96

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ARRDC1 antisense RNA 1 ARRDC1-AS1 C9orf37 -144.1kb Anti-sense Chr. 9 - 137615332 - 137618895 on NCBI Build GRCh38
          Downstream microRNA 602 MIR602 MIRN602,
          hsa-mir-602
          73.6kb Sense Chr. 9 - 137838419 - 137838516 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0900009318.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC09000853.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P93736 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 5203 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 60829 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P93736 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P93736 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014050 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014068 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014025 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014036 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16280707 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366820 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014041 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014078 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366856 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014022 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014053 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014034 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366827 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014046 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014043 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014062 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014051 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366840 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014063 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366812 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014073 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366838 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014080 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014045 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366845 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366818 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014075 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014052 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014066 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366810 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014077 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366849 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16280711 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366806 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366860 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366826 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014027 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366808 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366811 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366805 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014033 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014055 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014020 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366817 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014077 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014052 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366805 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366849 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014034 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366860 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366845 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014046 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366810 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014075 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366812 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014033 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366827 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366817 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014073 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16280711 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014050 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014051 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014080 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014062 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014066 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366838 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014078 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014068 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014045 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366856 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014055 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014063 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014053 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366818 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16280707 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014025 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014041 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366806 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366840 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014043 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014027 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014036 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014022 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366820 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366826 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366811 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07014020 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27366808 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P93736 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P93736 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014045 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014013 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014036 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014047 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014043 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014050 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014022 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16280706 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014070 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014017 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014057 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014028 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014023 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16280711 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16280703 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014066 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014073 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014055 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014058 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014033 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16280720 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014063 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014032 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014037 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014077 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014025 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014078 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014068 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16280707 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014060 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014065 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014041 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014034 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014026 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014039 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014051 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014075 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07014020 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8159733 - -
          Affymetrix Human Gene 1.1 ST Array 8159732 - -
          Affymetrix HT HG-U133+ PM Array Plate 1559311_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0900009318.hg.1 ENST00000629335.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009318.hg.1 ENST00000629335.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009318.hg.1 ENST00000629335.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900009318.hg.1 NR_024341.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009318.hg.1 NR_024341.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0900009318.hg.1 NR_024341.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09011346.hg.1 NR_024341.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR09011346.hg.1 TC09000853.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0900009318.hg.1 ENST00000629335.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900009318.hg.1 ENST00000629335.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900009318.hg.1 ENST00000629335.1 Ensembl
          Affymetrix Human Clariom D Assay TC0900009318.hg.1 NR_024341.1 Refseq
          Affymetrix Human Clariom D Assay TC0900009318.hg.1 NR_024341.1 Refseq
          Affymetrix Human Clariom D Assay TC0900009318.hg.1 NR_024341.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900163963.hg.1 ENST00000629335.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900163963.hg.1 NR_024341.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900163963.hg.1 TC0900009318.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900163963.hg.1 NR_024341.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900163963.hg.1 ENST00000629335.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900163964.hg.1 ENST00000629335.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900163964.hg.1 NR_024341.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900163964.hg.1 TC0900009318.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900163964.hg.1 ENST00000629335.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900163964.hg.1 NR_024341.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900163962.hg.1 ENST00000629335.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900163962.hg.1 NR_024341.1 Refseq
          Affymetrix Human Clariom D Assay PSR0900163962.hg.1 TC0900009318.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0900163962.hg.1 ENST00000629335.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0900163962.hg.1 NR_024341.1 Refseq
          Human Genome U133 Plus 2.0 Array 1559311_at - -
          Human Genome U133 Plus 2.0 Array 1559311_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559311_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559311_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559311_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559311_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559311_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559311_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559311_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559311_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559311_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559311_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559311_at11 - -
          Human Genome U95 Set 42638_at - -
          Human Genome U95 Set 42638_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A039503 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A039503 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1707318 BC132857.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1707318 AK097611.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1707318 AK127990.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1707318 BC132855.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1689131 NR_024341.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1707318 BC132857.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1707318 AK097611.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1707318 AK127990.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1707318 BC132855.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1689131 NR_024341.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1689131 NR_024341.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1707318 BC132857.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1707318 AK097611.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1707318 AK127990.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1707318 BC132855.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1689131 NR_024341.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-f85dfd666-px2hc:80/100.66.130.62:80.
          git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0