Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs02811007_m1
See other ACBD5 GE Assays ›
Gene Symbol:
ACBD5
Gene Name:
acyl-CoA binding domain containing 5
Gene Aliases:
-
Chromosome Location:
Chr.10: 27182624 - 27248674 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs02811007_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ACBD5
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001042473.3
NP_001035938.1
4 - 5
446
85
NM_001271512.3
NP_001258441.1
3 - 4
760
85
NM_001301251.1
NP_001288180.1
3 - 4
325
85
NM_001301252.1
NP_001288181.1
2 - 3
639
85
NM_001301253.1
NP_001288182.1
3 - 4
315
85
NM_001301254.1
NP_001288183.1
2 - 3
639
85
NM_145698.4
NP_663736.2
4 - 5
571
85
XM_006717528.3
XP_006717591.3
4 - 5
6971
85
XM_006717530.3
XP_006717593.1
4 - 5
570
85
XM_006717531.2
XP_006717594.1
4 - 5
580
85
XM_006717535.2
XP_006717598.1
4 - 5
437
85
XM_017016884.1
XP_016872373.1
4 - 5
6971
85
XM_017016885.1
XP_016872374.1
4 - 5
6971
85
XM_017016886.1
XP_016872375.1
4 - 5
6971
85
XM_017016887.1
XP_016872376.1
4 - 5
501
85
XM_017016888.1
XP_016872377.1
4 - 5
566
85
XM_017016889.1
XP_016872378.1
4 - 5
501
85
XM_017016890.1
XP_016872379.1
4 - 5
6971
85
XM_017016891.1
XP_016872380.1
4 - 5
566
85
XM_017016892.1
XP_016872381.1
4 - 5
487
85
XM_017016893.1
XP_016872382.1
4 - 5
6971
85
XM_017016894.1
XP_016872383.1
4 - 5
6971
85
XM_017016895.1
XP_016872384.1
4 - 5
437
85
XM_017016896.1
XP_016872385.1
4 - 5
438
85
XM_017016897.1
XP_016872386.1
4 - 5
566
85
XM_017016898.1
XP_016872387.1
4 - 5
566
85
XM_017016899.1
XP_016872388.1
4 - 5
566
85
XM_017016900.1
XP_016872389.1
3 - 4
355
85
XM_017016901.1
XP_016872390.1
3 - 4
335
85
XM_017016902.1
XP_016872391.1
3 - 4
354
85
XM_017016903.1
XP_016872392.1
3 - 4
339
85
XM_017016904.1
XP_016872393.1
3 - 4
367
85
XM_017016905.1
XP_016872394.1
3 - 4
353
85
XM_017016906.1
XP_016872395.1
4 - 5
438
85
XM_017016907.1
XP_016872396.1
3 - 4
367
85
XM_017016908.1
XP_016872397.1
3 - 4
356
85
GenBank mRNA
AB082527.1
-
3 - 4
274
85
AF505653.1
-
3 - 4
312
85
AK057469.1
-
4 - 5
438
85
AK294510.1
-
2 - 3
557
85
AK295031.1
-
3 - 4
397
85
AK304842.1
-
2 - 3
290
85
BC030555.1
-
4 - 5
475
5218685
85
DC333545.1
-
4 - 5
449
85
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length between 71 and 85
Mammalian Gene Collection
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene ACBD5
Gene Ontology Categories:
Function(s)
Process(es)
transport
macroautophagy
autophagy of peroxisome
aggrephagy
fatty-acyl-CoA binding
molecular_function
lipid binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0