Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03003721_g1
          See other SRD5A2 GE Assays ›
          Gene Symbol:
          SRD5A2
          Gene Name:
          steroid 5 alpha-reductase 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 31522486 - 31665651 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03003721_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SRD5A2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000348.3 NP_000339.2 3 - 4 616 69
          XM_011533069.2 XP_011531371.1 3 - 4 1331 69
          XM_011533072.2 XP_011531374.1 6 - 7 761 69
          GenBank mRNA AK307235.1 - 3 - 4 352 69
          AK316597.1 - 3 - 4 586 69
          BC112252.1 - 3 - 4 603 8327720 69
          BC113641.1 - 3 - 4 591 8322693 69
          EF560740.1 - 3 - 4 584 69
          M74047.1 - 3 - 4 572 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200012172.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02001716.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SRD5A2

          Gene Ontology Categories:

          Function(s) Process(es)

          steroid biosynthetic process
          androgen biosynthetic process
          steroid catabolic process
          cell-cell signaling
          androgen metabolic process
          male gonad development
          response to xenobiotic stimulus
          biphenyl metabolic process
          dibenzo-p-dioxin metabolic process
          phthalate metabolic process
          hippocampus development
          hypothalamus development
          cell differentiation
          male genitalia development
          female genitalia development
          response to nutrient levels
          response to follicle-stimulating hormone
          response to testosterone
          response to peptide hormone
          response to steroid hormone
          bone development
          3-oxo-5-alpha-steroid 4-dehydrogenase activity
          sterol 5-alpha reductase activity
          amide binding
          cholestenone 5-alpha-reductase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02027702.hg.1 ENST00000233139.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02027702.hg.1 ENST00000405650.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02027702.hg.1 NM_000348.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02027702.hg.1 TC02001716.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 ENST00000233139.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 ENST00000233139.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 ENST00000233139.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 ENST00000233139.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 ENST00000233139.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 ENST00000233139.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 ENST00000233139.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 ENST00000405650.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 ENST00000405650.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 ENST00000405650.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 ENST00000405650.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 ENST00000405650.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 ENST00000405650.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 ENST00000405650.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 ENST00000405650.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 ENST00000405650.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 ENST00000405650.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 ENST00000405650.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 ENST00000622030.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 ENST00000622030.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 ENST00000622030.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 ENST00000622030.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 ENST00000622030.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 ENST00000622030.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 ENST00000622030.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 ENST00000622030.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 ENST00000622030.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 ENST00000622030.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 ENST00000622030.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 NM_000348.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 NM_000348.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 NM_000348.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 NM_000348.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 NM_000348.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 NM_000348.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 NM_000348.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 NM_000348.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200012172.hg.1 NM_000348.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200188859.hg.1 ENST00000233139.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200188859.hg.1 ENST00000405650.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200188859.hg.1 ENST00000622030.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200188859.hg.1 NM_000348.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200188859.hg.1 TC0200012172.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200188859.hg.1 NM_000348.3 Refseq
          Affymetrix Human Clariom D Assay PSR0200188859.hg.1 ENST00000233139.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200188859.hg.1 ENST00000405650.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200188859.hg.1 ENST00000622030.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 ENST00000233139.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 ENST00000233139.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 ENST00000233139.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 ENST00000233139.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 ENST00000233139.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 ENST00000233139.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 ENST00000233139.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 ENST00000405650.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 ENST00000405650.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 ENST00000405650.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 ENST00000405650.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 ENST00000405650.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 ENST00000405650.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 ENST00000405650.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 ENST00000405650.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 ENST00000405650.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 ENST00000405650.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 ENST00000405650.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 ENST00000622030.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 ENST00000622030.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 ENST00000622030.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 ENST00000622030.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 ENST00000622030.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 ENST00000622030.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 ENST00000622030.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 ENST00000622030.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 ENST00000622030.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 ENST00000622030.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 ENST00000622030.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 NM_000348.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 NM_000348.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 NM_000348.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 NM_000348.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 NM_000348.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 NM_000348.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 NM_000348.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 NM_000348.3 Refseq
          Affymetrix Human Clariom D Assay TC0200012172.hg.1 NM_000348.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-59df64f8c5-6gx82:80/100.66.130.62:80.
          git-commit: fe83b6adba5b7aa08a343fc2038c841d76c6a25b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0