Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018087.hg.1 |
ENST00000338487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018087.hg.1 |
NM_001003892.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018087.hg.1 |
TC10001427.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011070.hg.1 |
ENST00000338487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011070.hg.1 |
ENST00000338487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011070.hg.1 |
ENST00000338487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011070.hg.1 |
NM_001003892.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011070.hg.1 |
NM_001003892.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011070.hg.1 |
NM_001003892.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018086.hg.1 |
ENST00000338487.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018086.hg.1 |
NM_001003892.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10018086.hg.1 |
TC10001427.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000175497.hg.1 |
ENST00000338487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175497.hg.1 |
NM_001003892.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175497.hg.1 |
TC1000011070.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000175497.hg.1 |
NM_001003892.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175497.hg.1 |
ENST00000338487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011070.hg.1 |
ENST00000338487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011070.hg.1 |
ENST00000338487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011070.hg.1 |
ENST00000338487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011070.hg.1 |
NM_001003892.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011070.hg.1 |
NM_001003892.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011070.hg.1 |
NM_001003892.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175498.hg.1 |
ENST00000338487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175498.hg.1 |
NM_001003892.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175498.hg.1 |
TC1000011070.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000175498.hg.1 |
ENST00000338487.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175498.hg.1 |
NM_001003892.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175499.hg.1 |
TC1000011070.hg.1 |
Clariom D TC |