Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020022.hg.1 |
ENST00000194097.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020022.hg.1 |
ENST00000503719.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020022.hg.1 |
ENST00000508426.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020022.hg.1 |
ENST00000517649.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020022.hg.1 |
ENST00000523981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020022.hg.1 |
NM_004536.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020022.hg.1 |
NM_022892.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05020022.hg.1 |
TC05001455.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013322.hg.1 |
ENST00000512908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013322.hg.1 |
ENST00000512908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013322.hg.1 |
ENST00000512908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013322.hg.1 |
ENST00000512908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013322.hg.1 |
ENST00000512908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013322.hg.1 |
ENST00000512908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013322.hg.1 |
ENST00000512908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013322.hg.1 |
ENST00000512908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013322.hg.1 |
ENST00000512908.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013322.hg.1 |
NM_022892.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013322.hg.1 |
NM_022892.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013322.hg.1 |
NM_022892.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013322.hg.1 |
NM_022892.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013322.hg.1 |
NM_022892.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013322.hg.1 |
NM_022892.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013322.hg.1 |
NM_022892.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013322.hg.1 |
NM_022892.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013322.hg.1 |
NM_022892.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013322.hg.1 |
NM_022892.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013322.hg.1 |
NM_022892.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013322.hg.1 |
NM_022892.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013317.hg.1 |
ENST00000508650.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013317.hg.1 |
ENST00000508650.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013317.hg.1 |
ENST00000508650.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013317.hg.1 |
ENST00000508650.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013317.hg.1 |
ENST00000511830.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013317.hg.1 |
ENST00000511830.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013317.hg.1 |
ENST00000511830.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013317.hg.1 |
ENST00000511830.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013317.hg.1 |
ENST00000511830.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013317.hg.1 |
ENST00000511830.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013317.hg.1 |
ENST00000511830.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013317.hg.1 |
ENST00000511830.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013317.hg.1 |
ENST00000511830.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013317.hg.1 |
ENST00000511830.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013317.hg.1 |
ENST00000511830.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013317.hg.1 |
ENST00000511830.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013317.hg.1 |
NR_024054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013317.hg.1 |
NR_024054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013317.hg.1 |
NR_024054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013317.hg.1 |
NR_024054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013317.hg.1 |
NR_024054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013317.hg.1 |
NR_024054.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013317.hg.1 |
NR_024054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013322.hg.1 |
ENST00000512908.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013322.hg.1 |
ENST00000512908.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013322.hg.1 |
ENST00000512908.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013322.hg.1 |
ENST00000512908.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013322.hg.1 |
ENST00000512908.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013322.hg.1 |
ENST00000512908.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013322.hg.1 |
ENST00000512908.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013322.hg.1 |
ENST00000512908.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013322.hg.1 |
ENST00000512908.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013322.hg.1 |
NM_022892.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013322.hg.1 |
NM_022892.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013322.hg.1 |
NM_022892.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013322.hg.1 |
NM_022892.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013322.hg.1 |
NM_022892.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013322.hg.1 |
NM_022892.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013322.hg.1 |
NM_022892.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013322.hg.1 |
NM_022892.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013322.hg.1 |
NM_022892.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013322.hg.1 |
NM_022892.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013322.hg.1 |
NM_022892.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013322.hg.1 |
NM_022892.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013317.hg.1 |
ENST00000508650.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013317.hg.1 |
ENST00000508650.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013317.hg.1 |
ENST00000508650.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013317.hg.1 |
ENST00000508650.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013317.hg.1 |
ENST00000511830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013317.hg.1 |
ENST00000511830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013317.hg.1 |
ENST00000511830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013317.hg.1 |
ENST00000511830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013317.hg.1 |
ENST00000511830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013317.hg.1 |
ENST00000511830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013317.hg.1 |
ENST00000511830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013317.hg.1 |
ENST00000511830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013317.hg.1 |
ENST00000511830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013317.hg.1 |
ENST00000511830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013317.hg.1 |
ENST00000511830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013317.hg.1 |
ENST00000511830.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013317.hg.1 |
NR_024054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013317.hg.1 |
NR_024054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013317.hg.1 |
NR_024054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013317.hg.1 |
NR_024054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013317.hg.1 |
NR_024054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013317.hg.1 |
NR_024054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013317.hg.1 |
NR_024054.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500176785.hg.1 |
ENST00000194097.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500176785.hg.1 |
ENST00000315147.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500176785.hg.1 |
ENST00000503719.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500176785.hg.1 |
ENST00000508426.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500176785.hg.1 |
ENST00000517649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500176785.hg.1 |
ENST00000523981.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500176785.hg.1 |
TC0500011049.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500176785.hg.1 |
ENST00000503719.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500176785.hg.1 |
ENST00000194097.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500176785.hg.1 |
ENST00000315147.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500176785.hg.1 |
ENST00000508426.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500176785.hg.1 |
ENST00000517649.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500176785.hg.1 |
ENST00000523981.5 |
Ensembl |