Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03044957_g1
          See other RPL5 GE Assays ›
          Gene Symbol:
          RPL5
          Gene Name:
          ribosomal protein L5
          Gene Aliases:
          DBA6, L5, MSTP030, PPP1R135
          Chromosome Location:
          Chr.1: 92832037 - 92841924 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03044957_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RPL5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000969.3 NP_000960.2 4 - 5 401 149
          GenBank mRNA AB208980.1 - 4 - 5 344 149
          AF113210.1 - 4 - 5 380 149
          AK092963.1 - 3 - 4 1725 149
          AK095815.1 - 3 - 4 1696 149
          AK222604.1 - 4 - 5 365 149
          AK314720.1 - 4 - 5 398 149
          BC109370.1 - 4 - 5 385 5736022 149
          BC132970.1 - 4 - 5 370 40146992 149
          BC132972.1 - 4 - 5 370 40146994 149
          BG756697.1 - 4 - 5 386 149
          BU941919.1 - 4 - 5 402 149
          U14966.1 - 4 - 5 353 149

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100009046.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01000854.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RPL5

          Gene Ontology Categories:

          Function(s) Process(es)

          ribosomal large subunit assembly
          nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
          rRNA processing
          translation
          translational initiation
          SRP-dependent cotranslational protein targeting to membrane
          viral transcription
          ribosomal large subunit biogenesis
          RNA binding
          structural constituent of ribosome
          protein binding
          5S rRNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0500007599.hg.1 ENST00000509786.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500007599.hg.1 ENST00000509786.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01013588.hg.1 ENST00000315741.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01013588.hg.1 ENST00000370321.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01013588.hg.1 ENST00000432788.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01013588.hg.1 NM_000969.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01013588.hg.1 TC01000854.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01013591.hg.1 ENST00000315741.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01013591.hg.1 ENST00000370321.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01013591.hg.1 ENST00000432788.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01013591.hg.1 NM_000969.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01013591.hg.1 TC01000854.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1000012382.hg.1 ENST00000418634.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000012382.hg.1 ENST00000418634.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01013592.hg.1 ENST00000315741.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01013592.hg.1 ENST00000370321.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01013592.hg.1 ENST00000432788.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01013592.hg.1 NM_000969.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01013592.hg.1 TC01000854.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR01013589.hg.1 ENST00000315741.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01013589.hg.1 ENST00000370321.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01013589.hg.1 ENST00000432788.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01013589.hg.1 NM_000969.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01013589.hg.1 TC01000854.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000315741.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000315741.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000315741.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000315741.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000315741.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000315741.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000315741.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000370321.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000370321.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000370321.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000370321.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000370321.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000370321.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000370321.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000370321.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000370321.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000370321.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000370321.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000370321.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000383953.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000384792.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000461952.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000461952.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000461952.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000461952.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000461952.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000461952.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000461952.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000470843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000470843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000470843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000470843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000470843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000470843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000470843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000470843.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000497519.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000497519.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000497519.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000497519.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000497519.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000497519.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 NM_000969.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 NM_000969.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 NM_000969.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 NM_000969.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 NM_000969.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 NM_000969.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 NM_000969.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 NM_000969.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 NM_000969.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 NM_000969.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 NM_000969.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 NM_000969.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 NR_000006.8 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 NR_002444.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100009046.hg.1 ENST00000315741.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100164499.hg.1 ENST00000315741.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100164499.hg.1 ENST00000370321.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100164499.hg.1 ENST00000461952.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100164499.hg.1 ENST00000470843.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100164499.hg.1 NM_000969.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100164499.hg.1 TC0100009046.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100164499.hg.1 ENST00000470843.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100164499.hg.1 ENST00000315741.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100164499.hg.1 ENST00000370321.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100164499.hg.1 ENST00000461952.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100164499.hg.1 NM_000969.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100164500.hg.1 ENST00000315741.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100164500.hg.1 ENST00000370321.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100164500.hg.1 ENST00000461952.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100164500.hg.1 ENST00000470843.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100164500.hg.1 NM_000969.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100164500.hg.1 TC0100009046.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100164500.hg.1 ENST00000315741.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100164500.hg.1 ENST00000370321.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100164500.hg.1 ENST00000461952.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100164500.hg.1 ENST00000470843.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100164500.hg.1 NM_000969.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100164501.hg.1 ENST00000315741.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100164501.hg.1 ENST00000370321.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100164501.hg.1 ENST00000470843.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100164501.hg.1 NM_000969.4 Refseq
          Affymetrix Human Clariom D Assay PSR0100164501.hg.1 TC0100009046.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100164501.hg.1 ENST00000315741.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100164501.hg.1 ENST00000370321.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0100164501.hg.1 ENST00000470843.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0100164501.hg.1 NM_000969.4 Refseq
          Affymetrix Human Clariom D Assay TC1000012382.hg.1 ENST00000418634.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000012382.hg.1 ENST00000418634.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007599.hg.1 ENST00000509786.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500007599.hg.1 ENST00000509786.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000315741.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000315741.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000315741.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000315741.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000315741.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000315741.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000315741.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000370321.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000370321.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000370321.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000370321.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000370321.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000370321.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000370321.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000370321.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000370321.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000370321.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000370321.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000370321.7 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000383953.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000384792.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000461952.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000461952.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000461952.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000461952.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000461952.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000461952.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000461952.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000470843.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000470843.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000470843.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000470843.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000470843.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000470843.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000470843.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000470843.5 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000497519.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000497519.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000497519.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000497519.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000497519.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000497519.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 NM_000969.4 Refseq
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 NM_000969.4 Refseq
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 NM_000969.4 Refseq
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 NM_000969.4 Refseq
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 NM_000969.4 Refseq
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 NM_000969.4 Refseq
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 NM_000969.4 Refseq
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 NM_000969.4 Refseq
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 NM_000969.4 Refseq
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 NM_000969.4 Refseq
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 NM_000969.4 Refseq
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 NM_000969.4 Refseq
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 NR_000006.8 Refseq
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 NR_002444.2 Refseq
          Affymetrix Human Clariom D Assay TC0100009046.hg.1 ENST00000315741.5 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-ztd5s:80/100.66.134.6:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0