Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03045104_m1
          See other HLA-DPB1 GE Assays ›
          Gene Symbol:
          HLA-DPB1
          Gene Name:
          major histocompatibility complex, class II, DP beta 1
          Gene Aliases:
          DPB1, HLA-DP, HLA-DP1B, HLA-DPB
          Chromosome Location:
          Chr.6: 33075926 - 33089696 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03045104_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HLA-DPB1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002121.5 NP_002112.3 3 - 4 759 93
          GenBank mRNA AB208978.1 - 3 - 4 675 93
          AK291986.1 - 3 - 4 736 93
          AK313305.1 - 3 - 4 697 93
          AY656678.1 - 3 - 4 643 93
          AY656679.1 - 3 - 4 643 93
          AY656680.1 - 3 - 4 643 93
          AY804132.1 - 3 - 4 690 93
          AY804133.1 - 3 - 4 652 93
          AY804134.1 - 3 - 4 680 93
          AY804135.1 - 3 - 4 680 93
          AY804136.1 - 3 - 4 646 93
          AY804137.1 - 3 - 4 680 93
          AY804138.1 - 3 - 4 680 93
          AY804139.1 - 3 - 4 680 93
          AY804140.1 - 3 - 4 681 93
          AY804141.1 - 3 - 4 93
          AY804142.1 - 3 - 4 690 93
          AY804143.1 - 3 - 4 679 93
          AY831401.1 - 3 - 4 691 93
          AY831402.1 - 3 - 4 691 93
          AY831403.1 - 3 - 4 691 93
          AY831404.1 - 3 - 4 643 93
          BC007963.2 - 3 - 4 676 4309717 93
          BC013184.1 - 3 - 4 694 4341117 93
          BC015000.1 - 3 - 4 676 4851472 93
          BT019780.1 - 3 - 4 643 93
          DQ284441.1 - 3 - 4 678 93
          FR775797.1 - 3 - 4 643 93
          FR775798.1 - 2 - 3 543 93
          GQ891549.1 - 3 - 4 643 93
          HE603611.1 - 3 - 4 643 93
          HE603612.1 - 3 - 4 643 93
          HE614584.1 - 3 - 4 643 93
          HE614585.1 - 3 - 4 643 93
          HE614586.1 - 3 - 4 643 93
          HE614587.1 - 3 - 4 643 93
          HE614588.1 - 3 - 4 643 93
          HE614589.1 - 3 - 4 643 93
          HE614590.1 - 3 - 4 643 93
          HE614591.1 - 3 - 4 643 93
          J03041.1 - 3 - 4 712 93
          K00409.1 - 2 - 3 414 93
          K01615.1 - 2 - 3 541 93
          KJ780721.1 - 3 - 4 643 93
          L29173.1 - 2 - 3 400 93
          M21886.1 - 2 - 3 556 93
          M28200.1 - 3 - 4 662 93
          M28201.1 - 2 - 3 541 93
          M28202.1 - 3 - 4 619 93
          M57466.1 - 3 - 4 695 93
          M83664.1 - 3 - 4 702 93
          X00532.1 - 2 - 3 543 93
          X01426.1 - 3 - 4 712 93
          X03067.1 - 3 - 4 691 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600007677.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06000408.hg.1, TC6_apd_hap1000123.hg.1, TC6_cox_hap2000228.hg.1, TC6_dbb_hap3000091.hg.1, TC6_mann_hap4000213.hg.1, TC6_mcf_hap5000227.hg.1, TC6_qbl_hap6000210.hg.1, TC6_ssto_hap7000085.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene HLA-DPB1

          Gene Ontology Categories:

          Function(s) Process(es)

          antigen processing and presentation of exogenous peptide antigen via MHC class II
          T cell costimulation
          positive regulation of interferon-gamma production
          positive regulation of T cell proliferation
          T cell receptor signaling pathway
          positive regulation of T cell activation
          interferon-gamma-mediated signaling pathway
          peptide antigen binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR06004798.hg.1 ENST00000411942.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06004798.hg.1 ENST00000418931.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06004798.hg.1 ENST00000422592.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06004798.hg.1 NM_002121.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06004798.hg.1 TC06000408.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_mann_hap4001484.hg.1 TC6_mann_hap4000213.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_mann_hap4001484.hg.1 NM_002121.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_ssto_hap7001693.hg.1 TC6_ssto_hap7000085.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_ssto_hap7001693.hg.1 NM_002121.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_ssto_hap7001694.hg.1 TC6_ssto_hap7000085.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_ssto_hap7001694.hg.1 NM_002121.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06004797.hg.1 ENST00000411942.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06004797.hg.1 ENST00000418931.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06004797.hg.1 ENST00000422592.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06004797.hg.1 NM_002121.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06004797.hg.1 TC06000408.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2001874.hg.1 TC6_cox_hap2000228.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2001874.hg.1 NM_002121.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_dbb_hap3001862.hg.1 TC6_dbb_hap3000091.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_dbb_hap3001862.hg.1 NM_002121.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_mcf_hap5001657.hg.1 TC6_mcf_hap5000227.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_mcf_hap5001657.hg.1 NM_002121.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6001837.hg.1 TC6_qbl_hap6000210.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6001837.hg.1 NM_002121.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_apd_hap1000719.hg.1 TC6_apd_hap1000123.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_apd_hap1000719.hg.1 NM_002121.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_mcf_hap5001658.hg.1 TC6_mcf_hap5000227.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_mcf_hap5001658.hg.1 NM_002121.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_apd_hap1000718.hg.1 TC6_apd_hap1000123.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_apd_hap1000718.hg.1 NM_002121.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_mann_hap4001483.hg.1 TC6_mann_hap4000213.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_mann_hap4001483.hg.1 NM_002121.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000416804.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000416804.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000416804.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000416804.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000416804.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000418931.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000418931.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000418931.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000418931.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000418931.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000418931.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000418931.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000418931.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000428835.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000428835.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000428835.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000428835.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000469120.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000469120.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000469120.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000469120.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000471184.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000471184.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000471184.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000471184.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000471184.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000471184.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000478189.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000478189.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000478189.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000488575.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000488575.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000488575.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000488575.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000498038.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000498038.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000498038.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000498038.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 ENST00000498038.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 NM_002121.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 NM_002121.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 NM_002121.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 NM_002121.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 NM_002121.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 NM_002121.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 NM_002121.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 NM_002121.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007677.hg.1 NM_002121.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06004800.hg.1 ENST00000411942.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06004800.hg.1 ENST00000418931.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06004800.hg.1 ENST00000422592.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR06004800.hg.1 NM_002121.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06004800.hg.1 TC06000408.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2001875.hg.1 TC6_cox_hap2000228.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2001875.hg.1 NM_002121.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_dbb_hap3001863.hg.1 TC6_dbb_hap3000091.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_dbb_hap3001863.hg.1 NM_002121.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6001838.hg.1 TC6_qbl_hap6000210.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6001838.hg.1 NM_002121.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007679.hg.1 ENST00000435074.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007679.hg.1 ENST00000435074.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007679.hg.1 ENST00000435074.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007679.hg.1 ENST00000435074.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007679.hg.1 ENST00000470997.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007679.hg.1 ENST00000470997.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007679.hg.1 ENST00000470997.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007679.hg.1 ENST00000470997.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600153062.hg.1 ENST00000416804.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600153062.hg.1 ENST00000418931.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600153062.hg.1 ENST00000428835.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600153062.hg.1 NM_002121.5 Refseq
          Affymetrix Human Clariom D Assay PSR0600153062.hg.1 TC0600007677.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600153062.hg.1 ENST00000416804.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600153062.hg.1 ENST00000418931.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0600153062.hg.1 ENST00000428835.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0600153062.hg.1 NM_002121.5 Refseq
          Affymetrix Human Clariom D Assay TC0600007679.hg.1 ENST00000435074.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007679.hg.1 ENST00000435074.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007679.hg.1 ENST00000435074.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007679.hg.1 ENST00000435074.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007679.hg.1 ENST00000470997.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007679.hg.1 ENST00000470997.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007679.hg.1 ENST00000470997.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007679.hg.1 ENST00000470997.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000416804.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000416804.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000416804.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000416804.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000416804.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000418931.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000418931.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000418931.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000418931.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000418931.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000418931.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000418931.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000418931.6 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000428835.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000428835.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000428835.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000428835.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000469120.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000469120.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000469120.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000469120.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000471184.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000471184.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000471184.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000471184.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000471184.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000471184.5 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000478189.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000478189.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000478189.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000488575.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000488575.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000488575.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000488575.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000498038.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000498038.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000498038.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000498038.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 ENST00000498038.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 NM_002121.5 Refseq
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 NM_002121.5 Refseq
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 NM_002121.5 Refseq
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 NM_002121.5 Refseq
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 NM_002121.5 Refseq
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 NM_002121.5 Refseq
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 NM_002121.5 Refseq
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 NM_002121.5 Refseq
          Affymetrix Human Clariom D Assay TC0600007677.hg.1 NM_002121.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc