Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03296927_s1
          See other PCDHB17P GE Assays ›
          Gene Symbol:
          PCDHB17P
          Gene Name:
          protocadherin beta 17 pseudogene
          Gene Aliases:
          ME4, PCDH-psi1, PCDHB17
          Chromosome Location:
          Chr.5: 141155999 - 141158409 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03296927_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PCDHB17P
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_001280.1 1 2349 108
          GenBank mRNA AF217751.1 - 1 2352 108
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AF152527.3 - 1 2349 Chr. 5 - 141155999 - 141158409 [+] on Build GRCh38 108

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream protocadherin beta 6 PCDHB6 PCDH-BETA6 -2.7kb Sense Chr. 5 - 141150022 - 141153287 on NCBI Build GRCh38
          Downstream protocadherin beta 7 PCDHB7 PCDH-BETA7 14.2kb Sense Chr. 5 - 141172621 - 141176376 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 35924 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 347528 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 109586 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 333435 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 253562 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 302796 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 224551 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P271773 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P271773 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 15619 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 45227 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P01351231 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P271773 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P271773 - -
          SurePrint G3 Human GE Microarray G4851A A_19_P00805852 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24504686 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326265 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24504689 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24504668 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24504674 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24504677 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326278 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326263 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326285 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326280 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326275 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24504682 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326292 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24504691 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326268 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24504673 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326291 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24504680 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326282 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24504668 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326280 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24504677 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24504682 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24504673 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326285 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326265 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326292 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326291 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326278 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326268 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24504691 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24504686 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24504674 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24504689 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326282 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326263 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326275 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24504680 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P271773 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P01351231 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P271773 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P31595704 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01351231 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326239 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326323 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326248 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326292 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326231 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326308 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326320 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326233 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326285 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326326 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326298 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326267 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326283 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326223 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326216 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326277 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326221 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326235 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326272 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326264 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326246 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326250 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326242 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326289 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326291 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326226 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326275 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326301 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326305 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326253 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326316 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326236 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326229 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326279 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326224 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326240 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326219 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326315 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326296 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326287 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326262 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326293 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326247 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326269 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326312 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326294 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326318 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P01351231 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NR_001280_at - -
          Affymetrix Human Gene 1.0 ST Array 8108707 - -
          Affymetrix Human Gene 1.1 ST Array 8108706 - -
          Affymetrix HT HG-U133+ PM Array Plate 216355_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 216313_PM_at - -
          Affymetrix Human Genome U219 Array 11745339_at - -
          Affymetrix Human Genome U219 Array 11745339_PM_at - -
          Affymetrix Human Genome U219 Array 11759954_PM_at - -
          Affymetrix Human Genome U219 Array 11759954_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0500008878.hg.1 ENST00000539533.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008878.hg.1 ENST00000623466.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008878.hg.1 ENST00000623466.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008878.hg.1 NR_001280.1 Refseq
          Affymetrix Human Clariom D Assay TC0500008878.hg.1 ENST00000539533.3 Ensembl
          Affymetrix Human Clariom D Assay TC0500008878.hg.1 ENST00000623466.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500008878.hg.1 ENST00000623466.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500008878.hg.1 NR_001280.1 Refseq
          Human Genome U133 Plus 2.0 Array 216355_at - -
          Human Genome U133 Plus 2.0 Array 216313_PM_at - -
          Human Genome U133 Plus 2.0 Array 216355_PM_at - -
          Human Genome U133 Plus 2.0 Array 216313_at - -
          Human Genome U133A 2.0 Array 216355_PM_at - -
          Human Genome U133A 2.0 Array 216313_at - -
          Human Genome U133A 2.0 Array 216355_at - -
          Human Genome U133A 2.0 Array 216313_PM_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NR_001280_at - -
          AffymetrixPathwork Diagnostics Pathchip pw509449 - -
          AffymetrixPathwork Diagnostics Pathchip pw545106 - -
          Human Genome U95 Set 1168_PM_at - -
          Human Genome U95 Set 1168_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216313_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216355_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216313_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216313_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216313_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216355_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216355_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216313_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216313_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216313_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216313_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216355_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216313_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216355_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216355_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216355_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216313_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216355_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216355_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216355_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216355_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 216313_at8 - -
          Human Genome U133 Set 216313_PM_at - -
          Human Genome U133 Set 216355_PM_at - -
          Human Genome U133 Set 216313_at - -
          Human Genome U133 Set 216355_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A028925 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A028925 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 218372 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_2185866 NR_001280.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2185866 AF217751.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1750376 NR_001280.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1750376 AF217751.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1750376 NR_001280.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1750376 AF217751.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2185866 NR_001280.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2185866 AF217751.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1750376 NR_001280.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1750376 AF217751.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2185866 NR_001280.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2185866 AF217751.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1750376 NR_001280.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1750376 AF217751.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2185866 NR_001280.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2185866 AF217751.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1750376 NR_001280.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1750376 AF217751.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1750376 NR_001280.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1750376 AF217751.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip ILMN_4433 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_4433 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2185866 NR_001280.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2185866 AF217751.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1750376 NR_001280.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1750376 AF217751.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2185866 NR_001280.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2185866 AF217751.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1750376 NR_001280.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1750376 AF217751.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2185866 NR_001280.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2185866 AF217751.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1750376 NR_001280.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1750376 AF217751.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2185866 NR_001280.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2185866 AF217751.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0