Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03296951_s1
          See other PCDHB18P GE Assays ›
          Gene Symbol:
          PCDHB18P
          Gene Name:
          protocadherin beta 18 pseudogene
          Gene Aliases:
          PCDH-psi2, PCDHB18
          Chromosome Location:
          Chr.5: 141234333 - 141237529 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03296951_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PCDHB18P
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_001281.2 1 2884 148
          GenBank mRNA AK095496.1 - 1 2853 148
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DB314968.1 - 1 Chr. 5 - 141237013 - 141237529 [-] on Build GRCh38 148
          GenBankmRNA DB310039.1 - 1 Chr. 5 - 141236995 - 141237522 [-] on Build GRCh38 148
          GenBankmRNA BE326289.1 - 1 195 Chr. 5 - 141237022 - 141237440 [+] on Build GRCh38 148

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream protocadherin beta 14 PCDHB14 PCDH-BETA14 -8.0kb Sense Chr. 5 - 141222326 - 141226288 on NCBI Build GRCh38
          Downstream protocadherin beta 19 pseudogene PCDHB19P PCDH-PSI5,
          PCDHB19
          2.6kb Sense Chr. 5 - 141240121 - 141244752 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          homophilic cell adhesion via plasma membrane adhesion molecules
          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P390958 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_23_P390958 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 24221 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 61354 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_23_P390958 - -
          SurePrint G3 Human GE Microarray G4851A A_23_P390958 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505126 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505115 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505102 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326594 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505108 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505113 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505111 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326611 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505098 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505109 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505147 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326585 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505103 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505114 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505112 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326609 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505150 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505128 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505133 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505136 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505105 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326596 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505149 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505101 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326586 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326603 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505096 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505146 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505121 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326575 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505122 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505114 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326586 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505146 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505108 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505150 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505112 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326609 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326575 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505098 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505136 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505149 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326594 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326596 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505126 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505113 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326611 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505128 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505109 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505096 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505121 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326603 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505105 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505102 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505115 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505101 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505111 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505122 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505103 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04326585 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505147 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24505133 - -
          Whole Human Genome Oligo Microarray G4112A A_23_P390958 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P17324933 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_23_P390958 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P17324933 - -
          Agilent Human 1B Microarray G4111A A_32_P357047 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326659 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326610 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326665 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326683 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326570 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326658 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326656 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326678 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326626 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326628 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326685 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326638 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326689 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326666 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326615 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326588 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326673 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326582 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326675 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326598 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326669 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326651 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326661 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326663 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326603 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15747294 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326618 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326635 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326676 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326586 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326579 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326631 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326648 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326600 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326567 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326684 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326620 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326591 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326576 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326616 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326653 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326589 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326652 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326608 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326634 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326622 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326578 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326596 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326594 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326613 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326574 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326671 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326584 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326573 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04326680 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P17324933 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NR_001281_at - -
          Affymetrix Human Gene 1.0 ST Array 8108751 - -
          Affymetrix Human Gene 1.0 ST Array 8108750 - -
          Affymetrix Human Gene 1.0 ST Array 8108752 - -
          Affymetrix Human Gene 1.1 ST Array 8108749 - -
          Affymetrix HT HG-U133+ PM Array Plate 234479_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 234724_PM_x_at - -
          Affymetrix Human Genome U219 Array 11746318_at - -
          Affymetrix Human Genome U219 Array 11746318_PM_at - -
          Human Genome U133 Plus 2.0 Array 234724_PM_x_at - -
          Human Genome U133 Plus 2.0 Array 234479_PM_at - -
          Human Genome U133 Plus 2.0 Array 234479_at - -
          Human Genome U133 Plus 2.0 Array 234724_x_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NR_001281_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234479_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234724_x_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234479_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234479_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234724_x_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234479_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234479_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234724_x_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234724_x_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234479_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234479_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234479_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234479_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234724_x_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234479_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234724_x_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234724_x_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234724_x_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234724_x_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234479_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234724_x_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 234724_x_at4 - -
          Human Genome U133 Set 234479_at - -
          Human Genome U133 Set 234724_PM_x_at - -
          Human Genome U133 Set 234479_PM_at - -
          Human Genome U133 Set 234724_x_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A028936 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A028935 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A028935 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A028936 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 544276 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_2170476 AF217743.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2170476 AK095496.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2170476 NR_001281.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1773040 - -
          HumanHT-12 V4.0 expression beadchip ILMN_2170476 AF217743.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2170476 AK095496.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2170476 NR_001281.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2170476 AF217743.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2170476 AK095496.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2170476 NR_001281.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2170476 AF217743.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2170476 AK095496.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2170476 NR_001281.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1773040 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 7160301 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_4529 - -
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2170476 AF217743.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2170476 AK095496.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2170476 NR_001281.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2170476 AF217743.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2170476 AK095496.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2170476 NR_001281.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2170476 AF217743.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2170476 AK095496.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2170476 NR_001281.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1773040 - -
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2170476 AF217743.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2170476 AK095496.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2170476 NR_001281.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0