Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03298478_s1
          See other KCNIP4-IT1 GE Assays ›
          Gene Symbol:
          KCNIP4-IT1
          Gene Name:
          KCNIP4 intronic transcript 1
          Gene Aliases:
          NCRNA00099, UM9(5), UM9-5
          Chromosome Location:
          Chr.4: 21843341 - 21853188 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03298478_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: KCNIP4-IT1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002813.1 1 9245 149
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BK000166.3 - 1 9245 Chr. 4 - 21843341 - 21853188 [-] on Build GRCh38 149

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC105374516 LOC105374516 -229.6kb Anti-sense Chr. 4 - 21582096 - 21613722 on NCBI Build GRCh38
          Downstream uncharacterized LOC100505912 LOC100505912 474.2kb Sense Chr. 4 - 22327354 - 22339666 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3352906 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3894 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3895 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 17071 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3352906 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3352906 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126783 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126791 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126823 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126794 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126782 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02973896 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02973924 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126822 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126785 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02973922 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126820 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02973901 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126776 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126816 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126810 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02973942 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02973929 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126806 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02973931 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126779 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126773 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126781 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126777 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02973905 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126818 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126815 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126807 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126800 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02973897 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02973914 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126812 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126817 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126786 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126805 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126809 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126789 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126772 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126780 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15551412 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126801 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126792 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126807 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126823 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126783 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02973929 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126812 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126801 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126822 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02973897 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126789 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126820 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15551412 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126806 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126805 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126817 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126773 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126794 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126780 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126810 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126781 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126800 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02973914 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126816 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02973922 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126776 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02973931 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02973924 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126777 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126791 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126772 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02973942 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126792 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126818 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126786 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126782 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02973905 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126809 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126785 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02973896 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126779 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P02973901 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23126815 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15551401 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973910 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973923 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973868 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973948 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973928 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973854 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973926 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973895 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973908 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15551402 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973898 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973953 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973891 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15551398 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973856 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973860 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973952 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973939 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973885 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15551399 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973944 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973862 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15551408 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973932 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15551411 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973869 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15551405 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973848 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973912 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973942 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15551412 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15551403 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973864 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973919 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973896 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973930 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973889 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973921 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973866 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973850 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973936 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973917 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973916 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973893 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973903 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973887 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973873 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973871 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973900 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973950 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973925 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15551396 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973876 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973901 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P02973906 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Genome U219 Array 11757234_PM_at - -
          Affymetrix Human Genome U219 Array 11757234_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A024012 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A024011 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A024012 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A024011 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3242530 NR_002813.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3242530 NR_002813.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3242530 NR_002813.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3242530 NR_002813.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0