Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03298625_s1
          See other TOB2P1 GE Assays ›
          Gene Symbol:
          TOB2P1
          Gene Name:
          transducer of ERBB2, 2 pseudogene 1
          Gene Aliases:
          TOB2P, TOB4p, p373c6.3
          Chromosome Location:
          Chr.6: 28215338 - 28218929 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03298625_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TOB2P1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002936.2 1 3441 132
          GenBank mRNA BC050562.1 - 1 3446 132
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AI424877.1 - 1 Chr. 6 - 28215338 - 28215762 [+] on Build GRCh38 132
          GenBankmRNA AW295192.1 - 1 Chr. 6 - 28215338 - 28215655 [+] on Build GRCh38 132

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream zinc finger protein 192 pseudogene 1 ZNF192P1 ZNF389,
          ZNF389P,
          dJ265C24.4
          -45.7kb Anti-sense Chr. 6 - 28161761 - 28169595 on NCBI Build GRCh38
          Downstream zinc finger with KRAB and SCAN domains 4 ZKSCAN4 P1P373C6,
          ZNF307,
          ZNF427,
          ZSCAN36
          22.8kb Sense Chr. 6 - 28241697 - 28259312 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600011246.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06001436.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24822030 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24822023 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04631787 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04631793 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04631818 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24822044 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24822013 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24822017 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04631848 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24822046 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04631847 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24821992 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24821989 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24822045 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04631806 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24822042 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04631829 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04631821 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04631804 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04631807 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24822038 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24821991 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04631822 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24822021 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24822019 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24822014 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04631792 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24822028 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24821988 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04631823 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24821999 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04631837 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24821993 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24822041 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04631798 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04631833 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24822015 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24822037 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04631825 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24822018 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24822003 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04631817 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24822006 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24821996 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04631828 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04631834 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24822049 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24821997 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04631797 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 643258 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 421595 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3348011 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P440095 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 4924 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 44117 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3348011 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3348011 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24822059 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631864 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631881 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631859 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24822070 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24822069 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631870 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631871 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24822067 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631862 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631873 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631876 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24822079 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631878 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631874 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24822082 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24822081 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24822063 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24822059 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631881 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631876 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24822063 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631862 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24822082 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631871 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24822070 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24822069 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24822079 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631874 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24822067 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631859 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631878 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631873 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631864 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04631870 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P24822081 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P440095 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P17490512 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_32_P440095 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01447682 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P17490512 - -
          Agilent Human 1B Microarray G4111A A_32_P440095 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631919 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631859 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631818 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631816 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631906 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15796790 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631827 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631923 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15796775 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631795 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631835 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631907 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631893 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631831 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631856 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631804 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15796787 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631822 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631793 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631789 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631844 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631903 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631921 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631809 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631781 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631891 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631909 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631851 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631895 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631842 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631783 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631840 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631876 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631862 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631800 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631824 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631910 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631853 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631846 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631881 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631810 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631880 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631913 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631883 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631798 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631901 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631878 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631904 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631870 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631820 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631785 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631889 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631852 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04631837 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P17490512 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NR_002936_at - -
          Affymetrix Human Gene 1.0 ST Array 8124552 - -
          Affymetrix Human Gene 1.1 ST Array 8124551 - -
          Affymetrix Human Genome U219 Array 11746349_at - -
          Affymetrix Human Genome U219 Array 11746349_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR06016385.hg.1 NR_002936.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06016385.hg.1 TC06001436.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0600175836.hg.1 ENST00000469761.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600175836.hg.1 NR_002936.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600175836.hg.1 TC0600011246.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600175836.hg.1 NR_002936.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600175836.hg.1 ENST00000469761.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0600175835.hg.1 NR_002936.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600175835.hg.1 TC0600011246.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600175835.hg.1 NR_002936.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600175837.hg.1 NR_002936.2 Refseq
          Affymetrix Human Clariom D Assay PSR0600175837.hg.1 TC0600011246.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600175837.hg.1 NR_002936.2 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A029585 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A029584 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A029585 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A029584 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 230432 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1657832 BC039027.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1657832 BC050562.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1657832 NR_002936.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1657832 BC015543.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1657832 BC039027.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1657832 BC050562.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1657832 NR_002936.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1657832 BC015543.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1657832 BC039027.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1657832 BC050562.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1657832 NR_002936.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1657832 BC015543.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1657832 BC039027.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1657832 BC050562.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1657832 NR_002936.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1657832 BC015543.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1657832 BC039027.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1657832 BC050562.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1657832 NR_002936.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1657832 BC015543.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1657832 BC039027.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1657832 BC050562.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1657832 NR_002936.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1657832 BC015543.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1657832 BC039027.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1657832 BC050562.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1657832 NR_002936.2 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1657832 BC015543.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0