Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03298802_s1
          See other SNORD89 GE Assays ›
          Gene Symbol:
          SNORD89 RNF149
          Gene Name:
          small nucleolar RNA, C/D box 89
          ring finger protein 149
          Gene Aliases:
          HBII-289
          DNAPTP2
          Chromosome Location:
          Chr.2: 101272936 - 101273049 on Build GRCh38
          Chromosome Location:
          Chr.2: 101271213 - 101308716 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs03298802_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SNORD89
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003070.1 1 57 72
          GenBank mRNA AK126261.1 - 1 448 72
          Gene Symbol: RNF149
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_005263921.3 XP_005263978.1 7 1390 72
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AW161820.1 - 1 Chr. 2 - 101272583 - 101273070 [+] on Build GRCh38 72
          GenBankmRNA BU164391.1 - 1 41 Chr. 2 - 101272198 - 101273033 [-] on Build GRCh38 72
          GenBankmRNA DN949208.1 - 1 38 Chr. 2 - 101272782 - 101273030 [-] on Build GRCh38 72
          GenBankmRNA BM311163.1 - 1 52 Chr. 2 - 101272554 - 101273034 [-] on Build GRCh38 72
          GenBankmRNA DB193060.1 - 1 448 Chr. 2 - 101272863 - 101273440 [-] on Build GRCh38 72
          GenBankmRNA BM457579.1 - 54 Chr. - - [-] on Build 72
          GenBankmRNA BF893393.1 - 1 Chr. 2 - 101272831 - 101273209 [+] on Build GRCh38 72
          GenBankmRNA BU942069.1 - 1 56 Chr. 2 - 101272306 - 101273046 [-] on Build GRCh38 72
          GenBankmRNA BU198638.1 - 57 Chr. - - [-] on Build 72

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream CCR4-NOT transcription complex subunit 11 CNOT11 C2orf29,
          C40
          -2.6kb Anti-sense Chr. 2 - 101252883 - 101270316 on NCBI Build GRCh38
          Downstream microRNA 5696 MIR5696 36.4kb Anti-sense Chr. 2 - 101309450 - 101309534 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200016719.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02002145.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          protein ubiquitination
          regulation of protein stability
          negative regulation of MAPK cascade
          cellular response to xenobiotic stimulus
          zinc ion binding
          ligase activity
          ubiquitin protein ligase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 493556 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4092 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21465428 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01361950 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21465425 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01361959 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01361960 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21465430 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01361942 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01361958 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21465427 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21465438 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01361952 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21465422 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01361956 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21465421 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21465437 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21465446 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21465429 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21465441 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21465425 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21465437 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21465430 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01361960 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21465421 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01361942 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21465441 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21465427 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01361952 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21465446 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21465428 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21465438 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21465429 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01361950 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01361956 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01361959 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21465422 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01361958 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15759165 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01361941 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01361957 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01361955 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01361961 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01361958 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15292714 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01361949 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01361945 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01361964 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01361952 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01361947 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15292711 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15292710 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 235536_PM_at - -
          Affymetrix Human Genome U219 Array 11757177_PM_s_at - -
          Affymetrix Human Genome U219 Array 11757177_s_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR02035049.hg.1 ENST00000390981.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02035049.hg.1 NR_003070.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02035049.hg.1 TC02002145.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0200016719.hg.1 ENST00000390981.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016719.hg.1 ENST00000485752.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016719.hg.1 ENST00000485752.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200016719.hg.1 NR_003070.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200227593.hg.1 ENST00000390981.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200227593.hg.1 NR_003070.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200227593.hg.1 TC0200016719.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200227593.hg.1 NR_003070.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200227593.hg.1 ENST00000390981.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016719.hg.1 ENST00000390981.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016719.hg.1 ENST00000485752.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016719.hg.1 ENST00000485752.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200016719.hg.1 NR_003070.1 Refseq
          Human Genome U133 Plus 2.0 Array 235536_at - -
          Human Genome U133 Plus 2.0 Array 235536_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235536_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235536_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235536_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235536_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235536_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235536_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235536_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235536_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235536_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235536_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235536_at8 - -
          Human Genome U95 Set 63906_PM_f_at - -
          Human Genome U95 Set 63437_PM_r_at - -
          Human Genome U95 Set 57770_PM_at - -
          Human Genome U95 Set 63705_PM_f_at - -
          Human Genome U95 Set 63705_f_at - -
          Human Genome U95 Set 63437_r_at - -
          Human Genome U95 Set 63436_s_at - -
          Human Genome U95 Set 63436_PM_s_at - -
          Human Genome U95 Set 57770_at - -
          Human Genome U95 Set 63906_f_at - -
          Human Genome U95 Set 70957_r_at - -
          Human Genome U95 Set 70957_PM_r_at - -
          Human Genome U133 Set 235536_at - -
          Human Genome U133 Set 235536_PM_at - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3238662 NR_003070.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3238662 AK126261.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3238662 NR_003070.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3238662 AK126261.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3238662 NR_003070.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3238662 AK126261.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3238662 NR_003070.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3238662 AK126261.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0