Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03298806_s1
          See other SNORD92 GE Assays ›
          Gene Symbol:
          SNORD92
          Gene Name:
          small nucleolar RNA, C/D box 92
          Gene Aliases:
          HBII-316
          Chromosome Location:
          Chr.2: 28913662 - 28913750 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03298806_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SNORD92
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003074.1 1 42 80

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream tRNA methyltransferase 61B TRMT61B -43.4kb Anti-sense Chr. 2 - 28849821 - 28870309 on NCBI Build GRCh38
          Downstream small nucleolar RNA,
          C/D box 53
          SNORD53 RNU53,
          SNORD53A,
          U53
          13.3kb Sense Chr. 2 - 28927067 - 28927142 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200007112.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02000188.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4094 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150591 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150606 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150594 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150581 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150615 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150585 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150584 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150586 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150612 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01071041 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150596 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01071040 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01071062 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150600 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150573 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01071047 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01071050 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150597 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150607 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01071057 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150589 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01071056 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150605 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01071028 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01071034 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01071025 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01071059 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150603 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01071065 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01071065 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150573 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01071056 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150585 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150605 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150594 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01071028 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150591 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01071041 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150586 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150603 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150612 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150597 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01071057 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01071034 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150589 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150607 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150584 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01071025 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150581 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01071059 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01071050 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150615 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150606 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150596 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01071047 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01071062 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P01071040 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P21150600 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01071122 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01071106 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01071104 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01071049 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01071059 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01071028 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01071108 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01071052 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01071113 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01071060 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01071033 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01071063 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01071031 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01071117 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01071047 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01071041 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01071025 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01071111 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01071021 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01071038 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01071121 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01071054 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01071036 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01071056 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01071044 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P01071119 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR02002745.hg.1 NR_003074.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02002745.hg.1 TC02000188.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0200007112.hg.1 ENST00000585078.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200007112.hg.1 NR_003074.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200149041.hg.1 ENST00000585078.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200149041.hg.1 NR_003074.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200149041.hg.1 TC0200007112.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200149041.hg.1 NR_003074.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200149041.hg.1 ENST00000585078.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200007112.hg.1 ENST00000585078.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200007112.hg.1 NR_003074.1 Refseq
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3245759 NR_003074.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3245759 NR_003074.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3245759 NR_003074.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3245759 NR_003074.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7f58f9545f-46b6w:80/100.66.130.62:80.
          git-commit: c36c52929dfaa9f2956040e6abd275853d6e1b95
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline