Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03299045_g1
          See other SNHG9 GE Assays ›
          Gene Symbol:
          SNHG9
          Gene Name:
          small nucleolar RNA host gene 9
          Gene Aliases:
          NCRNA00062
          Chromosome Location:
          Chr.16: 1964996 - 1965504 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03299045_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SNHG9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003142.2 137 76
          GenBank mRNA CR749453.1 - 138 76
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AW958849.1 - Chr. - - [-] on Build 76
          GenBankmRNA AV762892.1 - 116 Chr. - - [-] on Build 76
          GenBankmRNA AI830540.1 - 1 - 2 Chr. 16 - 1964959 - 1965502 [-] on Build GRCh38 76
          GenBankmRNA AI335153.1 - 1 - 2 67 Chr. 16 - 1965066 - 1965505 [+] on Build GRCh38 76
          GenBankmRNA AI862260.1 - 1 - 2 69 Chr. 16 - 1965068 - 1965501 [+] on Build GRCh38 76
          GenBankmRNA CB110750.1 - 1 - 2 102 Chr. 16 - 1965031 - 1965508 [+] on Build GRCh38 76
          GenBankmRNA BI963236.1 - 1 - 2 Chr. 16 - 1965067 - 1965501 [-] on Build GRCh38 76
          GenBankmRNA BF131329.1 - 108 Chr. - - [-] on Build 76
          GenBankmRNA BU960491.1 - 1 - 2 77 Chr. 16 - 1965059 - 1965504 [+] on Build GRCh38 76
          GenBankmRNA AW157547.1 - 1 - 2 Chr. 16 - 1965074 - 1965508 [-] on Build GRCh38 76
          GenBankmRNA AA516122.1 - 1 - 2 Chr. 16 - 1964994 - 1965509 [-] on Build GRCh38 76
          GenBankmRNA CD174859.1 - 1 - 2 129 Chr. 16 - 1965028 - 1965504 [+] on Build GRCh38 76
          GenBankmRNA BI966761.1 - 1 - 2 Chr. 16 - 1965022 - 1965504 [-] on Build GRCh38 76
          GenBankmRNA DW442228.1 - Chr. - - [-] on Build 76
          GenBankmRNA AA357636.1 - 1 - 2 Chr. 16 - 1964979 - 1965502 [+] on Build GRCh38 76
          GenBankmRNA AI670727.1 - 1 - 2 Chr. 16 - 1964992 - 1965502 [-] on Build GRCh38 76
          GenBankmRNA DW438767.1 - 1 - 2 Chr. 16 - 1965025 - 1965499 [+] on Build GRCh38 76
          GenBankmRNA BX096205.1 - 1 - 2 173 Chr. 16 - 1964960 - 1965509 [+] on Build GRCh38 76
          GenBankmRNA BU587988.1 - Chr. - - [-] on Build 76
          GenBankmRNA BI962931.1 - 1 - 2 Chr. 16 - 1964992 - 1965504 [+] on Build GRCh38 76
          GenBankmRNA BU849390.1 - 1 - 2 107 Chr. 16 - 1965026 - 1965504 [+] on Build GRCh38 76
          GenBankmRNA AA661893.1 - 1 - 2 Chr. 16 - 1965013 - 1965508 [+] on Build GRCh38 76
          GenBankmRNA DW437557.1 - Chr. - - [-] on Build 76
          GenBankmRNA BI964920.1 - 1 - 2 97 Chr. 16 - 1965036 - 1965504 [+] on Build GRCh38 76
          GenBankmRNA DW441990.1 - 91 Chr. - - [-] on Build 76
          GenBankmRNA BG940943.1 - 1 - 2 Chr. 16 - 1965016 - 1965508 [-] on Build GRCh38 76
          GenBankmRNA BF217948.1 - 1 - 2 Chr. 16 - 1965016 - 1965497 [+] on Build GRCh38 76
          GenBankmRNA BU565055.1 - 1 - 2 144 Chr. 16 - 1964992 - 1965504 [+] on Build GRCh38 76

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream small nucleolar RNA,
          H/ACA box 78
          SNORA78 ACA64 Anti-sense Chr. 16 - 1965184 - 1965310 on NCBI Build GRCh38
          Downstream ring finger protein 151 RNF151 1.3kb Anti-sense Chr. 16 - 1966639 - 1968975 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1600006576.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC16000051.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3280965 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4113 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4114 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 45617 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3280965 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3280965 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09888609 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09888610 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09888609 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09888610 - -
          Agilent Human 1B Microarray G4111A A_32_P84349 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16841461 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09888610 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09888587 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16841490 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09888606 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 228645_PM_at - -
          Affymetrix Human Genome U219 Array 11763697_s_at - -
          Affymetrix Human Genome U219 Array 11763697_PM_s_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR16001123.hg.1 ENST00000531523.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16001123.hg.1 NR_003142.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16001123.hg.1 TC16000051.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR16001122.hg.1 ENST00000531523.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR16001122.hg.1 NR_003142.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR16001122.hg.1 TC16000051.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1600006576.hg.1 ENST00000459373.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006576.hg.1 ENST00000531523.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006576.hg.1 ENST00000531523.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006576.hg.1 ENST00000531523.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006576.hg.1 ENST00000531523.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006576.hg.1 ENST00000564014.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006576.hg.1 ENST00000564014.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006576.hg.1 ENST00000564014.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006576.hg.1 ENST00000564014.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006576.hg.1 ENST00000564014.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1600006576.hg.1 NR_003020.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600006576.hg.1 NR_003142.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600006576.hg.1 NR_003142.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1600006576.hg.1 NR_003142.2 Refseq
          Affymetrix Human Clariom D Assay TC1600006576.hg.1 ENST00000459373.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006576.hg.1 ENST00000531523.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006576.hg.1 ENST00000531523.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006576.hg.1 ENST00000531523.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006576.hg.1 ENST00000531523.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006576.hg.1 ENST00000564014.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006576.hg.1 ENST00000564014.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006576.hg.1 ENST00000564014.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006576.hg.1 ENST00000564014.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006576.hg.1 ENST00000564014.1 Ensembl
          Affymetrix Human Clariom D Assay TC1600006576.hg.1 NR_003020.1 Refseq
          Affymetrix Human Clariom D Assay TC1600006576.hg.1 NR_003142.2 Refseq
          Affymetrix Human Clariom D Assay TC1600006576.hg.1 NR_003142.2 Refseq
          Affymetrix Human Clariom D Assay TC1600006576.hg.1 NR_003142.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600146013.hg.1 ENST00000531523.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600146013.hg.1 NR_003142.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600146013.hg.1 TC1600006576.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600146013.hg.1 NR_003142.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600146013.hg.1 ENST00000531523.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600146015.hg.1 ENST00000531523.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600146015.hg.1 ENST00000564014.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600146015.hg.1 NR_003142.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600146015.hg.1 TC1600006576.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600146015.hg.1 ENST00000531523.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600146015.hg.1 ENST00000564014.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600146015.hg.1 NR_003142.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600146020.hg.1 ENST00000531523.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600146020.hg.1 ENST00000564014.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600146020.hg.1 NR_003142.2 Refseq
          Affymetrix Human Clariom D Assay PSR1600146020.hg.1 TC1600006576.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1600146020.hg.1 ENST00000531523.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600146020.hg.1 ENST00000564014.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1600146020.hg.1 NR_003142.2 Refseq
          Human Genome U133 Plus 2.0 Array 228645_PM_at - -
          Human Genome U133 Plus 2.0 Array 228645_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228645_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228645_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228645_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228645_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228645_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228645_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228645_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228645_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228645_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228645_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 228645_at8 - -
          Human Genome U95 Set 89811_PM_at - -
          Human Genome U95 Set 89811_at - -
          Human Genome U133 Set 228645_PM_at - -
          Human Genome U133 Set 228645_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A054175 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A054175 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 140133 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3238785 CR749453.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3238785 NR_003142.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3238785 CR749453.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3238785 NR_003142.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3238785 CR749453.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3238785 NR_003142.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3238785 CR749453.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3238785 NR_003142.2 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0