Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03299132_s1
          See other SNORD114-31 GE Assays ›
          Gene Symbol:
          SNORD114-31
          Gene Name:
          small nucleolar RNA, C/D box 114-31
          Gene Aliases:
          14q(II-31)
          Chromosome Location:
          Chr.14: 100993236 - 100993309 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03299132_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SNORD114-31
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003224.1 1 34 69

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream small nucleolar RNA,
          C/D box 114-30
          SNORD114-30 14q(II-30) -1.2kb Sense Chr. 14 - 100991919 - 100991989 on NCBI Build GRCh38
          Downstream microRNA 379 MIR379 MIRN379,
          hsa-mir-379,
          mir-379
          28.8kb Sense Chr. 14 - 101022066 - 101022132 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1400010689.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC14000711.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4142 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4141 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979878 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09517566 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979890 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09517551 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979869 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979889 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979883 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979874 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979887 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979879 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979871 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979876 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979868 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09517564 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979885 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979870 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09517560 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979889 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09517560 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979871 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09517551 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979885 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979870 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979869 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979874 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09517564 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979876 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09517566 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979878 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979883 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979868 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979890 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979887 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29979879 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517564 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517496 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517551 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517578 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517524 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517597 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517575 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517585 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517486 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517538 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517489 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517599 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517536 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16734936 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517561 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517554 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517517 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517609 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517618 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517525 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517576 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517472 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517535 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16734938 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517573 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517571 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517559 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517587 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517589 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517568 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517557 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517616 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517593 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517614 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517514 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517606 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517493 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517480 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517595 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517588 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517570 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517478 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517583 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517543 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517611 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517581 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517613 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517546 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517501 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517498 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517601 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517522 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517602 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517604 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09517505 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR14008163.hg.1 ENST00000363219.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14008163.hg.1 NR_003224.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14008163.hg.1 TC14000711.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1400010689.hg.1 ENST00000363219.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400010689.hg.1 NR_003224.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400180765.hg.1 ENST00000363219.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400180765.hg.1 NR_003224.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400180765.hg.1 TC1400010689.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1400180765.hg.1 NR_003224.1 Refseq
          Affymetrix Human Clariom D Assay PSR1400180765.hg.1 ENST00000363219.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010689.hg.1 ENST00000363219.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400010689.hg.1 NR_003224.1 Refseq
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3310451 NR_003224.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3310451 NR_003224.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3310451 NR_003224.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3310451 NR_003224.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc