Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03301915_sH
          See other IGBP1P1 GE Assays ›
          Gene Symbol:
          IGBP1P1
          Gene Name:
          immunoglobulin (CD79A) binding protein 1 pseudogene 1
          Gene Aliases:
          C14orf19
          Chromosome Location:
          Chr.14: 34939922 - 34940496 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03301915_sH
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: IGBP1P1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002937.2 1 352 102
          GenBank mRNA AY168620.1 - 1 106

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream bromodomain adjacent to zinc finger domain 1A BAZ1A ACF1,
          WALp1,
          WCRF180,
          hACF1
          -64.3kb Anti-sense Chr. 14 - 34752731 - 34875647 on NCBI Build GRCh38
          Downstream signal recognition particle 54 SRP54 42.2kb Sense Chr. 14 - 34982645 - 35029567 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1400006886.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC14000218.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09220596 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09220621 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678402 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678392 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678425 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678407 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678431 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09220619 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678433 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678383 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678442 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678389 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678386 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09220601 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678398 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678428 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678426 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678410 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678373 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678397 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678436 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678393 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16693623 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09220580 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678424 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678375 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678441 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09220563 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09220602 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09220566 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09220628 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09220590 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09220605 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678384 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16693613 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09220625 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09220561 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09220604 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678432 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09220581 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09220579 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678388 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678438 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09220603 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678372 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678387 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678371 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09220599 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09220586 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09220595 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678412 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09220627 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678429 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29678390 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3353581 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678393 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09220605 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678412 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678392 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678400 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09220580 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09220566 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678390 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09220602 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09220607 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09220596 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09220590 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678398 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678397 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678386 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678388 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678410 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678377 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678417 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678384 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678405 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678383 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16693610 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678402 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678387 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678379 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09220603 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678424 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678389 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678407 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678402 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678384 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678412 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09220566 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678398 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678405 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678397 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678417 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678410 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678424 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678387 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678392 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678393 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09220607 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09220590 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678379 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09220603 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16693610 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09220580 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678400 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09220602 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678389 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678388 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678383 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09220605 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678377 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678386 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09220596 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678390 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29678407 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220608 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220598 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220606 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220566 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220576 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220624 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220610 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220562 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220588 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220567 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220579 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220629 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220593 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220616 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220564 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220619 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220614 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220581 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220578 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220595 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220572 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220584 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220560 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220611 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220569 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220604 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220586 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220574 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220557 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16693622 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16693623 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220627 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220602 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220590 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09220621 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NR_002937_at - -
          Affymetrix Human Gene 1.0 ST Array 7973901 - -
          Affymetrix Human Gene 1.1 ST Array 7973900 - -
          Affymetrix Human Transcriptome Array 2.0 PSR14002322.hg.1 ENST00000427756.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14002322.hg.1 NR_002937.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14002322.hg.1 TC14000218.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1400147803.hg.1 ENST00000554337.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400147803.hg.1 NR_002937.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400147803.hg.1 TC1400006886.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1400147803.hg.1 NR_002937.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400147803.hg.1 ENST00000554337.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400147804.hg.1 NR_002937.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400147804.hg.1 TC1400006886.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1400147804.hg.1 NR_002937.2 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A051365 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A051365 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_2157709 AY168620.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_2157709 NR_002937.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1799563 AY168620.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1799563 NR_002937.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1799563 AY168620.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1799563 NR_002937.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1799563 AY168620.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1799563 NR_002937.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2157709 AY168620.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_2157709 NR_002937.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2157709 AY168620.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_2157709 NR_002937.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2157709 AY168620.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_2157709 NR_002937.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1799563 AY168620.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1799563 NR_002937.2 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2157709 AY168620.1 human_transcriptome
          Illumina HumanRef-8 v3.0 expression beadchip ILMN_2157709 NR_002937.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2157709 AY168620.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_2157709 NR_002937.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1799563 AY168620.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1799563 NR_002937.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1799563 AY168620.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1799563 NR_002937.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2157709 AY168620.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_2157709 NR_002937.2 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1799563 AY168620.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1799563 NR_002937.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2157709 AY168620.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_2157709 NR_002937.2 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0