Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03309488_s1
          See other SNORA5C GE Assays ›
          Gene Symbol:
          SNORA5C
          Gene Name:
          small nucleolar RNA, H/ACA box 5C
          Gene Aliases:
          ACA5c
          Chromosome Location:
          Chr.7: 45104906 - 45105042 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03309488_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SNORA5C
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002991.1 1 69 77

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream small nucleolar RNA,
          H/ACA box 5A
          SNORA5A ACA5,
          ACA5A/B/C
          -424.0 Sense Chr. 7 - 45104349 - 45104482 on NCBI Build GRCh38
          Downstream small nucleolar RNA,
          H/ACA box 5B
          SNORA5B ACA5b 926.0 Sense Chr. 7 - 45105968 - 45106099 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700010949.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07001353.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4004 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4005 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05460096 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05460099 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05460097 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05460091 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25682247 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05460091 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05460096 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05460097 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P25682247 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P05460099 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05460111 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15936666 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05460126 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05460117 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05460100 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05460112 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05460091 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05460114 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05460115 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05460109 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05460132 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05460092 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05460107 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05460130 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05460128 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05460096 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05460135 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05460105 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15936661 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05460094 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05460104 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05460099 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05460125 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05460121 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05460123 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05460133 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15936663 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15936664 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05460102 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P05460098 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NR_002991_at - -
          Affymetrix Human Gene 1.0 ST Array 8139485 - -
          Affymetrix Human Gene 1.1 ST Array 8139484 - -
          Affymetrix Human Transcriptome Array 2.0 PSR07021143.hg.1 ENST00000364902.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07021143.hg.1 NR_002991.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07021143.hg.1 TC07001353.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0700010949.hg.1 ENST00000364902.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010949.hg.1 ENST00000364902.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700010949.hg.1 NR_002991.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700010949.hg.1 NR_002991.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700178306.hg.1 ENST00000364902.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700178306.hg.1 NR_002991.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700178306.hg.1 NR_002991.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700178306.hg.1 TC0700010949.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700178306.hg.1 ENST00000364902.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010949.hg.1 ENST00000364902.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010949.hg.1 ENST00000364902.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700010949.hg.1 NR_002991.1 Refseq
          Affymetrix Human Clariom D Assay TC0700010949.hg.1 NR_002991.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700178305.hg.1 ENST00000364902.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700178305.hg.1 NR_002991.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700178305.hg.1 TC0700010949.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700178305.hg.1 ENST00000364902.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700178305.hg.1 NR_002991.1 Refseq
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3246708 NR_002991.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3246708 AM055746.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3246708 NR_002991.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3246708 AM055746.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3246708 NR_002991.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3246708 AM055746.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3246708 NR_002991.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3246708 AM055746.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0