Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03405518_m1
          See other PLA2G2C GE Assays ›
          Gene Symbol:
          PLA2G2C
          Gene Name:
          phospholipase A2 group IIC
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 20163085 - 20177364 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03405518_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PLA2G2C
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001316722.1 NP_001303651.1 3 - 4 275 65

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 Homo sapiens Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid catabolic process
          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20095395 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078151 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20095450 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078168 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078161 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078154 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078121 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078126 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078189 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20095419 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078181 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078165 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078186 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078110 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20095453 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078173 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078159 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078178 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078122 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078149 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15035206 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078115 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078131 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20095459 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15035192 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15035211 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078147 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078128 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078170 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078133 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15035198 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078162 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20095397 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078118 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15035209 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20095386 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078172 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078120 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078139 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078113 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078188 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078143 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20095460 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20095434 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078111 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078135 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078191 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20095446 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20095437 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15035194 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20095400 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078180 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20095461 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20095381 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20095468 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078160 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20095410 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078114 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20095417 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20095413 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20095469 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078140 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078125 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078167 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20095415 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078145 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20095475 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078152 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20095455 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078132 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20095458 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15035195 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078108 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20095402 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078134 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00078142 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 119081 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 536869 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 454647 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 278405 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 423364 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3344156 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1715 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_1714 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 3233 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P35051846 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P00025379 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P00025384 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3344156 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3344156 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00078190 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00078181 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15035209 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00078188 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00078172 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00078161 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20095464 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00078162 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20095468 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00078168 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00078173 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20095450 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00078159 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20095447 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15035211 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20095475 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20095437 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20095461 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00078189 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00078165 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20095469 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15035207 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20095460 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20095446 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00078178 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00078180 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20095453 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20095455 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20095464 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00078190 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15035211 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15035207 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20095461 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00078178 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20095447 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00078188 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00078165 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00078181 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20095453 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20095469 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00078180 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20095460 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00078189 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00078172 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00078173 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20095437 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00078159 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15035209 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20095475 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00078168 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20095450 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20095468 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20095446 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00078162 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00078161 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20095455 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P35051846 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00025370 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00025379 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00025368 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00025381 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078137 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078186 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078140 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078181 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078175 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15035209 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078132 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078128 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078115 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078120 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078154 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15035206 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078190 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078122 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078116 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078177 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078143 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078126 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078171 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078150 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15035192 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078149 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078172 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078208 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078166 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078159 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078184 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15035207 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078141 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15035220 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15035217 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078142 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078176 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078169 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078157 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15035198 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078130 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078205 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078173 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15035193 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078134 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078119 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078121 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078145 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078161 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078203 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078210 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078188 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078152 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078200 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078201 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078197 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078118 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078124 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078163 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078192 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078194 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078179 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00078113 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P00025370 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NM_001105572_at - -
          Affymetrix Human Gene 1.0 ST Array 7913236 - -
          Affymetrix Human Gene 1.1 ST Array 7913235 - -
          Affymetrix Human Genome U219 Array 11743249_at - -
          Affymetrix Human Genome U219 Array 11743249_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A022996 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A022996 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3237030 NM_001105572.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1659370 - -
          HumanHT-12 V4.0 expression beadchip ILMN_3237030 NM_001105572.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3237030 NM_001105572.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3237030 NM_001105572.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1659370 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1656867 NM_001105572.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1659370 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1656867 NM_001105572.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1659370 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0