Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03454032_s1
          See other OR5E1P GE Assays ›
          Gene Symbol:
          OR5E1P
          Gene Name:
          olfactory receptor family 5 subfamily E member 1 pseudogene
          Gene Aliases:
          HSTPCR24, OR5E1, TPCR24
          Chromosome Location:
          Chr.11: 7849051 - 7849571 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03454032_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: OR5E1P
          Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027711.1
          1 453 78
          GenBank mRNA AF309699.1 - LIT2079
          OR5E1P.
          1 453 78

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream olfactory receptor family 5 subfamily P member 3 OR5P3 JCG1 -23.1kb Anti-sense Chr. 11 - 7825037 - 7825972 on NCBI Build GRCh38
          Downstream uncharacterized LOC283299 LOC283299 1.2kb Anti-sense Chr. 11 - 7850751 - 7905955 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1100006768.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC11000153.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28024240 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28024243 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07625147 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16417645 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07625150 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28024234 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07625149 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28024241 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28024232 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28024242 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28024226 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07625160 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07625164 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07625161 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28024224 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07625159 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07625165 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28024228 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07625148 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28024227 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28024222 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28024239 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P07625170 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3575529 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_5195 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_5196 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 5923 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3575529 - -
          SurePrint G3 Human GE Microarray G4851A A_19_P00323558 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07625170 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07625161 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07625171 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07625149 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07625168 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07625160 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28024222 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28024241 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07625159 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28024243 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28024245 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28024239 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28024242 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07625156 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28024234 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28024224 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28024226 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07625154 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28024223 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28024248 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28024228 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28024247 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28024232 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07625148 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07625147 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07625159 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07625171 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07625168 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28024232 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28024248 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07625156 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07625149 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28024243 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28024239 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28024228 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07625170 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28024245 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07625160 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28024241 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28024222 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07625161 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28024226 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07625154 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07625147 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28024224 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28024223 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P07625148 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28024247 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28024242 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P28024234 - -
          Agilent Human 1B Microarray G4111A A_32_P917758 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16417658 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16417656 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625173 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625148 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625207 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625243 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625160 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625186 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625166 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625152 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625245 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625163 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625240 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625184 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625168 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625224 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625197 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625235 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625223 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625174 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625227 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625237 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625170 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16417650 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625239 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625154 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625161 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625216 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625206 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625247 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625204 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625199 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625157 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625158 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625177 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625201 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625192 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625219 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625230 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625182 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625189 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625156 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625171 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625178 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625180 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625195 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625225 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625233 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P07625211 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7938262 - -
          Affymetrix Human Gene 1.1 ST Array 7938261 - -
          Affymetrix HT HG-U133+ PM Array Plate 1566277_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC1100006768.hg.1 ENST00000305754.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006768.hg.1 ENST00000305754.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006768.hg.1 ENST00000305754.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100006768.hg.1 NR_027711.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11002169.hg.1 NR_027711.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR11002169.hg.1 TC11000153.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC1100006768.hg.1 ENST00000305754.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100006768.hg.1 ENST00000305754.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100006768.hg.1 ENST00000305754.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100006768.hg.1 NR_027711.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100147446.hg.1 ENST00000305754.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1100147446.hg.1 NR_027711.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100147446.hg.1 TC1100006768.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1100147446.hg.1 NR_027711.1 Refseq
          Affymetrix Human Clariom D Assay PSR1100147446.hg.1 ENST00000305754.2 Ensembl
          Human Genome U133 Plus 2.0 Array 1566277_PM_at - -
          Human Genome U133 Plus 2.0 Array 1566277_at - -
          Human Genome U133 Plus 2.0 Array 1566276_at - -
          Human Genome U133 Plus 2.0 Array 1566276_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566277_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566277_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566277_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566276_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566276_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566277_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566276_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566277_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566277_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566276_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566276_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566276_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566277_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566276_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566277_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566276_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566277_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566276_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566277_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566277_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566276_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566276_at7 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046109 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A046109 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 145459 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix