Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03464002_s1
          See other SNORD8 GE Assays ›
          Gene Symbol:
          SNORD8
          Gene Name:
          small nucleolar RNA, C/D box 8
          Gene Aliases:
          RNU6C, mgU6-53
          Chromosome Location:
          Chr.14: 21397293 - 21397401 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03464002_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SNORD8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_002916.2 1 68 106
          GenBank mRNA AJ243222.1 - 1 67 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1400008633.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC14000919.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_3922 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09158472 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615252 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615274 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09158428 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615264 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615261 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615286 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615295 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615237 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615243 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615234 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09158468 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615272 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615249 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09158475 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615251 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09158457 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615256 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09158448 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615276 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615279 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09158441 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615268 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615271 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615280 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615253 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615284 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615240 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615291 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09158454 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09158433 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09158436 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615245 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615242 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615277 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09158424 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615235 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615285 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09158474 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615293 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615293 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615280 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615240 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615285 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615251 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615261 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615279 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09158448 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615237 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09158472 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09158424 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615291 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615271 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615245 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615256 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09158441 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09158474 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615274 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615286 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615284 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615276 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615277 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615295 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09158457 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615272 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615242 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09158454 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09158433 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615235 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615264 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09158428 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615243 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615268 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615252 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615249 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615234 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09158436 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P29615253 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09158475 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09158468 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158431 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158466 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158430 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16684026 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158452 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158456 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158399 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158472 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158453 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158488 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158422 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158461 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16684031 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158426 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158480 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158476 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158447 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158481 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158396 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158470 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158420 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158485 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158483 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158402 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158474 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158450 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158443 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158459 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158478 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158437 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158440 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158395 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158455 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158390 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158424 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158465 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158393 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16684025 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158435 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P09158432 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P16684046 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NR_002916_at - -
          Affymetrix Human Gene 1.0 ST Array 7977733 - -
          Affymetrix Human Gene 1.1 ST Array 7977732 - -
          Affymetrix HT HG-U133+ PM Array Plate 1566340_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1566339_PM_at - -
          Affymetrix Human Genome U219 Array 11763909_x_at - -
          Affymetrix Human Genome U219 Array 11763909_PM_x_at - -
          Affymetrix Human Genome U219 Array 11763908_at - -
          Affymetrix Human Genome U219 Array 11763908_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR14010044.hg.1 ENST00000363915.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR14010044.hg.1 NR_002916.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR14010044.hg.1 TC14000919.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1400008633.hg.1 ENST00000363915.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1400008633.hg.1 NR_002916.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400162414.hg.1 ENST00000363915.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400162414.hg.1 NR_002916.2 Refseq
          Affymetrix Human Clariom D Assay PSR1400162414.hg.1 TC1400008633.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1400162414.hg.1 ENST00000363915.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1400162414.hg.1 NR_002916.2 Refseq
          Affymetrix Human Clariom D Assay TC1400008633.hg.1 ENST00000363915.1 Ensembl
          Affymetrix Human Clariom D Assay TC1400008633.hg.1 NR_002916.2 Refseq
          Human Genome U133 Plus 2.0 Array 1566339_at - -
          Human Genome U133 Plus 2.0 Array 1566340_PM_at - -
          Human Genome U133 Plus 2.0 Array 1566340_at - -
          Human Genome U133 Plus 2.0 Array 1566339_PM_at - -
          Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) NR_002916_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566340_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566339_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566339_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566340_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566340_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566340_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566340_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566339_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566339_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566339_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566339_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566340_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566339_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566339_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566340_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566339_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566340_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566339_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566340_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566340_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566339_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566340_at9 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3248874 NR_002916.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3248874 NR_002916.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3248874 NR_002916.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3248874 NR_002916.2 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0