Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000361092.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000361092.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000361092.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000361092.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000361092.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000361092.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000361092.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000361092.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000361092.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000361092.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000361092.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000376808.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000376808.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000376808.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000376808.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000376808.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000376808.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000376808.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000376808.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000376808.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000376808.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000376811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000376811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000376811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000376811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000376811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000376811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000376811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000376811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000376811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000376811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000376811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000376811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000376811.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000466137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000466137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000466137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000466137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000482537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000482537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000482537.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000490321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000490321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000490321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000490321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000494066.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000494066.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000494066.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000494066.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000494066.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000494066.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000494066.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000494066.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000494066.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000494066.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000494066.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000494066.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
ENST00000494066.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NM_001127603.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NM_001127603.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NM_001127603.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NM_001127603.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NM_001127603.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NM_001127603.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NM_001127603.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NM_001127603.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NM_001127603.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NM_001127603.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NM_017881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NM_017881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NM_017881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NM_017881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NM_017881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NM_017881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NM_017881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NM_017881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NM_017881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NM_017881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NM_017881.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NR_023352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NR_023352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NR_023352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NR_023352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NR_023352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NR_023352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NR_023352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NR_023352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NR_023352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NR_023352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NR_023352.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010445.hg.1 |
NR_023352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000361092.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000361092.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000361092.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000361092.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000361092.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000361092.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000361092.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000361092.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000361092.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000361092.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000361092.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000376808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000376808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000376808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000376808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000376808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000376808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000376808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000376808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000376808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000376808.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000376811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000376811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000376811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000376811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000376811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000376811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000376811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000376811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000376811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000376811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000376811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000376811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000376811.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000466137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000466137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000466137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000466137.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000482537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000482537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000482537.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000490321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000490321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000490321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000490321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000494066.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000494066.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000494066.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000494066.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000494066.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000494066.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000494066.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000494066.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000494066.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000494066.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000494066.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000494066.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
ENST00000494066.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NM_001127603.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NM_001127603.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NM_001127603.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NM_001127603.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NM_001127603.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NM_001127603.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NM_001127603.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NM_001127603.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NM_001127603.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NM_001127603.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NM_017881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NM_017881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NM_017881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NM_017881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NM_017881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NM_017881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NM_017881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NM_017881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NM_017881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NM_017881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NM_017881.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NR_023352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NR_023352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NR_023352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NR_023352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NR_023352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NR_023352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NR_023352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NR_023352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NR_023352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NR_023352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NR_023352.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010445.hg.1 |
NR_023352.1 |
Refseq |