Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03654441_s1
          See other RNA28S5 GE Assays ›
          Gene Symbol:
          RNA28S5 RNA45S5
          Gene Name:
          RNA, 28S ribosomal 5
          RNA, 45S pre-ribosomal 5
          Gene Aliases:
          RN28S1
          RN45S
          Chromosome Location:
          Chr.Un|NT_167214: 113348 - 118417 on Build GRCh38
          Chromosome Location:
          Chr.Un|NT_167214: 105424 - 118780 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs03654441_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RNA28S5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003287.2 1 1268 98
          Gene Symbol: RNA45S5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046235.1 1 9192 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2100006470.hg.1, TC2100006488.hg.1

          Set Membership:

          Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3244165 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3420259 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3346552 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3336632 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4163 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4164 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 54255 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 57065 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 60699 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 40777 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3244165 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3346552 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3420259 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3336632 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3336632 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3420259 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3244165 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3346552 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array NR_003287_at - -
          Affymetrix Human Gene 1.0 ST Array 7917646 - -
          Affymetrix Human Gene 1.0 ST Array 8169351 - -
          Affymetrix Human Gene 1.0 ST Array 7942876 - -
          Affymetrix Human Gene 1.0 ST Array 7967621 - -
          Affymetrix Human Gene 1.0 ST Array 8151235 - -
          Affymetrix Human Gene 1.0 ST Array 7942792 - -
          Affymetrix Human Gene 1.0 ST Array 8059577 - -
          Affymetrix Human Gene 1.1 ST Array 7942791 - -
          Affymetrix Human Gene 1.1 ST Array 7917645 - -
          Affymetrix Human Gene 1.1 ST Array 7942875 - -
          Affymetrix Human Gene 1.1 ST Array 7967620 - -
          Affymetrix Human Gene 1.1 ST Array 8169350 - -
          Affymetrix Human Gene 1.1 ST Array 8151234 - -
          Affymetrix Human Gene 1.1 ST Array 8059576 - -
          Affymetrix Human Transcriptome Array 2.0 TC22_KI270733v1_random00006435.hg.1 ENST00000610594.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC22_KI270733v1_random00006435.hg.1 ENST00000611782.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC22_KI270733v1_random00006435.hg.1 ENST00000611802.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC22_KI270733v1_random00006435.hg.1 ENST00000611919.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC22_KI270733v1_random00006435.hg.1 ENST00000611934.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC22_KI270733v1_random00006435.hg.1 ENST00000616292.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006470.hg.1 ENST00000611393.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006470.hg.1 ENST00000617236.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006470.hg.1 NR_003287.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006470.hg.1 NR_003287.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006470.hg.1 NR_003287.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006470.hg.1 NR_003287.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006470.hg.1 NR_003287.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006470.hg.1 NR_003287.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006470.hg.1 NR_003287.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TCUn_GL000220v100006442.hg.1 ENST00000612047.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TCUn_GL000220v100006432.hg.1 ENST00000611431.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TCUn_GL000220v100006432.hg.1 ENST00000611446.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TCUn_GL000220v100006432.hg.1 ENST00000614470.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TCUn_GL000220v100006432.hg.1 ENST00000614928.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TCUn_GL000220v100006432.hg.1 ENST00000615115.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TCUn_GL000220v100006432.hg.1 ENST00000617883.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TCUn_GL000220v100006432.hg.1 ENST00000619779.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC22_KI270733v1_random00006443.hg.1 ENST00000612700.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006488.hg.1 ENST00000612173.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006488.hg.1 ENST00000613635.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006488.hg.1 ENST00000614525.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006488.hg.1 ENST00000616418.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006488.hg.1 ENST00000616457.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006488.hg.1 ENST00000619471.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100144281.hg.1 NR_003287.4 Refseq
          Affymetrix Human Clariom D Assay PSR2100144281.hg.1 TC2100006470.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100144281.hg.1 NR_003287.4 Refseq
          Affymetrix Human Clariom D Assay PSR2100144284.hg.1 NR_003287.4 Refseq
          Affymetrix Human Clariom D Assay PSR2100144284.hg.1 TC2100006470.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100144284.hg.1 NR_003287.4 Refseq
          Affymetrix Human Clariom D Assay PSR2100144328.hg.1 NR_046235.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100144328.hg.1 TC2100006488.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100144328.hg.1 NR_046235.3 Refseq
          Affymetrix Human Clariom D Assay TCUn_GL000220v100006442.hg.1 ENST00000612047.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100144282.hg.1 ENST00000611393.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100144282.hg.1 NR_003287.4 Refseq
          Affymetrix Human Clariom D Assay PSR2100144282.hg.1 TC2100006470.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100144282.hg.1 ENST00000611393.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100144282.hg.1 NR_003287.4 Refseq
          Affymetrix Human Clariom D Assay PSR2100144332.hg.1 NR_046235.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100144332.hg.1 TC2100006488.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100144332.hg.1 NR_046235.3 Refseq
          Affymetrix Human Clariom D Assay TCUn_GL000220v100006432.hg.1 ENST00000611431.1 Ensembl
          Affymetrix Human Clariom D Assay TCUn_GL000220v100006432.hg.1 ENST00000611446.1 Ensembl
          Affymetrix Human Clariom D Assay TCUn_GL000220v100006432.hg.1 ENST00000614470.1 Ensembl
          Affymetrix Human Clariom D Assay TCUn_GL000220v100006432.hg.1 ENST00000614928.1 Ensembl
          Affymetrix Human Clariom D Assay TCUn_GL000220v100006432.hg.1 ENST00000615115.1 Ensembl
          Affymetrix Human Clariom D Assay TCUn_GL000220v100006432.hg.1 ENST00000617883.1 Ensembl
          Affymetrix Human Clariom D Assay TCUn_GL000220v100006432.hg.1 ENST00000619779.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100144283.hg.1 NR_003287.4 Refseq
          Affymetrix Human Clariom D Assay PSR2100144283.hg.1 TC2100006470.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100144283.hg.1 NR_003287.4 Refseq
          Affymetrix Human Clariom D Assay PSR2100144329.hg.1 ENST00000616457.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100144329.hg.1 NR_046235.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100144329.hg.1 TC2100006488.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100144329.hg.1 ENST00000616457.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100144329.hg.1 NR_046235.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100144331.hg.1 ENST00000612173.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100144331.hg.1 NR_046235.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100144331.hg.1 TC2100006488.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100144331.hg.1 ENST00000612173.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100144331.hg.1 NR_046235.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006470.hg.1 ENST00000611393.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006470.hg.1 ENST00000617236.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006470.hg.1 NR_003287.4 Refseq
          Affymetrix Human Clariom D Assay TC2100006470.hg.1 NR_003287.4 Refseq
          Affymetrix Human Clariom D Assay TC2100006470.hg.1 NR_003287.4 Refseq
          Affymetrix Human Clariom D Assay TC2100006470.hg.1 NR_003287.4 Refseq
          Affymetrix Human Clariom D Assay TC2100006470.hg.1 NR_003287.4 Refseq
          Affymetrix Human Clariom D Assay TC2100006470.hg.1 NR_003287.4 Refseq
          Affymetrix Human Clariom D Assay TC2100006470.hg.1 NR_003287.4 Refseq
          Affymetrix Human Clariom D Assay TC2100006488.hg.1 ENST00000612173.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006488.hg.1 ENST00000613635.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006488.hg.1 ENST00000614525.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006488.hg.1 ENST00000616418.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006488.hg.1 ENST00000616457.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006488.hg.1 ENST00000619471.1 Ensembl
          Affymetrix Human Clariom D Assay TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Clariom D Assay TC2100006488.hg.1 NR_046235.3 Refseq
          Affymetrix Human Clariom D Assay TC22_KI270733v1_random00006435.hg.1 ENST00000610594.1 Ensembl
          Affymetrix Human Clariom D Assay TC22_KI270733v1_random00006435.hg.1 ENST00000611782.1 Ensembl
          Affymetrix Human Clariom D Assay TC22_KI270733v1_random00006435.hg.1 ENST00000611802.1 Ensembl
          Affymetrix Human Clariom D Assay TC22_KI270733v1_random00006435.hg.1 ENST00000611919.1 Ensembl
          Affymetrix Human Clariom D Assay TC22_KI270733v1_random00006435.hg.1 ENST00000611934.1 Ensembl
          Affymetrix Human Clariom D Assay TC22_KI270733v1_random00006435.hg.1 ENST00000616292.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100144285.hg.1 ENST00000617236.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100144285.hg.1 NR_003287.4 Refseq
          Affymetrix Human Clariom D Assay PSR2100144285.hg.1 TC2100006470.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100144285.hg.1 ENST00000617236.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100144285.hg.1 NR_003287.4 Refseq
          Affymetrix Human Clariom D Assay PSR2100144333.hg.1 NR_046235.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100144333.hg.1 TC2100006488.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100144333.hg.1 NR_046235.3 Refseq
          Affymetrix Human Clariom D Assay TC22_KI270733v1_random00006443.hg.1 ENST00000612700.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2100144330.hg.1 NR_046235.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100144330.hg.1 TC2100006488.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100144330.hg.1 NR_046235.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100144280.hg.1 NR_003287.4 Refseq
          Affymetrix Human Clariom D Assay PSR2100144280.hg.1 TC2100006470.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100144280.hg.1 NR_003287.4 Refseq
          Affymetrix Human Clariom D Assay PSR2100144286.hg.1 NR_003287.4 Refseq
          Affymetrix Human Clariom D Assay PSR2100144286.hg.1 TC2100006470.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100144286.hg.1 NR_003287.4 Refseq
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3251587 NR_003287.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3251587 AK057879.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1733559 NR_003287.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1733559 AF023158.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3251587 NR_003287.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3251587 AK057879.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1733559 NR_003287.2 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1733559 AF023158.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1733559 NR_003287.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_1733559 AF023158.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3251587 NR_003287.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3251587 AK057879.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1733559 NR_003287.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_1733559 AF023158.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3251587 NR_003287.2 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3251587 AK057879.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1733559 NR_003287.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1733559 AF023158.1 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1733559 NR_003287.2 human_transcriptome
          Illumina humanRef-8 v2.0 expression beadchip ILMN_1733559 AF023158.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1733559 NR_003287.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1733559 AF023158.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1733559 NR_003287.2 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1733559 AF023158.1 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1733559 NR_003287.2 human_transcriptome
          Illumina HumanRef-8 WG-DASL v3.0 ILMN_1733559 AF023158.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0