Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03654883_s1
          See other LOC100130761 GE Assays ›
          Gene Symbol:
          LOC100130761
          Gene Name:
          uncharacterized LOC100130761
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03654883_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100130761
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK095511.1 - 1 2741 102

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream T-box,
          brain 1
          TBR1 TBR-1,
          TES-56
          -8.8kb Sense Chr. 2 - 161416094 - 161425867 on NCBI Build GRCh38
          Downstream AT-hook containing transcription factor 1 pseudogene 1 AHCTF1P1 AHCTF1P,
          ELYS-like
          61.6kb Anti-sense Chr. 2 - 161499067 - 161507902 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747510 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747572 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747554 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747578 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747530 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01623342 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747531 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747523 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01623386 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747540 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01623393 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747576 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747556 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747536 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747505 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01623399 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747515 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747567 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01623344 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01623351 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747564 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747580 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747522 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747507 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747526 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747581 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01623383 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01623402 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01623367 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747551 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01623385 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747539 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747541 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747546 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747506 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747555 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747509 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01623331 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747553 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747545 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01623355 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747517 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747579 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01623403 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747549 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747529 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01623394 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747532 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747525 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747583 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747512 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747516 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747550 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747519 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747535 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01623362 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747552 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747562 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747565 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01623364 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747577 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747570 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747547 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747569 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747513 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747544 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21747568 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 107162 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 368629 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3283897 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 48059 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P00513051 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3283897 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P00513051 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P30748145 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00513051 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P00513051 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0200009817.hg.1 ENST00000624969.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009817.hg.1 ENST00000624969.1 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0