Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs03654900_s1
          See other LOC100131053 GE Assays ›
          Gene Symbol:
          LOC100131053
          Gene Name:
          uncharacterized LOC100131053
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs03654900_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100131053
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK095564.1 - 1 1850 90
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DB312536.1 - 1 Chr. 1 - 19333972 - 19334460 [-] on Build GRCh38 90
          GenBankmRNA H25413.1 - 1 Chr. 1 - 19334091 - 19334452 [-] on Build GRCh38 90
          GenBankmRNA AA927130.1 - 1 Chr. 1 - 19334169 - 19334469 [-] on Build GRCh38 90
          GenBankmRNA DB315267.1 - 1 Chr. 1 - 19333936 - 19334444 [-] on Build GRCh38 90
          GenBankmRNA BG057784.1 - 1 Chr. 1 - 19334152 - 19334481 [-] on Build GRCh38 90
          GenBankmRNA BU684388.1 - 1 Chr. 1 - 19333142 - 19334463 [-] on Build GRCh38 90
          GenBankmRNA AA404325.1 - 1 Chr. 1 - 19333971 - 19334463 [-] on Build GRCh38 90
          GenBankmRNA AA812831.1 - 1 Chr. 1 - 19334057 - 19334469 [-] on Build GRCh38 90

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream PQ loop repeat containing 2 PQLC2 Sense Chr. 1 - 19312246 - 19332611 on NCBI Build GRCh38
          Downstream capping actin protein of muscle Z-line beta subunit CAPZB CAPB,
          CAPPB,
          CAPZ
          4.3kb Anti-sense Chr. 1 - 19338773 - 19485641 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human CGH Microarray 4x180K 96868 - -
          Agilent SurePrint G3 Human CGH Microarray 8x60K 353 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 836389 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3209167 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P912228 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 18582 - -
          Agilent Human Genome CGH Microarray 44B A_14_P107721 - -
          Agilent Human Genome CGH Microarray 44A A_14_P107721 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 14139 - -
          Agilent Human Genome CGH + SNP Microarray A_14_P107721 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3209167 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P912228 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_14_P107721 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_14_P107721 - -
          Agilent Human Genome CGH Microarray 4x44K A_14_P107721 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P912228 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 110959 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 77302 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00024331 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_14_P107721 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_14_P107721 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 239546_PM_at - -
          Human Genome U133 Plus 2.0 Array 239546_at - -
          Human Genome U133 Plus 2.0 Array 239546_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239546_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239546_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239546_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239546_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239546_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239546_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239546_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239546_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239546_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239546_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 239546_at3 - -
          Human Genome U95 Set 76538_at - -
          Human Genome U95 Set 76538_PM_at - -
          Human Genome U133 Set 239546_at - -
          Human Genome U133 Set 239546_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0